Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6922019-6922292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO2
Entrez Gene ID
2026
Gene Name
enolase 2
Gene Aliases
HEL-S-279, NSE
UniGene
Hs.511915
Gene Chromosome Location
Chr.12: 6914450-6923696 on Build GRCh38
Transcripts
NM_001975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4044557 c.1145C>T p.A382V Chr.12: 6922133-6922133 on Build GRCh38
4958619 c.1096G>A p.G366R Chr.12: 6922084-6922084 on Build GRCh38
4044558 c.1161G>A p.G387G Chr.12: 6922149-6922149 on Build GRCh38
4147429 c.1134C>A p.D378E Chr.12: 6922122-6922122 on Build GRCh38
6021049 c.1099G>C p.V367L Chr.12: 6922087-6922087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781845923 Chr.12: 6922047-6922047 on Build GRCh38
rs376310491 Chr.12: 6922193-6922193 on Build GRCh38
rs979580672 Chr.12: 6922205-6922205 on Build GRCh38
rs782469877 Chr.12: 6922048-6922048 on Build GRCh38
rs781924277 Chr.12: 6922146-6922146 on Build GRCh38
rs782552065 Chr.12: 6922066-6922066 on Build GRCh38
rs782608998 Chr.12: 6922088-6922088 on Build GRCh38
rs782209242 Chr.12: 6922098-6922098 on Build GRCh38
rs190347711 Chr.12: 6922143-6922143 on Build GRCh38
rs924141275 Chr.12: 6922221-6922221 on Build GRCh38
rs782820907 Chr.12: 6922185-6922185 on Build GRCh38
rs782434657 Chr.12: 6922078-6922078 on Build GRCh38
rs782730997 Chr.12: 6922173-6922173 on Build GRCh38
rs782678932 Chr.12: 6922068-6922068 on Build GRCh38
rs11550252 Chr.12: 6922083-6922083 on Build GRCh38
rs782693220 Chr.12: 6922210-6922210 on Build GRCh38
rs782003553 Chr.12: 6922175-6922175 on Build GRCh38
rs782286397 Chr.12: 6922127-6922127 on Build GRCh38
rs770346242 Chr.12: 6922196-6922197 on Build GRCh38
rs782104497 Chr.12: 6922147-6922147 on Build GRCh38
rs181064177 Chr.12: 6922263-6922263 on Build GRCh38
rs782117672 Chr.12: 6922181-6922181 on Build GRCh38
rs781862516 Chr.12: 6922057-6922057 on Build GRCh38
rs782772138 Chr.12: 6922053-6922053 on Build GRCh38
rs782471113 Chr.12: 6922209-6922209 on Build GRCh38
rs782205190 Chr.12: 6922076-6922076 on Build GRCh38

View Full Product Details