Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44703821-44704078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM16
Entrez Gene ID
388551
Gene Name
carcinoembryonic antigen related cell adhesion molecule 16
Gene Aliases
CEAL2, DFNA4B
UniGene
Hs.456381
Gene Chromosome Location
Chr.19: 44699151-44710714 on Build GRCh38
Transcripts
NM_001039213.3 XM_017026795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5731274 c.616G>A p.A206T Chr.19: 44704056-44704056 on Build GRCh38
3707210 c.578-7C>T p.? Chr.19: 44704011-44704011 on Build GRCh38
4671314 c.631C>T p.R211* Chr.19: 44704071-44704071 on Build GRCh38
3535715 c.604G>C p.A202P Chr.19: 44704044-44704044 on Build GRCh38
5580599 c.596C>A p.T199K Chr.19: 44704036-44704036 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762882576 Chr.19: 44703962-44703962 on Build GRCh38
rs387907149 Chr.19: 44704053-44704053 on Build GRCh38
rs527797823 Chr.19: 44704049-44704049 on Build GRCh38
rs192922176 Chr.19: 44703902-44703902 on Build GRCh38
rs1036098369 Chr.19: 44703865-44703865 on Build GRCh38
rs777437787 Chr.19: 44704005-44704005 on Build GRCh38
rs765825648 Chr.19: 44703991-44703991 on Build GRCh38
rs371499861 Chr.19: 44704035-44704035 on Build GRCh38
rs761152852 Chr.19: 44703983-44703983 on Build GRCh38
rs767224799 Chr.19: 44703976-44703976 on Build GRCh38
rs775017974 Chr.19: 44704056-44704056 on Build GRCh38
rs565350614 Chr.19: 44704023-44704023 on Build GRCh38
rs888791396 Chr.19: 44703918-44703918 on Build GRCh38
rs1007523407 Chr.19: 44703928-44703928 on Build GRCh38
rs1018545602 Chr.19: 44703959-44703959 on Build GRCh38
rs762421371 Chr.19: 44703990-44703990 on Build GRCh38
rs764390124 Chr.19: 44703984-44703984 on Build GRCh38
rs755184535 Chr.19: 44704002-44704002 on Build GRCh38
rs764381305 Chr.19: 44704026-44704026 on Build GRCh38
rs531964496 Chr.19: 44703996-44703996 on Build GRCh38
rs1051907463 Chr.19: 44703903-44703903 on Build GRCh38
rs897607071 Chr.19: 44703876-44703876 on Build GRCh38
rs374156875 Chr.19: 44703972-44703972 on Build GRCh38
rs368338468 Chr.19: 44703997-44703997 on Build GRCh38
rs1031181464 Chr.19: 44703950-44703950 on Build GRCh38
rs754163867 Chr.19: 44703986-44703986 on Build GRCh38
rs774975434 Chr.19: 44703978-44703978 on Build GRCh38
rs751750847 Chr.19: 44704001-44704001 on Build GRCh38
rs201855875 Chr.19: 44704013-44704013 on Build GRCh38
rs977761111 Chr.19: 44703933-44703933 on Build GRCh38
rs879833225 Chr.19: 44703988-44703988 on Build GRCh38
rs957777348 Chr.19: 44703951-44703951 on Build GRCh38

View Full Product Details