Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9754534-9754759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD69
Entrez Gene ID
969
Gene Name
CD69 molecule
Gene Aliases
AIM, BL-AC/P26, CLEC2C, EA1, GP32/28, MLR-3
UniGene
Hs.208854
Gene Chromosome Location
Chr.12: 9752486-9760901 on Build GRCh38
Transcripts
NM_001781.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
944897 c.478G>T p.E160* Chr.12: 9754600-9754600 on Build GRCh38
432039 c.400C>T p.R134* Chr.12: 9754678-9754678 on Build GRCh38
5842095 c.482T>G p.F161C Chr.12: 9754596-9754596 on Build GRCh38
2047237 c.460A>G p.K154E Chr.12: 9754618-9754618 on Build GRCh38
5842096 c.445C>T p.P149S Chr.12: 9754633-9754633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762487537 Chr.12: 9754636-9754636 on Build GRCh38
rs201675276 Chr.12: 9754676-9754676 on Build GRCh38
rs748250928 Chr.12: 9754706-9754706 on Build GRCh38
rs774956184 Chr.12: 9754629-9754629 on Build GRCh38
rs540178434 Chr.12: 9754622-9754622 on Build GRCh38
rs144700824 Chr.12: 9754627-9754627 on Build GRCh38
rs774397998 Chr.12: 9754588-9754588 on Build GRCh38
rs770769958 Chr.12: 9754621-9754621 on Build GRCh38
rs372400857 Chr.12: 9754673-9754673 on Build GRCh38
rs767215920 Chr.12: 9754665-9754665 on Build GRCh38
rs376663052 Chr.12: 9754578-9754578 on Build GRCh38
rs199731208 Chr.12: 9754632-9754632 on Build GRCh38
rs771393996 Chr.12: 9754730-9754730 on Build GRCh38
rs772672364 Chr.12: 9754568-9754577 on Build GRCh38
rs749269880 Chr.12: 9754678-9754678 on Build GRCh38
rs763688311 Chr.12: 9754639-9754639 on Build GRCh38
rs867305691 Chr.12: 9754651-9754651 on Build GRCh38
rs755061113 Chr.12: 9754693-9754693 on Build GRCh38
rs966323258 Chr.12: 9754644-9754644 on Build GRCh38
rs772404503 Chr.12: 9754711-9754711 on Build GRCh38
rs1029242014 Chr.12: 9754714-9754714 on Build GRCh38
rs748425489 Chr.12: 9754605-9754605 on Build GRCh38
rs774103338 Chr.12: 9754606-9754606 on Build GRCh38
rs756942837 Chr.12: 9754645-9754645 on Build GRCh38
rs1004860841 Chr.12: 9754623-9754623 on Build GRCh38
rs113155724 Chr.12: 9754591-9754591 on Build GRCh38
rs142870511 Chr.12: 9754672-9754672 on Build GRCh38
rs747421625 Chr.12: 9754729-9754729 on Build GRCh38
rs773433527 Chr.12: 9754720-9754720 on Build GRCh38
rs779064267 Chr.12: 9754704-9754704 on Build GRCh38
rs756191117 Chr.12: 9754576-9754576 on Build GRCh38
rs749431406 Chr.12: 9754583-9754583 on Build GRCh38
rs751119124 Chr.12: 9754641-9754641 on Build GRCh38
rs768970736 Chr.12: 9754587-9754587 on Build GRCh38
rs745729153 Chr.12: 9754567-9754567 on Build GRCh38

View Full Product Details