Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7064046-7064271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRPEL1
Entrez Gene ID
80273
Gene Name
GrpE like 1, mitochondrial
Gene Aliases
GrpE, HMGE, mt-GrpE#1
UniGene
Hs.443723
Gene Chromosome Location
Chr.4: 7058895-7068210 on Build GRCh38
Transcripts
NM_025196.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1057017 c.132G>T p.M44I Chr.4: 7064154-7064154 on Build GRCh38
2829932 c.198G>T p.K66N Chr.4: 7064088-7064088 on Build GRCh38
6296255 c.90C>A p.A30A Chr.4: 7064196-7064196 on Build GRCh38
6259020 c.129C>A p.D43E Chr.4: 7064157-7064157 on Build GRCh38
1430760 c.92C>T p.T31M Chr.4: 7064194-7064194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767288952 Chr.4: 7064156-7064156 on Build GRCh38
rs752297045 Chr.4: 7064123-7064123 on Build GRCh38
rs774609129 Chr.4: 7064088-7064088 on Build GRCh38
rs749540530 Chr.4: 7064065-7064065 on Build GRCh38
rs374845101 Chr.4: 7064105-7064105 on Build GRCh38
rs758688203 Chr.4: 7064244-7064244 on Build GRCh38
rs756000781 Chr.4: 7064173-7064173 on Build GRCh38
rs772346740 Chr.4: 7064193-7064193 on Build GRCh38
rs914220754 Chr.4: 7064148-7064148 on Build GRCh38
rs753276386 Chr.4: 7064212-7064212 on Build GRCh38
rs751222701 Chr.4: 7064128-7064128 on Build GRCh38
rs113063443 Chr.4: 7064232-7064232 on Build GRCh38
rs777267396 Chr.4: 7064207-7064207 on Build GRCh38
rs962342453 Chr.4: 7064230-7064230 on Build GRCh38
rs772479973 Chr.4: 7064100-7064100 on Build GRCh38
rs146360027 Chr.4: 7064213-7064213 on Build GRCh38
rs761215457 Chr.4: 7064120-7064120 on Build GRCh38
rs901602998 Chr.4: 7064165-7064165 on Build GRCh38
rs750347151 Chr.4: 7064171-7064171 on Build GRCh38
rs975881524 Chr.4: 7064068-7064068 on Build GRCh38
rs747347955 Chr.4: 7064194-7064194 on Build GRCh38
rs201079395 Chr.4: 7064081-7064081 on Build GRCh38
rs1016553712 Chr.4: 7064222-7064222 on Build GRCh38
rs189680001 Chr.4: 7064241-7064241 on Build GRCh38
rs945766651 Chr.4: 7064153-7064153 on Build GRCh38
rs142193388 Chr.4: 7064124-7064124 on Build GRCh38
rs762140695 Chr.4: 7064092-7064092 on Build GRCh38
rs761475422 Chr.4: 7064215-7064215 on Build GRCh38
rs769307748 Chr.4: 7064240-7064240 on Build GRCh38
rs773596752 Chr.4: 7064110-7064110 on Build GRCh38
rs771616178 Chr.4: 7064202-7064202 on Build GRCh38
rs200577196 Chr.4: 7064155-7064155 on Build GRCh38
rs757011343 Chr.4: 7064154-7064154 on Build GRCh38
rs927168309 Chr.4: 7064091-7064091 on Build GRCh38
rs576819542 Chr.4: 7064238-7064238 on Build GRCh38
rs569245761 Chr.4: 7064180-7064180 on Build GRCh38
rs140117073 Chr.4: 7064176-7064176 on Build GRCh38
rs1028420499 Chr.4: 7064099-7064099 on Build GRCh38
rs755877944 Chr.4: 7064228-7064228 on Build GRCh38
rs1050565985 Chr.4: 7064179-7064179 on Build GRCh38
rs764871556 Chr.4: 7064122-7064122 on Build GRCh38
rs767242721 Chr.4: 7064227-7064227 on Build GRCh38
rs779932846 Chr.4: 7064175-7064175 on Build GRCh38
rs746858574 Chr.4: 7064242-7064242 on Build GRCh38
rs191407428 Chr.4: 7064209-7064209 on Build GRCh38

View Full Product Details