Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27531133-27531404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIBADH
Entrez Gene ID
11112
Gene Name
3-hydroxyisobutyrate dehydrogenase
Gene Aliases
NS5ATP1
UniGene
Hs.406758
Gene Chromosome Location
Chr.7: 27525440-27663001 on Build GRCh38
Transcripts
NM_152740.3 XM_017011719.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5509133 c.721G>T p.A241S Chr.7: 27531323-27531323 on Build GRCh38
334770 c.789G>T p.V263V Chr.7: 27531255-27531255 on Build GRCh38
4361824 c.799G>A p.V267I Chr.7: 27531245-27531245 on Build GRCh38
1088899 c.808G>A p.A270T Chr.7: 27531236-27531236 on Build GRCh38
5358498 c.711A>G p.P237P Chr.7: 27531333-27531333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970616917 Chr.7: 27531296-27531296 on Build GRCh38
rs151261943 Chr.7: 27531154-27531154 on Build GRCh38
rs140422741 Chr.7: 27531323-27531323 on Build GRCh38
rs754420248 Chr.7: 27531200-27531200 on Build GRCh38
rs748892207 Chr.7: 27531345-27531346 on Build GRCh38
rs779715913 Chr.7: 27531225-27531226 on Build GRCh38
rs1039826470 Chr.7: 27531266-27531266 on Build GRCh38
rs942960750 Chr.7: 27531258-27531259 on Build GRCh38
rs367901321 Chr.7: 27531295-27531295 on Build GRCh38
rs755006413 Chr.7: 27531300-27531300 on Build GRCh38
rs145731776 Chr.7: 27531246-27531246 on Build GRCh38
rs370669214 Chr.7: 27531377-27531377 on Build GRCh38
rs779809483 Chr.7: 27531189-27531189 on Build GRCh38
rs371636646 Chr.7: 27531328-27531328 on Build GRCh38
rs112104797 Chr.7: 27531175-27531175 on Build GRCh38
rs777580186 Chr.7: 27531229-27531229 on Build GRCh38
rs141047822 Chr.7: 27531238-27531238 on Build GRCh38
rs35454595 Chr.7: 27531196-27531196 on Build GRCh38
rs147879630 Chr.7: 27531287-27531287 on Build GRCh38
rs375072886 Chr.7: 27531351-27531357 on Build GRCh38
rs778718460 Chr.7: 27531304-27531304 on Build GRCh38
rs3735571 Chr.7: 27531273-27531273 on Build GRCh38
rs375108307 Chr.7: 27531361-27531361 on Build GRCh38
rs755232994 Chr.7: 27531351-27531351 on Build GRCh38
rs1032507621 Chr.7: 27531254-27531254 on Build GRCh38
rs761861649 Chr.7: 27531231-27531231 on Build GRCh38
rs529685275 Chr.7: 27531248-27531248 on Build GRCh38
rs767672213 Chr.7: 27531286-27531286 on Build GRCh38
rs752194399 Chr.7: 27531320-27531320 on Build GRCh38
rs147714836 Chr.7: 27531237-27531237 on Build GRCh38
rs770032420 Chr.7: 27531336-27531336 on Build GRCh38
rs762899269 Chr.7: 27531162-27531162 on Build GRCh38
rs199794378 Chr.7: 27531168-27531168 on Build GRCh38
rs775828480 Chr.7: 27531245-27531245 on Build GRCh38
rs140501381 Chr.7: 27531225-27531225 on Build GRCh38
rs750153535 Chr.7: 27531185-27531185 on Build GRCh38
rs780525690 Chr.7: 27531348-27531348 on Build GRCh38
rs764273166 Chr.7: 27531167-27531167 on Build GRCh38
rs755920763 Chr.7: 27531188-27531188 on Build GRCh38
rs753848362 Chr.7: 27531192-27531192 on Build GRCh38
rs73287942 Chr.7: 27531160-27531160 on Build GRCh38
rs760350852 Chr.7: 27531291-27531291 on Build GRCh38
rs767296159 Chr.7: 27531183-27531183 on Build GRCh38
rs747796694 Chr.7: 27531205-27531205 on Build GRCh38
rs770225736 Chr.7: 27531239-27531239 on Build GRCh38
rs898631087 Chr.7: 27531163-27531163 on Build GRCh38
rs146545289 Chr.7: 27531372-27531372 on Build GRCh38
rs746699632 Chr.7: 27531332-27531332 on Build GRCh38
rs774853912 Chr.7: 27531381-27531381 on Build GRCh38
rs923951250 Chr.7: 27531166-27531166 on Build GRCh38

View Full Product Details