Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 129900830-129901377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM10
Entrez Gene ID
56980
Gene Name
PR domain 10
Gene Aliases
PFM7
UniGene
Hs.275086
Gene Chromosome Location
Chr.11: 129899706-130003097 on Build GRCh38
Transcripts
NM_020228.2 NM_199437.1 NM_199438.1 NM_199439.1 XM_011542912.2 XM_017018023.1

SNP Detail

SNP ID Chromosome Location
rs145568613 Chr.11: 129901213-129901213 on Build GRCh38
rs1004161396 Chr.11: 129901075-129901075 on Build GRCh38
rs192024075 Chr.11: 129901116-129901116 on Build GRCh38
rs386758837 Chr.11: 129901212-129901213 on Build GRCh38
rs531523428 Chr.11: 129901113-129901113 on Build GRCh38
rs562397036 Chr.11: 129900999-129900999 on Build GRCh38
rs1023791135 Chr.11: 129900895-129900895 on Build GRCh38
rs371252997 Chr.11: 129901150-129901152 on Build GRCh38
rs888567437 Chr.11: 129901074-129901074 on Build GRCh38
rs914327390 Chr.11: 129900887-129900887 on Build GRCh38
rs943716863 Chr.11: 129901344-129901344 on Build GRCh38
rs137866324 Chr.11: 129901235-129901235 on Build GRCh38
rs955955356 Chr.11: 129901015-129901015 on Build GRCh38
rs968487717 Chr.11: 129901215-129901215 on Build GRCh38
rs565109306 Chr.11: 129900892-129900892 on Build GRCh38
rs991203878 Chr.11: 129900885-129900885 on Build GRCh38
rs896764227 Chr.11: 129900877-129900877 on Build GRCh38
rs1056365941 Chr.11: 129900867-129900867 on Build GRCh38
rs938178059 Chr.11: 129900878-129900878 on Build GRCh38
rs907970612 Chr.11: 129901284-129901284 on Build GRCh38
rs1032771383 Chr.11: 129901002-129901003 on Build GRCh38
rs1026985321 Chr.11: 129901027-129901027 on Build GRCh38
rs1617778 Chr.11: 129901212-129901212 on Build GRCh38
rs577000705 Chr.11: 129901128-129901128 on Build GRCh38
rs1041380265 Chr.11: 129900952-129900952 on Build GRCh38
rs1001404361 Chr.11: 129901000-129901000 on Build GRCh38
rs1015624557 Chr.11: 129901090-129901090 on Build GRCh38
rs751084445 Chr.11: 129900868-129900869 on Build GRCh38
rs971368148 Chr.11: 129901110-129901110 on Build GRCh38
rs146755037 Chr.11: 129900874-129900874 on Build GRCh38
rs532714728 Chr.11: 129901290-129901290 on Build GRCh38
rs374839534 Chr.11: 129900983-129900983 on Build GRCh38
rs547176019 Chr.11: 129901214-129901214 on Build GRCh38
rs1041417190 Chr.11: 129900925-129900925 on Build GRCh38
rs35751590 Chr.11: 129901221-129901222 on Build GRCh38
rs959489729 Chr.11: 129901150-129901150 on Build GRCh38
rs899637329 Chr.11: 129900979-129900979 on Build GRCh38
rs576624426 Chr.11: 129900942-129900942 on Build GRCh38
rs760253995 Chr.11: 129900861-129900861 on Build GRCh38
rs545633256 Chr.11: 129900944-129900944 on Build GRCh38
rs943994914 Chr.11: 129900947-129900947 on Build GRCh38
rs878911317 Chr.11: 129901120-129901120 on Build GRCh38
rs996642096 Chr.11: 129900992-129900992 on Build GRCh38
rs947033190 Chr.11: 129900921-129900921 on Build GRCh38

View Full Product Details