Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 129914601-129915150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM10
Entrez Gene ID
56980
Gene Name
PR domain 10
Gene Aliases
PFM7
UniGene
Hs.275086
Gene Chromosome Location
Chr.11: 129899706-130003097 on Build GRCh38
Transcripts
NM_020228.2 NM_199437.1 NM_199438.1 NM_199439.1 XM_011542912.2 XM_017018023.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
171638 c.2548C>T p.R850* Chr.11: 129914997-129914997 on Build GRCh38
6067922 c.2761C>A p.Q921K Chr.11: 129914784-129914784 on Build GRCh38
6754913 c.2691A>G p.Q897Q Chr.11: 129914854-129914854 on Build GRCh38
687270 c.2830C>T p.Q944* Chr.11: 129914715-129914715 on Build GRCh38
924944 c.2783C>T p.S928L Chr.11: 129914762-129914762 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751518878 Chr.11: 129914676-129914676 on Build GRCh38
rs115260710 Chr.11: 129914666-129914666 on Build GRCh38
rs370949794 Chr.11: 129914713-129914713 on Build GRCh38
rs780510554 Chr.11: 129914851-129914852 on Build GRCh38
rs375447334 Chr.11: 129914891-129914891 on Build GRCh38
rs183483249 Chr.11: 129914699-129914699 on Build GRCh38
rs145298028 Chr.11: 129914842-129914842 on Build GRCh38
rs781166079 Chr.11: 129914830-129914830 on Build GRCh38
rs769670890 Chr.11: 129914854-129914854 on Build GRCh38
rs773373139 Chr.11: 129914638-129914638 on Build GRCh38
rs370360512 Chr.11: 129914895-129914895 on Build GRCh38
rs111630660 Chr.11: 129914750-129914750 on Build GRCh38
rs745561964 Chr.11: 129914802-129914802 on Build GRCh38
rs780703897 Chr.11: 129915055-129915055 on Build GRCh38
rs775876571 Chr.11: 129914873-129914873 on Build GRCh38
rs757686056 Chr.11: 129914787-129914787 on Build GRCh38
rs746880596 Chr.11: 129914813-129914813 on Build GRCh38
rs561725124 Chr.11: 129914768-129914768 on Build GRCh38
rs779552249 Chr.11: 129914810-129914810 on Build GRCh38
rs765995026 Chr.11: 129914997-129914997 on Build GRCh38
rs201852274 Chr.11: 129915022-129915022 on Build GRCh38
rs73016844 Chr.11: 129915071-129915071 on Build GRCh38
rs746970319 Chr.11: 129914704-129914704 on Build GRCh38
rs777640644 Chr.11: 129914934-129914934 on Build GRCh38
rs375699368 Chr.11: 129914857-129914857 on Build GRCh38
rs373946342 Chr.11: 129914697-129914697 on Build GRCh38
rs201058090 Chr.11: 129914860-129914860 on Build GRCh38
rs904090758 Chr.11: 129914756-129914756 on Build GRCh38
rs892622338 Chr.11: 129915108-129915108 on Build GRCh38
rs367829574 Chr.11: 129914893-129914893 on Build GRCh38
rs918412104 Chr.11: 129914890-129914890 on Build GRCh38
rs199649951 Chr.11: 129914970-129914970 on Build GRCh38
rs751587746 Chr.11: 129914655-129914655 on Build GRCh38
rs75558641 Chr.11: 129914761-129914761 on Build GRCh38
rs375197801 Chr.11: 129914955-129914955 on Build GRCh38
rs368035644 Chr.11: 129914915-129914915 on Build GRCh38
rs1013583605 Chr.11: 129914762-129914762 on Build GRCh38
rs376264867 Chr.11: 129914684-129914684 on Build GRCh38
rs774682934 Chr.11: 129914759-129914759 on Build GRCh38
rs942192305 Chr.11: 129914956-129914956 on Build GRCh38
rs775900388 Chr.11: 129914942-129914942 on Build GRCh38
rs776413906 Chr.11: 129914719-129914719 on Build GRCh38
rs750585931 Chr.11: 129914758-129914758 on Build GRCh38
rs763285097 Chr.11: 129914986-129914986 on Build GRCh38
rs541579876 Chr.11: 129915121-129915122 on Build GRCh38
rs187939540 Chr.11: 129914944-129914944 on Build GRCh38
rs760976287 Chr.11: 129915046-129915046 on Build GRCh38
rs1029453562 Chr.11: 129914812-129914812 on Build GRCh38
rs923013617 Chr.11: 129914651-129914651 on Build GRCh38
rs768574831 Chr.11: 129914825-129914825 on Build GRCh38
rs955004876 Chr.11: 129914824-129914824 on Build GRCh38
rs1002257356 Chr.11: 129915105-129915105 on Build GRCh38
rs931705877 Chr.11: 129914937-129914937 on Build GRCh38
rs753210538 Chr.11: 129914770-129914770 on Build GRCh38
rs756704362 Chr.11: 129914774-129914774 on Build GRCh38
rs752522111 Chr.11: 129915070-129915070 on Build GRCh38
rs778395388 Chr.11: 129914792-129914792 on Build GRCh38
rs1038580217 Chr.11: 129914694-129914694 on Build GRCh38
rs997716293 Chr.11: 129915109-129915109 on Build GRCh38
rs192670392 Chr.11: 129915086-129915086 on Build GRCh38
rs371170571 Chr.11: 129914863-129914863 on Build GRCh38
rs746448744 Chr.11: 129914836-129914836 on Build GRCh38
rs764489350 Chr.11: 129914778-129914778 on Build GRCh38
rs763907491 Chr.11: 129914634-129914634 on Build GRCh38
rs371498835 Chr.11: 129914931-129914931 on Build GRCh38
rs374123981 Chr.11: 129914900-129914900 on Build GRCh38
rs754238642 Chr.11: 129914786-129914786 on Build GRCh38
rs775718691 Chr.11: 129914866-129914866 on Build GRCh38
rs144412505 Chr.11: 129914941-129914941 on Build GRCh38
rs748894919 Chr.11: 129914687-129914687 on Build GRCh38
rs764673235 Chr.11: 129914882-129914882 on Build GRCh38
rs1036567267 Chr.11: 129914985-129914985 on Build GRCh38
rs750795035 Chr.11: 129914977-129914977 on Build GRCh38
rs754434690 Chr.11: 129914885-129914885 on Build GRCh38
rs763174361 Chr.11: 129914640-129914640 on Build GRCh38
rs754532021 Chr.11: 129914925-129914925 on Build GRCh38
rs201951007 Chr.11: 129914705-129914705 on Build GRCh38
rs147190509 Chr.11: 129914809-129914809 on Build GRCh38
rs929858864 Chr.11: 129914898-129914898 on Build GRCh38
rs867970026 Chr.11: 129914777-129914777 on Build GRCh38
rs766414155 Chr.11: 129914648-129914648 on Build GRCh38
rs768886734 Chr.11: 129914959-129914959 on Build GRCh38
rs1052744586 Chr.11: 129915093-129915093 on Build GRCh38
rs549722947 Chr.11: 129914940-129914940 on Build GRCh38
rs767669456 Chr.11: 129914767-129914767 on Build GRCh38
rs756510238 Chr.11: 129914674-129914674 on Build GRCh38
rs754521037 Chr.11: 129914669-129914669 on Build GRCh38
rs370489624 Chr.11: 129914708-129914708 on Build GRCh38
rs376705432 Chr.11: 129915050-129915050 on Build GRCh38
rs368250108 Chr.11: 129914969-129914969 on Build GRCh38
rs760426995 Chr.11: 129914629-129914629 on Build GRCh38
rs753298257 Chr.11: 129914670-129914670 on Build GRCh38
rs369106793 Chr.11: 129914696-129914696 on Build GRCh38
rs1045272951 Chr.11: 129914751-129914751 on Build GRCh38
rs755145654 Chr.11: 129914664-129914664 on Build GRCh38
rs761665298 Chr.11: 129914734-129914734 on Build GRCh38
rs758074195 Chr.11: 129914914-129914914 on Build GRCh38
rs565673192 Chr.11: 129915077-129915077 on Build GRCh38
rs117750366 Chr.11: 129914869-129914869 on Build GRCh38
rs765451854 Chr.11: 129914975-129914975 on Build GRCh38
rs781078066 Chr.11: 129914926-129914926 on Build GRCh38
rs748149601 Chr.11: 129914927-129914927 on Build GRCh38

View Full Product Details