Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27312439-27312900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK38L
Entrez Gene ID
23012
Gene Name
serine/threonine kinase 38 like
Gene Aliases
NDR2
UniGene
Hs.184523
Gene Chromosome Location
Chr.12: 27244143-27325959 on Build GRCh38
Transcripts
NM_015000.3 XM_005253342.3 XM_006719058.3 XM_006719059.3 XM_011520613.2 XM_017019036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
21105 c.438T>C p.D146D Chr.12: 27312593-27312593 on Build GRCh38
938389 c.487C>A p.L163I Chr.12: 27312642-27312642 on Build GRCh38
3459805 c.406C>T p.R136* Chr.12: 27312561-27312561 on Build GRCh38
4546287 c.396G>T p.V132V Chr.12: 27312551-27312551 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs138365234 Chr.12: 27312669-27312669 on Build GRCh38
rs113318303 Chr.12: 27312634-27312634 on Build GRCh38
rs778752373 Chr.12: 27312670-27312670 on Build GRCh38
rs775413598 Chr.12: 27312532-27312532 on Build GRCh38
rs957415973 Chr.12: 27312766-27312766 on Build GRCh38
rs377425691 Chr.12: 27312722-27312722 on Build GRCh38
rs755586232 Chr.12: 27312593-27312593 on Build GRCh38
rs757083298 Chr.12: 27312562-27312562 on Build GRCh38
rs773900950 Chr.12: 27312508-27312508 on Build GRCh38
rs184586101 Chr.12: 27312538-27312538 on Build GRCh38
rs748924373 Chr.12: 27312617-27312617 on Build GRCh38
rs765059568 Chr.12: 27312620-27312620 on Build GRCh38
rs769323446 Chr.12: 27312510-27312511 on Build GRCh38
rs576095733 Chr.12: 27312815-27312815 on Build GRCh38
rs751847420 Chr.12: 27312804-27312804 on Build GRCh38
rs565839422 Chr.12: 27312777-27312777 on Build GRCh38
rs759688667 Chr.12: 27312542-27312542 on Build GRCh38
rs987119059 Chr.12: 27312853-27312853 on Build GRCh38
rs554121560 Chr.12: 27312773-27312773 on Build GRCh38
rs779445140 Chr.12: 27312610-27312610 on Build GRCh38
rs1024350437 Chr.12: 27312558-27312558 on Build GRCh38
rs776842753 Chr.12: 27312539-27312539 on Build GRCh38
rs894737758 Chr.12: 27312469-27312469 on Build GRCh38
rs1041370466 Chr.12: 27312574-27312574 on Build GRCh38
rs766437985 Chr.12: 27312685-27312687 on Build GRCh38
rs767429248 Chr.12: 27312581-27312581 on Build GRCh38
rs890473732 Chr.12: 27312468-27312468 on Build GRCh38
rs971378267 Chr.12: 27312638-27312638 on Build GRCh38
rs751385924 Chr.12: 27312544-27312544 on Build GRCh38
rs754620405 Chr.12: 27312628-27312628 on Build GRCh38
rs989995156 Chr.12: 27312767-27312767 on Build GRCh38
rs750427518 Chr.12: 27312590-27312590 on Build GRCh38
rs1031739852 Chr.12: 27312748-27312748 on Build GRCh38
rs888185627 Chr.12: 27312742-27312742 on Build GRCh38
rs189555638 Chr.12: 27312865-27312865 on Build GRCh38
rs554607721 Chr.12: 27312791-27312791 on Build GRCh38
rs536059649 Chr.12: 27312782-27312782 on Build GRCh38
rs1020496022 Chr.12: 27312822-27312822 on Build GRCh38
rs573397338 Chr.12: 27312730-27312730 on Build GRCh38
rs766433520 Chr.12: 27312526-27312526 on Build GRCh38
rs762904824 Chr.12: 27312523-27312523 on Build GRCh38
rs772634226 Chr.12: 27312522-27312522 on Build GRCh38
rs896953858 Chr.12: 27312630-27312630 on Build GRCh38
rs760820253 Chr.12: 27312524-27312524 on Build GRCh38
rs200136083 Chr.12: 27312543-27312543 on Build GRCh38
rs772679618 Chr.12: 27312507-27312507 on Build GRCh38

View Full Product Details