Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51590926-51591428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN8A
Entrez Gene ID
6334
Gene Name
sodium voltage-gated channel alpha subunit 8
Gene Aliases
CERIII, CIAT, EIEE13, MED, NaCh6, Nav1.6, PN4
UniGene
Hs.436550
Gene Chromosome Location
Chr.12: 51589959-51812865 on Build GRCh38
Transcripts
NM_001177984.2 NM_014191.3 XM_006719556.3 XM_011538650.2 XM_011538651.2 XM_017019794.1 XM_017019795.1 XM_017019796.1

SNP Detail

SNP ID Chromosome Location
rs886049579 Chr.12: 51591312-51591312 on Build GRCh38
rs1033829069 Chr.12: 51591305-51591305 on Build GRCh38
rs1021067316 Chr.12: 51591335-51591335 on Build GRCh38
rs925195746 Chr.12: 51591150-51591150 on Build GRCh38
rs1028334121 Chr.12: 51590964-51590964 on Build GRCh38
rs966694569 Chr.12: 51591339-51591339 on Build GRCh38
rs967126960 Chr.12: 51591123-51591124 on Build GRCh38
rs1002493155 Chr.12: 51591287-51591287 on Build GRCh38
rs917251322 Chr.12: 51591127-51591170 on Build GRCh38
rs886049578 Chr.12: 51591257-51591257 on Build GRCh38
rs566535586 Chr.12: 51591324-51591324 on Build GRCh38
rs888034727 Chr.12: 51591286-51591286 on Build GRCh38
rs1049276460 Chr.12: 51591273-51591273 on Build GRCh38
rs866798180 Chr.12: 51591259-51591259 on Build GRCh38
rs1013647099 Chr.12: 51591314-51591314 on Build GRCh38
rs992268305 Chr.12: 51591075-51591075 on Build GRCh38
rs985255489 Chr.12: 51591195-51591195 on Build GRCh38
rs931182250 Chr.12: 51591134-51591134 on Build GRCh38
rs374729451 Chr.12: 51591347-51591347 on Build GRCh38
rs1043168573 Chr.12: 51591376-51591376 on Build GRCh38
rs934858767 Chr.12: 51591408-51591408 on Build GRCh38
rs932470768 Chr.12: 51591170-51591170 on Build GRCh38
rs918515992 Chr.12: 51591129-51591129 on Build GRCh38
rs927092513 Chr.12: 51591145-51591145 on Build GRCh38
rs1033730587 Chr.12: 51591047-51591047 on Build GRCh38
rs915945238 Chr.12: 51591354-51591354 on Build GRCh38
rs1002786946 Chr.12: 51590968-51590968 on Build GRCh38
rs887926558 Chr.12: 51590967-51590967 on Build GRCh38
rs552823069 Chr.12: 51591293-51591293 on Build GRCh38
rs1032376116 Chr.12: 51591353-51591353 on Build GRCh38
rs930992822 Chr.12: 51591257-51591257 on Build GRCh38
rs912437977 Chr.12: 51591219-51591220 on Build GRCh38
rs960778414 Chr.12: 51591068-51591068 on Build GRCh38
rs937145939 Chr.12: 51591226-51591226 on Build GRCh38
rs943773417 Chr.12: 51591220-51591220 on Build GRCh38
rs1001279580 Chr.12: 51591350-51591350 on Build GRCh38
rs534924909 Chr.12: 51591371-51591371 on Build GRCh38
rs1036771869 Chr.12: 51591229-51591229 on Build GRCh38
rs1056927957 Chr.12: 51591312-51591312 on Build GRCh38
rs992792503 Chr.12: 51591351-51591351 on Build GRCh38
rs886049580 Chr.12: 51591319-51591319 on Build GRCh38
rs568745199 Chr.12: 51591018-51591019 on Build GRCh38
rs912384249 Chr.12: 51591387-51591387 on Build GRCh38
rs992809334 Chr.12: 51591082-51591097 on Build GRCh38
rs983987118 Chr.12: 51591138-51591138 on Build GRCh38
rs926099486 Chr.12: 51591379-51591379 on Build GRCh38
rs948807327 Chr.12: 51591251-51591252 on Build GRCh38

View Full Product Details