Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53276542-53277046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESPL1
Entrez Gene ID
9700
Gene Name
extra spindle pole bodies like 1, separase
Gene Aliases
ESP1, SEPA
UniGene
Hs.153479
Gene Chromosome Location
Chr.12: 53268257-53293643 on Build GRCh38
Transcripts
NM_012291.4 XM_006719705.3 XM_011539024.2 XM_011539025.2 XM_017020253.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6389564 c.1706T>C p.L569P Chr.12: 53276625-53276625 on Build GRCh38
4403126 c.1934C>T p.T645I Chr.12: 53276853-53276853 on Build GRCh38
4516280 c.1749_1750CC>TT p.(=) Chr.12: 53276668-53276669 on Build GRCh38
6656344 c.1930C>T p.Q644* Chr.12: 53276849-53276849 on Build GRCh38
468571 c.1753C>T p.L585L Chr.12: 53276672-53276672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758968472 Chr.12: 53276771-53276771 on Build GRCh38
rs776254146 Chr.12: 53276590-53276590 on Build GRCh38
rs368065388 Chr.12: 53276898-53276898 on Build GRCh38
rs555983912 Chr.12: 53276710-53276710 on Build GRCh38
rs781254044 Chr.12: 53276791-53276791 on Build GRCh38
rs760378660 Chr.12: 53276881-53276881 on Build GRCh38
rs763335435 Chr.12: 53276770-53276770 on Build GRCh38
rs974331508 Chr.12: 53276985-53276985 on Build GRCh38
rs113972522 Chr.12: 53276927-53276927 on Build GRCh38
rs770745247 Chr.12: 53276882-53276882 on Build GRCh38
rs762256992 Chr.12: 53276909-53276909 on Build GRCh38
rs756350037 Chr.12: 53276806-53276806 on Build GRCh38
rs201587677 Chr.12: 53276581-53276581 on Build GRCh38
rs147987096 Chr.12: 53276701-53276701 on Build GRCh38
rs769705862 Chr.12: 53276614-53276614 on Build GRCh38
rs367676774 Chr.12: 53276897-53276897 on Build GRCh38
rs370317688 Chr.12: 53276875-53276875 on Build GRCh38
rs767240091 Chr.12: 53276665-53276665 on Build GRCh38
rs765713040 Chr.12: 53276689-53276689 on Build GRCh38
rs934122165 Chr.12: 53276994-53276994 on Build GRCh38
rs975693063 Chr.12: 53276914-53276914 on Build GRCh38
rs925967215 Chr.12: 53276695-53276695 on Build GRCh38
rs373872335 Chr.12: 53276889-53276889 on Build GRCh38
rs1318648 Chr.12: 53276761-53276761 on Build GRCh38
rs759357321 Chr.12: 53276895-53276896 on Build GRCh38
rs773148878 Chr.12: 53276961-53276961 on Build GRCh38
rs775293865 Chr.12: 53276735-53276735 on Build GRCh38
rs748052663 Chr.12: 53276844-53276844 on Build GRCh38
rs545594149 Chr.12: 53276780-53276780 on Build GRCh38
rs113474898 Chr.12: 53277009-53277009 on Build GRCh38
rs574428519 Chr.12: 53276743-53276743 on Build GRCh38
rs866722873 Chr.12: 53276747-53276747 on Build GRCh38
rs982534165 Chr.12: 53276942-53276942 on Build GRCh38
rs200043098 Chr.12: 53276667-53276667 on Build GRCh38
rs922641228 Chr.12: 53276993-53276993 on Build GRCh38
rs867325327 Chr.12: 53276658-53276658 on Build GRCh38
rs762631048 Chr.12: 53276918-53276918 on Build GRCh38
rs1014520644 Chr.12: 53276645-53276646 on Build GRCh38
rs149272357 Chr.12: 53276652-53276652 on Build GRCh38
rs1049879834 Chr.12: 53277007-53277007 on Build GRCh38
rs199988679 Chr.12: 53276845-53276845 on Build GRCh38
rs557990780 Chr.12: 53276836-53276836 on Build GRCh38
rs143192971 Chr.12: 53276706-53276706 on Build GRCh38
rs147544896 Chr.12: 53276711-53276711 on Build GRCh38
rs200635387 Chr.12: 53276649-53276649 on Build GRCh38
rs773703245 Chr.12: 53276744-53276744 on Build GRCh38
rs956211053 Chr.12: 53276970-53276970 on Build GRCh38
rs753191986 Chr.12: 53276700-53276700 on Build GRCh38
rs201291279 Chr.12: 53276907-53276907 on Build GRCh38
rs140179562 Chr.12: 53276731-53276731 on Build GRCh38
rs778970591 Chr.12: 53276837-53276837 on Build GRCh38
rs755455888 Chr.12: 53276668-53276668 on Build GRCh38
rs779803929 Chr.12: 53276809-53276809 on Build GRCh38
rs868583594 Chr.12: 53276762-53276762 on Build GRCh38
rs144474323 Chr.12: 53276653-53276653 on Build GRCh38
rs570883809 Chr.12: 53276765-53276765 on Build GRCh38
rs61760168 Chr.12: 53276779-53276779 on Build GRCh38
rs747148602 Chr.12: 53276705-53276705 on Build GRCh38
rs1023514288 Chr.12: 53276899-53276899 on Build GRCh38
rs749176917 Chr.12: 53276741-53276741 on Build GRCh38
rs775348881 Chr.12: 53276635-53276635 on Build GRCh38
rs771546244 Chr.12: 53276870-53276870 on Build GRCh38
rs777689637 Chr.12: 53276702-53276702 on Build GRCh38
rs750602674 Chr.12: 53276800-53276800 on Build GRCh38
rs749148102 Chr.12: 53276823-53276823 on Build GRCh38
rs777915986 Chr.12: 53276577-53276577 on Build GRCh38
rs916467919 Chr.12: 53276591-53276591 on Build GRCh38
rs745726284 Chr.12: 53276606-53276606 on Build GRCh38
rs908186354 Chr.12: 53276958-53276958 on Build GRCh38
rs761453624 Chr.12: 53276753-53276753 on Build GRCh38
rs759919863 Chr.12: 53276755-53276755 on Build GRCh38
rs764217873 Chr.12: 53276982-53276982 on Build GRCh38
rs546423065 Chr.12: 53276954-53276954 on Build GRCh38
rs763429092 Chr.12: 53276895-53276895 on Build GRCh38
rs761349808 Chr.12: 53276659-53276659 on Build GRCh38
rs772945895 Chr.12: 53276758-53276758 on Build GRCh38
rs563274512 Chr.12: 53276763-53276763 on Build GRCh38

View Full Product Details