Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57724201-57724697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP2
Entrez Gene ID
116986
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 2
Gene Aliases
CENTG1, GGAP2, PIKE
UniGene
Hs.302435
Gene Chromosome Location
Chr.12: 57723765-57742201 on Build GRCh38
Transcripts
NM_001122772.2 NM_014770.3 XM_017018772.1

SNP Detail

SNP ID Chromosome Location
rs2269720 Chr.12: 57724465-57724465 on Build GRCh38
rs946381019 Chr.12: 57724654-57724654 on Build GRCh38
rs982376733 Chr.12: 57724536-57724536 on Build GRCh38
rs772949083 Chr.12: 57724229-57724229 on Build GRCh38
rs759346066 Chr.12: 57724602-57724602 on Build GRCh38
rs970691444 Chr.12: 57724438-57724438 on Build GRCh38
rs77903952 Chr.12: 57724275-57724275 on Build GRCh38
rs183415660 Chr.12: 57724570-57724570 on Build GRCh38
rs553760444 Chr.12: 57724292-57724296 on Build GRCh38
rs952717440 Chr.12: 57724477-57724477 on Build GRCh38
rs776508087 Chr.12: 57724466-57724466 on Build GRCh38
rs77566780 Chr.12: 57724380-57724380 on Build GRCh38
rs970343185 Chr.12: 57724641-57724641 on Build GRCh38
rs901452126 Chr.12: 57724378-57724378 on Build GRCh38
rs554546501 Chr.12: 57724537-57724537 on Build GRCh38
rs191258052 Chr.12: 57724400-57724400 on Build GRCh38
rs999197865 Chr.12: 57724319-57724319 on Build GRCh38
rs560276737 Chr.12: 57724299-57724299 on Build GRCh38
rs893462975 Chr.12: 57724246-57724246 on Build GRCh38
rs560993926 Chr.12: 57724343-57724343 on Build GRCh38
rs1043407458 Chr.12: 57724546-57724546 on Build GRCh38
rs888841399 Chr.12: 57724281-57724281 on Build GRCh38
rs1052557647 Chr.12: 57724243-57724243 on Build GRCh38
rs956246717 Chr.12: 57724372-57724372 on Build GRCh38
rs983436336 Chr.12: 57724668-57724668 on Build GRCh38
rs530550220 Chr.12: 57724647-57724647 on Build GRCh38
rs75960044 Chr.12: 57724519-57724519 on Build GRCh38
rs952057720 Chr.12: 57724667-57724667 on Build GRCh38
rs1014393685 Chr.12: 57724632-57724632 on Build GRCh38
rs1031804436 Chr.12: 57724330-57724330 on Build GRCh38
rs1024864057 Chr.12: 57724633-57724633 on Build GRCh38
rs531218242 Chr.12: 57724388-57724388 on Build GRCh38
rs988070863 Chr.12: 57724555-57724555 on Build GRCh38
rs999144528 Chr.12: 57724566-57724566 on Build GRCh38
rs960425938 Chr.12: 57724600-57724600 on Build GRCh38
rs867930074 Chr.12: 57724489-57724489 on Build GRCh38
rs536424933 Chr.12: 57724587-57724587 on Build GRCh38
rs969420876 Chr.12: 57724528-57724528 on Build GRCh38
rs920850917 Chr.12: 57724666-57724666 on Build GRCh38
rs980138385 Chr.12: 57724457-57724457 on Build GRCh38
rs764433482 Chr.12: 57724544-57724544 on Build GRCh38
rs1007083669 Chr.12: 57724282-57724282 on Build GRCh38
rs903491650 Chr.12: 57724564-57724564 on Build GRCh38
rs1019764126 Chr.12: 57724297-57724297 on Build GRCh38
rs915120262 Chr.12: 57724649-57724649 on Build GRCh38
rs368821325 Chr.12: 57724554-57724554 on Build GRCh38
rs907949080 Chr.12: 57724514-57724514 on Build GRCh38
rs900436125 Chr.12: 57724530-57724531 on Build GRCh38
rs550580367 Chr.12: 57724658-57724658 on Build GRCh38
rs1030742972 Chr.12: 57724579-57724579 on Build GRCh38
rs187072180 Chr.12: 57724326-57724326 on Build GRCh38
rs139048084 Chr.12: 57724225-57724225 on Build GRCh38
rs532416578 Chr.12: 57724486-57724486 on Build GRCh38
rs150054599 Chr.12: 57724247-57724247 on Build GRCh38
rs992949462 Chr.12: 57724385-57724385 on Build GRCh38
rs1023688185 Chr.12: 57724520-57724520 on Build GRCh38

View Full Product Details