Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 71132586-71133036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN8
Entrez Gene ID
7103
Gene Name
tetraspanin 8
Gene Aliases
CO-029, TM4SF3
UniGene
Hs.170563
Gene Chromosome Location
Chr.12: 71125093-71157999 on Build GRCh38
Transcripts
NM_004616.2 XM_006719583.3 XM_017019913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
416080 c.574G>C p.E192Q Chr.12: 71132695-71132695 on Build GRCh38
3355867 c.468T>A p.N156K Chr.12: 71132801-71132801 on Build GRCh38
4416509 c.456C>T p.C152C Chr.12: 71132813-71132813 on Build GRCh38
6810671 c.543C>T p.C181C Chr.12: 71132726-71132726 on Build GRCh38
3464966 c.485G>A p.G162E Chr.12: 71132784-71132784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111590488 Chr.12: 71132845-71132845 on Build GRCh38
rs754568907 Chr.12: 71132654-71132654 on Build GRCh38
rs951367172 Chr.12: 71132920-71132920 on Build GRCh38
rs373629291 Chr.12: 71132844-71132844 on Build GRCh38
rs777730838 Chr.12: 71132679-71132679 on Build GRCh38
rs147276837 Chr.12: 71132704-71132704 on Build GRCh38
rs3816942 Chr.12: 71132929-71132929 on Build GRCh38
rs747620614 Chr.12: 71132659-71132659 on Build GRCh38
rs773591960 Chr.12: 71132862-71132862 on Build GRCh38
rs757239809 Chr.12: 71132873-71132873 on Build GRCh38
rs778971183 Chr.12: 71132876-71132876 on Build GRCh38
rs962988663 Chr.12: 71132626-71132626 on Build GRCh38
rs1007596874 Chr.12: 71132786-71132786 on Build GRCh38
rs187378419 Chr.12: 71132733-71132733 on Build GRCh38
rs2270588 Chr.12: 71132634-71132634 on Build GRCh38
rs764084767 Chr.12: 71132794-71132794 on Build GRCh38
rs752377778 Chr.12: 71132867-71132867 on Build GRCh38
rs370472870 Chr.12: 71132771-71132771 on Build GRCh38
rs375451226 Chr.12: 71132864-71132864 on Build GRCh38
rs760787994 Chr.12: 71132723-71132723 on Build GRCh38
rs767112082 Chr.12: 71132785-71132785 on Build GRCh38
rs925516747 Chr.12: 71132790-71132790 on Build GRCh38
rs760185143 Chr.12: 71132868-71132868 on Build GRCh38
rs774229763 Chr.12: 71132695-71132695 on Build GRCh38
rs1048927106 Chr.12: 71132979-71132979 on Build GRCh38
rs868432453 Chr.12: 71132926-71132926 on Build GRCh38
rs762144605 Chr.12: 71132732-71132732 on Build GRCh38
rs749879513 Chr.12: 71132801-71132801 on Build GRCh38
rs926659432 Chr.12: 71132919-71132919 on Build GRCh38
rs969445459 Chr.12: 71133005-71133005 on Build GRCh38
rs878877779 Chr.12: 71132832-71132832 on Build GRCh38
rs777739682 Chr.12: 71132838-71132838 on Build GRCh38
rs1029893432 Chr.12: 71132846-71132846 on Build GRCh38
rs2270587 Chr.12: 71132813-71132813 on Build GRCh38
rs778881284 Chr.12: 71132823-71132823 on Build GRCh38
rs539173427 Chr.12: 71132937-71132937 on Build GRCh38
rs769084520 Chr.12: 71132729-71132729 on Build GRCh38
rs753825850 Chr.12: 71132812-71132812 on Build GRCh38
rs879098249 Chr.12: 71132691-71132691 on Build GRCh38
rs780689350 Chr.12: 71132658-71132658 on Build GRCh38
rs898381030 Chr.12: 71132895-71132895 on Build GRCh38
rs968163884 Chr.12: 71132880-71132880 on Build GRCh38
rs1045112179 Chr.12: 71132636-71132636 on Build GRCh38
rs1048832652 Chr.12: 71132782-71132782 on Build GRCh38
rs749102050 Chr.12: 71132688-71132688 on Build GRCh38
rs751607880 Chr.12: 71132886-71132890 on Build GRCh38
rs868757282 Chr.12: 71132789-71132789 on Build GRCh38
rs182077813 Chr.12: 71132967-71132967 on Build GRCh38
rs777192427 Chr.12: 71132730-71132730 on Build GRCh38
rs140845131 Chr.12: 71132768-71132768 on Build GRCh38
rs766635069 Chr.12: 71132687-71132688 on Build GRCh38
rs1397566 Chr.12: 71132897-71132897 on Build GRCh38
rs779537041 Chr.12: 71132819-71132819 on Build GRCh38
rs370498795 Chr.12: 71132854-71132854 on Build GRCh38
rs777626110 Chr.12: 71132811-71132811 on Build GRCh38
rs869025229 Chr.12: 71132741-71132741 on Build GRCh38
rs763662189 Chr.12: 71132870-71132870 on Build GRCh38
rs943145456 Chr.12: 71132994-71132994 on Build GRCh38
rs775733045 Chr.12: 71132721-71132721 on Build GRCh38
rs759732327 Chr.12: 71132618-71132618 on Build GRCh38
rs926494496 Chr.12: 71132653-71132653 on Build GRCh38
rs534378530 Chr.12: 71132949-71132949 on Build GRCh38
rs750722789 Chr.12: 71132752-71132752 on Build GRCh38
rs1025622306 Chr.12: 71132908-71132908 on Build GRCh38
rs550197908 Chr.12: 71132802-71132802 on Build GRCh38
rs897682341 Chr.12: 71132712-71132712 on Build GRCh38
rs770249468 Chr.12: 71132858-71132858 on Build GRCh38
rs878959174 Chr.12: 71132830-71132830 on Build GRCh38
rs772379057 Chr.12: 71132714-71132714 on Build GRCh38
rs751785765 Chr.12: 71132712-71132712 on Build GRCh38
rs756050814 Chr.12: 71132662-71132662 on Build GRCh38
rs879097073 Chr.12: 71132692-71132692 on Build GRCh38
rs992060202 Chr.12: 71132931-71132931 on Build GRCh38
rs887488293 Chr.12: 71132987-71132987 on Build GRCh38
rs939292049 Chr.12: 71132680-71132680 on Build GRCh38
rs879236411 Chr.12: 71132828-71132829 on Build GRCh38
rs755221103 Chr.12: 71132755-71132756 on Build GRCh38
rs143898955 Chr.12: 71132694-71132694 on Build GRCh38
rs745609135 Chr.12: 71132831-71132831 on Build GRCh38
rs878911183 Chr.12: 71132827-71132827 on Build GRCh38
rs768769333 Chr.12: 71132851-71132851 on Build GRCh38
rs757095965 Chr.12: 71132856-71132856 on Build GRCh38

View Full Product Details