Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 8330528-8331022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC45A1
Entrez Gene ID
50651
Gene Name
solute carrier family 45 member 1
Gene Aliases
DNB5
UniGene
Hs.463036
Gene Chromosome Location
Chr.1: 8318085-8344167 on Build GRCh38
Transcripts
NM_001080397.2 XM_011541530.1 XM_011541531.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1602703 c.1109C>T p.T370M Chr.1: 8330602-8330602 on Build GRCh38
5442858 c.1481C>T p.P494L Chr.1: 8330872-8330872 on Build GRCh38
4460645 c.1276C>T p.P426S Chr.1: 8330667-8330667 on Build GRCh38
464990 c.1383C>T p.D461D Chr.1: 8330876-8330876 on Build GRCh38
5442857 c.1379C>T p.P460L Chr.1: 8330872-8330872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971870519 Chr.1: 8330556-8330556 on Build GRCh38
rs746396535 Chr.1: 8330786-8330786 on Build GRCh38
rs757302694 Chr.1: 8330691-8330691 on Build GRCh38
rs919719059 Chr.1: 8330652-8330652 on Build GRCh38
rs1004912560 Chr.1: 8330683-8330683 on Build GRCh38
rs964395673 Chr.1: 8330913-8330913 on Build GRCh38
rs529790748 Chr.1: 8330819-8330819 on Build GRCh38
rs987549963 Chr.1: 8330698-8330698 on Build GRCh38
rs893702427 Chr.1: 8330826-8330826 on Build GRCh38
rs763684329 Chr.1: 8330687-8330687 on Build GRCh38
rs201113608 Chr.1: 8330841-8330841 on Build GRCh38
rs764318169 Chr.1: 8330883-8330883 on Build GRCh38
rs1024426806 Chr.1: 8330551-8330551 on Build GRCh38
rs776965975 Chr.1: 8330718-8330718 on Build GRCh38
rs780271461 Chr.1: 8330915-8330915 on Build GRCh38
rs149921947 Chr.1: 8330777-8330777 on Build GRCh38
rs368992612 Chr.1: 8330642-8330642 on Build GRCh38
rs1020384798 Chr.1: 8330956-8330956 on Build GRCh38
rs535344724 Chr.1: 8330898-8330898 on Build GRCh38
rs760878880 Chr.1: 8330580-8330580 on Build GRCh38
rs143396132 Chr.1: 8330872-8330872 on Build GRCh38
rs747821410 Chr.1: 8330969-8330969 on Build GRCh38
rs749529510 Chr.1: 8330882-8330882 on Build GRCh38
rs771683470 Chr.1: 8330970-8330970 on Build GRCh38
rs201437087 Chr.1: 8330617-8330617 on Build GRCh38
rs571135364 Chr.1: 8330663-8330663 on Build GRCh38
rs920192932 Chr.1: 8330948-8330948 on Build GRCh38
rs200223590 Chr.1: 8330857-8330857 on Build GRCh38
rs749492882 Chr.1: 8330638-8330638 on Build GRCh38
rs372616601 Chr.1: 8330569-8330569 on Build GRCh38
rs777954848 Chr.1: 8330712-8330712 on Build GRCh38
rs200437676 Chr.1: 8330755-8330755 on Build GRCh38
rs368154825 Chr.1: 8330608-8330608 on Build GRCh38
rs762717907 Chr.1: 8330730-8330730 on Build GRCh38
rs748512128 Chr.1: 8330772-8330772 on Build GRCh38
rs200084739 Chr.1: 8330649-8330649 on Build GRCh38
rs139151699 Chr.1: 8330721-8330721 on Build GRCh38
rs267598735 Chr.1: 8330666-8330666 on Build GRCh38
rs753437408 Chr.1: 8330586-8330586 on Build GRCh38
rs910178683 Chr.1: 8330699-8330699 on Build GRCh38
rs775964927 Chr.1: 8330795-8330795 on Build GRCh38
rs755211086 Chr.1: 8330731-8330731 on Build GRCh38
rs374431547 Chr.1: 8330749-8330749 on Build GRCh38
rs373884958 Chr.1: 8330686-8330686 on Build GRCh38
rs377250283 Chr.1: 8330602-8330602 on Build GRCh38
rs767163607 Chr.1: 8330837-8330837 on Build GRCh38
rs954614646 Chr.1: 8330876-8330876 on Build GRCh38
rs747481559 Chr.1: 8330716-8330716 on Build GRCh38
rs754672835 Chr.1: 8330599-8330599 on Build GRCh38
rs137918218 Chr.1: 8330643-8330643 on Build GRCh38
rs138729525 Chr.1: 8330720-8330720 on Build GRCh38
rs2289731 Chr.1: 8330828-8330828 on Build GRCh38
rs770045434 Chr.1: 8330827-8330827 on Build GRCh38
rs761492222 Chr.1: 8330579-8330579 on Build GRCh38
rs201577337 Chr.1: 8330704-8330704 on Build GRCh38
rs149392083 Chr.1: 8330618-8330618 on Build GRCh38
rs199714755 Chr.1: 8330889-8330889 on Build GRCh38
rs768910859 Chr.1: 8330951-8330951 on Build GRCh38
rs748228656 Chr.1: 8330894-8330895 on Build GRCh38
rs757071373 Chr.1: 8330619-8330619 on Build GRCh38
rs764927970 Chr.1: 8330676-8330676 on Build GRCh38
rs758684449 Chr.1: 8330766-8330766 on Build GRCh38
rs937355657 Chr.1: 8330873-8330873 on Build GRCh38
rs753730878 Chr.1: 8330877-8330877 on Build GRCh38
rs748429750 Chr.1: 8330708-8330708 on Build GRCh38
rs201094126 Chr.1: 8330717-8330717 on Build GRCh38
rs746345039 Chr.1: 8330902-8330902 on Build GRCh38
rs771993387 Chr.1: 8330709-8330709 on Build GRCh38
rs503079 Chr.1: 8330601-8330601 on Build GRCh38
rs772188244 Chr.1: 8330907-8330909 on Build GRCh38
rs370439504 Chr.1: 8330740-8330740 on Build GRCh38
rs145948582 Chr.1: 8330697-8330697 on Build GRCh38
rs756713316 Chr.1: 8330909-8330909 on Build GRCh38
rs767970873 Chr.1: 8330573-8330573 on Build GRCh38
rs752399812 Chr.1: 8330678-8330678 on Build GRCh38
rs766399771 Chr.1: 8330982-8330982 on Build GRCh38
rs764843488 Chr.1: 8330585-8330585 on Build GRCh38
rs376067386 Chr.1: 8330739-8330739 on Build GRCh38
rs893208126 Chr.1: 8330933-8330933 on Build GRCh38
rs774558124 Chr.1: 8330800-8330803 on Build GRCh38
rs908479385 Chr.1: 8330782-8330782 on Build GRCh38
rs973021594 Chr.1: 8330944-8330944 on Build GRCh38
rs773547912 Chr.1: 8330648-8330648 on Build GRCh38
rs781133523 Chr.1: 8330696-8330696 on Build GRCh38
rs751431761 Chr.1: 8330689-8330689 on Build GRCh38
rs757968411 Chr.1: 8330610-8330610 on Build GRCh38
rs142956849 Chr.1: 8330981-8330981 on Build GRCh38
rs768696228 Chr.1: 8330669-8330670 on Build GRCh38
rs749611806 Chr.1: 8330918-8330918 on Build GRCh38
rs199862899 Chr.1: 8330671-8330671 on Build GRCh38
rs774616329 Chr.1: 8330570-8330570 on Build GRCh38
rs778931295 Chr.1: 8330705-8330705 on Build GRCh38
rs887015026 Chr.1: 8330682-8330682 on Build GRCh38
rs757279390 Chr.1: 8330897-8330897 on Build GRCh38
rs759183998 Chr.1: 8330675-8330675 on Build GRCh38
rs755928603 Chr.1: 8330741-8330741 on Build GRCh38
rs751151036 Chr.1: 8330737-8330737 on Build GRCh38
rs540833665 Chr.1: 8330778-8330778 on Build GRCh38
rs535551148 Chr.1: 8330986-8330986 on Build GRCh38
rs975526404 Chr.1: 8330738-8330738 on Build GRCh38
rs752215389 Chr.1: 8330603-8330603 on Build GRCh38
rs150269149 Chr.1: 8330565-8330565 on Build GRCh38
rs769069066 Chr.1: 8330639-8330639 on Build GRCh38
rs762510680 Chr.1: 8330816-8330816 on Build GRCh38
rs202228969 Chr.1: 8330787-8330787 on Build GRCh38
rs777708048 Chr.1: 8330771-8330771 on Build GRCh38
rs774957284 Chr.1: 8330729-8330729 on Build GRCh38
rs772886052 Chr.1: 8330977-8330977 on Build GRCh38
rs942982282 Chr.1: 8330710-8330710 on Build GRCh38
rs781183365 Chr.1: 8330626-8330626 on Build GRCh38
rs201632307 Chr.1: 8330952-8330952 on Build GRCh38

View Full Product Details