Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9971636-9972144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMNAT1
Entrez Gene ID
64802
Gene Name
nicotinamide nucleotide adenylyltransferase 1
Gene Aliases
LCA9, NMNAT, PNAT1
UniGene
Hs.633762
Gene Chromosome Location
Chr.1: 9942923-9996739 on Build GRCh38
Transcripts
NM_001297778.1 NM_001297779.1 NM_022787.3 XM_011541971.2 XM_017002107.1 XM_017002108.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5001602 c.20C>A p.T7N Chr.1: 9972093-9972093 on Build GRCh38
5001601 c.20C>A p.T7N Chr.1: 9972093-9972093 on Build GRCh38
892886 c.12C>T p.S4S Chr.1: 9972085-9972085 on Build GRCh38
4463668 c.12C>T p.S4S Chr.1: 9972085-9972085 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200072546 Chr.1: 9972115-9972115 on Build GRCh38
rs1040521574 Chr.1: 9971790-9971790 on Build GRCh38
rs971801735 Chr.1: 9971908-9971908 on Build GRCh38
rs981573175 Chr.1: 9971942-9971942 on Build GRCh38
rs1032161100 Chr.1: 9971947-9971947 on Build GRCh38
rs914837846 Chr.1: 9971674-9971674 on Build GRCh38
rs148490817 Chr.1: 9972085-9972085 on Build GRCh38
rs533286126 Chr.1: 9971933-9971933 on Build GRCh38
rs932879619 Chr.1: 9971679-9971679 on Build GRCh38
rs986108441 Chr.1: 9972107-9972107 on Build GRCh38
rs543224218 Chr.1: 9971781-9971781 on Build GRCh38
rs748626122 Chr.1: 9971689-9971689 on Build GRCh38
rs751746188 Chr.1: 9972038-9972038 on Build GRCh38
rs112971844 Chr.1: 9972016-9972016 on Build GRCh38
rs887698762 Chr.1: 9971829-9971829 on Build GRCh38
rs201852975 Chr.1: 9972105-9972105 on Build GRCh38
rs387907294 Chr.1: 9972098-9972098 on Build GRCh38
rs566784324 Chr.1: 9972072-9972072 on Build GRCh38
rs944350491 Chr.1: 9972116-9972116 on Build GRCh38
rs1023001918 Chr.1: 9971884-9971884 on Build GRCh38
rs200890715 Chr.1: 9971973-9971974 on Build GRCh38
rs1027467115 Chr.1: 9971819-9971819 on Build GRCh38
rs61740429 Chr.1: 9972096-9972096 on Build GRCh38
rs944504525 Chr.1: 9971779-9971779 on Build GRCh38
rs897679399 Chr.1: 9971792-9971792 on Build GRCh38
rs551727721 Chr.1: 9971950-9971950 on Build GRCh38
rs757458161 Chr.1: 9972050-9972050 on Build GRCh38
rs138613460 Chr.1: 9972110-9972110 on Build GRCh38
rs189075646 Chr.1: 9971832-9971832 on Build GRCh38
rs1012808082 Chr.1: 9971867-9971867 on Build GRCh38
rs756649389 Chr.1: 9972074-9972074 on Build GRCh38
rs374109763 Chr.1: 9972037-9972037 on Build GRCh38
rs747825699 Chr.1: 9972086-9972086 on Build GRCh38
rs755650392 Chr.1: 9971936-9971936 on Build GRCh38
rs993316108 Chr.1: 9971800-9971800 on Build GRCh38
rs956096374 Chr.1: 9971974-9971974 on Build GRCh38
rs746522578 Chr.1: 9972101-9972101 on Build GRCh38
rs542611263 Chr.1: 9971974-9971974 on Build GRCh38
rs201044653 Chr.1: 9972112-9972112 on Build GRCh38
rs755365836 Chr.1: 9972082-9972082 on Build GRCh38
rs199561165 Chr.1: 9972091-9972091 on Build GRCh38
rs75644044 Chr.1: 9971870-9971870 on Build GRCh38
rs1019528916 Chr.1: 9971858-9971858 on Build GRCh38
rs991119721 Chr.1: 9971986-9971986 on Build GRCh38
rs749720958 Chr.1: 9972078-9972078 on Build GRCh38
rs367701663 Chr.1: 9972063-9972063 on Build GRCh38
rs759311025 Chr.1: 9972028-9972028 on Build GRCh38
rs77955144 Chr.1: 9971862-9971862 on Build GRCh38
rs531358447 Chr.1: 9971688-9971688 on Build GRCh38

View Full Product Details