Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9985135-9985638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMNAT1
Entrez Gene ID
64802
Gene Name
nicotinamide nucleotide adenylyltransferase 1
Gene Aliases
LCA9, NMNAT, PNAT1
UniGene
Hs.633762
Gene Chromosome Location
Chr.1: 9942923-9996739 on Build GRCh38
Transcripts
NM_001297778.1 NM_022787.3 XM_011541971.2 XM_017002108.1

SNP Detail

SNP ID Chromosome Location
rs916616133 Chr.1: 9985310-9985311 on Build GRCh38
rs868312331 Chr.1: 9985494-9985494 on Build GRCh38
rs567372758 Chr.1: 9985592-9985592 on Build GRCh38
rs778791137 Chr.1: 9985391-9985391 on Build GRCh38
rs1129990 Chr.1: 9985252-9985252 on Build GRCh38
rs966012710 Chr.1: 9985534-9985534 on Build GRCh38
rs1129991 Chr.1: 9985281-9985281 on Build GRCh38
rs527867747 Chr.1: 9985527-9985527 on Build GRCh38
rs948054530 Chr.1: 9985594-9985594 on Build GRCh38
rs1034718463 Chr.1: 9985408-9985408 on Build GRCh38
rs770868280 Chr.1: 9985364-9985364 on Build GRCh38
rs920493015 Chr.1: 9985249-9985249 on Build GRCh38
rs919233638 Chr.1: 9985230-9985230 on Build GRCh38
rs376429006 Chr.1: 9985232-9985232 on Build GRCh38
rs1129989 Chr.1: 9985219-9985219 on Build GRCh38
rs1017853671 Chr.1: 9985532-9985532 on Build GRCh38
rs1129984 Chr.1: 9985160-9985160 on Build GRCh38
rs1052640931 Chr.1: 9985438-9985438 on Build GRCh38
rs953125220 Chr.1: 9985189-9985189 on Build GRCh38
rs551997167 Chr.1: 9985319-9985319 on Build GRCh38
rs1010510545 Chr.1: 9985176-9985176 on Build GRCh38
rs1013046271 Chr.1: 9985293-9985293 on Build GRCh38
rs370825608 Chr.1: 9985395-9985395 on Build GRCh38
rs888786269 Chr.1: 9985473-9985473 on Build GRCh38
rs11808892 Chr.1: 9985503-9985503 on Build GRCh38
rs1008579413 Chr.1: 9985477-9985477 on Build GRCh38
rs991411614 Chr.1: 9985551-9985551 on Build GRCh38
rs533700517 Chr.1: 9985258-9985258 on Build GRCh38
rs1129992 Chr.1: 9985291-9985291 on Build GRCh38
rs930598172 Chr.1: 9985253-9985253 on Build GRCh38
rs867864490 Chr.1: 9985185-9985185 on Build GRCh38
rs529236396 Chr.1: 9985223-9985223 on Build GRCh38
rs944614764 Chr.1: 9985161-9985161 on Build GRCh38
rs1129986 Chr.1: 9985171-9985171 on Build GRCh38
rs982163288 Chr.1: 9985607-9985607 on Build GRCh38
rs397832540 Chr.1: 9985179-9985179 on Build GRCh38
rs1028583307 Chr.1: 9985465-9985465 on Build GRCh38
rs961851938 Chr.1: 9985523-9985523 on Build GRCh38
rs987190845 Chr.1: 9985193-9985193 on Build GRCh38
rs976392943 Chr.1: 9985540-9985540 on Build GRCh38
rs548765254 Chr.1: 9985528-9985528 on Build GRCh38
rs962206940 Chr.1: 9985226-9985226 on Build GRCh38
rs563150764 Chr.1: 9985240-9985240 on Build GRCh38
rs894046430 Chr.1: 9985445-9985445 on Build GRCh38
rs1129988 Chr.1: 9985186-9985186 on Build GRCh38
rs935511284 Chr.1: 9985431-9985431 on Build GRCh38
rs887822987 Chr.1: 9985274-9985274 on Build GRCh38
rs916544462 Chr.1: 9985573-9985573 on Build GRCh38
rs1011148608 Chr.1: 9985454-9985454 on Build GRCh38
rs1027526212 Chr.1: 9985530-9985530 on Build GRCh38
rs956372031 Chr.1: 9985486-9985486 on Build GRCh38
rs948083824 Chr.1: 9985314-9985314 on Build GRCh38

View Full Product Details