Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100503203-100503707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR1H4
Entrez Gene ID
9971
Gene Name
nuclear receptor subfamily 1 group H member 4
Gene Aliases
BAR, FXR, HRR-1, HRR1, RIP14
UniGene
Hs.282735
Gene Chromosome Location
Chr.12: 100473773-100563867 on Build GRCh38
Transcripts
NM_001206977.1 NM_001206978.1 NM_001206979.1 NM_001206992.1 NM_001206993.1 NM_005123.3 NR_135146.1 XM_006719719.2 XM_011539040.2 XM_011539041.2 XM_011539042.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3954027 c.44G>T p.S15I Chr.12: 100503431-100503431 on Build GRCh38
691717 c.93T>A p.S31R Chr.12: 100503480-100503480 on Build GRCh38
5625895 c.104C>T p.A35V Chr.12: 100503491-100503491 on Build GRCh38
7234892 c.44G>A p.S15N Chr.12: 100503431-100503431 on Build GRCh38
4037903 c.45T>C p.S15S Chr.12: 100503432-100503432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762091357 Chr.12: 100503382-100503382 on Build GRCh38
rs889813610 Chr.12: 100503322-100503322 on Build GRCh38
rs746952981 Chr.12: 100503625-100503625 on Build GRCh38
rs1056577979 Chr.12: 100503444-100503444 on Build GRCh38
rs754545354 Chr.12: 100503406-100503406 on Build GRCh38
rs569366944 Chr.12: 100503320-100503320 on Build GRCh38
rs891754942 Chr.12: 100503513-100503513 on Build GRCh38
rs7973216 Chr.12: 100503631-100503631 on Build GRCh38
rs889887628 Chr.12: 100503500-100503500 on Build GRCh38
rs1018676728 Chr.12: 100503520-100503520 on Build GRCh38
rs776845009 Chr.12: 100503381-100503381 on Build GRCh38
rs551129842 Chr.12: 100503263-100503263 on Build GRCh38
rs995644565 Chr.12: 100503299-100503299 on Build GRCh38
rs267603737 Chr.12: 100503479-100503479 on Build GRCh38
rs953239450 Chr.12: 100503321-100503321 on Build GRCh38
rs753523572 Chr.12: 100503527-100503527 on Build GRCh38
rs543990670 Chr.12: 100503606-100503606 on Build GRCh38
rs981087811 Chr.12: 100503590-100503590 on Build GRCh38
rs757084154 Chr.12: 100503528-100503528 on Build GRCh38
rs760766214 Chr.12: 100503370-100503370 on Build GRCh38
rs770729903 Chr.12: 100503436-100503436 on Build GRCh38
rs767116594 Chr.12: 100503507-100503507 on Build GRCh38
rs151248248 Chr.12: 100503604-100503604 on Build GRCh38
rs778750541 Chr.12: 100503533-100503533 on Build GRCh38
rs747025458 Chr.12: 100503452-100503452 on Build GRCh38
rs188325336 Chr.12: 100503512-100503512 on Build GRCh38
rs770945144 Chr.12: 100503340-100503340 on Build GRCh38
rs955997789 Chr.12: 100503329-100503329 on Build GRCh38
rs751060328 Chr.12: 100503398-100503398 on Build GRCh38
rs745794005 Chr.12: 100503540-100503540 on Build GRCh38
rs904464785 Chr.12: 100503241-100503241 on Build GRCh38
rs962006668 Chr.12: 100503663-100503663 on Build GRCh38
rs750847995 Chr.12: 100503386-100503386 on Build GRCh38
rs17030240 Chr.12: 100503438-100503438 on Build GRCh38
rs781439729 Chr.12: 100503302-100503302 on Build GRCh38
rs755684009 Chr.12: 100503524-100503524 on Build GRCh38
rs781654919 Chr.12: 100503399-100503399 on Build GRCh38
rs895643208 Chr.12: 100503448-100503448 on Build GRCh38
rs777272110 Chr.12: 100503624-100503624 on Build GRCh38
rs17030243 Chr.12: 100503670-100503670 on Build GRCh38
rs1037209643 Chr.12: 100503254-100503254 on Build GRCh38
rs777402336 Chr.12: 100503429-100503429 on Build GRCh38
rs748525265 Chr.12: 100503489-100503489 on Build GRCh38
rs750737066 Chr.12: 100503328-100503328 on Build GRCh38
rs373472692 Chr.12: 100503383-100503383 on Build GRCh38
rs1008165847 Chr.12: 100503323-100503323 on Build GRCh38
rs968603097 Chr.12: 100503564-100503564 on Build GRCh38
rs968945783 Chr.12: 100503349-100503349 on Build GRCh38
rs913947211 Chr.12: 100503649-100503649 on Build GRCh38
rs1037687738 Chr.12: 100503439-100503439 on Build GRCh38
rs17030242 Chr.12: 100503453-100503453 on Build GRCh38
rs756448155 Chr.12: 100503336-100503336 on Build GRCh38
rs761917707 Chr.12: 100503460-100503460 on Build GRCh38
rs1000228738 Chr.12: 100503248-100503248 on Build GRCh38
rs370621536 Chr.12: 100503491-100503491 on Build GRCh38
rs76198224 Chr.12: 100503618-100503618 on Build GRCh38
rs776748418 Chr.12: 100503459-100503459 on Build GRCh38
rs749143699 Chr.12: 100503434-100503434 on Build GRCh38
rs1021945924 Chr.12: 100503561-100503561 on Build GRCh38
rs201042707 Chr.12: 100503449-100503449 on Build GRCh38
rs760186422 Chr.12: 100503505-100503505 on Build GRCh38
rs376884786 Chr.12: 100503421-100503421 on Build GRCh38
rs766941311 Chr.12: 100503397-100503397 on Build GRCh38
rs1001385595 Chr.12: 100503617-100503617 on Build GRCh38
rs898620734 Chr.12: 100503289-100503289 on Build GRCh38
rs777504030 Chr.12: 100503526-100503526 on Build GRCh38
rs763337383 Chr.12: 100503389-100503389 on Build GRCh38
rs773408709 Chr.12: 100503474-100503474 on Build GRCh38
rs536694582 Chr.12: 100503373-100503373 on Build GRCh38
rs920154439 Chr.12: 100503675-100503675 on Build GRCh38
rs775396936 Chr.12: 100503341-100503341 on Build GRCh38
rs766734563 Chr.12: 100503490-100503490 on Build GRCh38
rs757714308 Chr.12: 100503270-100503270 on Build GRCh38
rs992506803 Chr.12: 100503416-100503416 on Build GRCh38
rs952267704 Chr.12: 100503544-100503544 on Build GRCh38
rs202244290 Chr.12: 100503412-100503412 on Build GRCh38
rs144715621 Chr.12: 100503473-100503473 on Build GRCh38
rs199956839 Chr.12: 100503492-100503492 on Build GRCh38
rs555887115 Chr.12: 100503614-100503614 on Build GRCh38

View Full Product Details