Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112405107-112405613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL6
Entrez Gene ID
6128
Gene Name
ribosomal protein L6
Gene Aliases
L6, SHUJUN-2, TAXREB107, TXREB1
UniGene
Hs.528668
Gene Chromosome Location
Chr.12: 112405181-112418850 on Build GRCh38
Transcripts
NM_000970.4 NM_001024662.2 NM_001320137.1 NM_001320138.1 NM_001320139.1 NM_001320140.1 NM_001320141.1 NM_001320142.1 XM_017019781.1 XM_017019782.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6419797 c.832A>G p.T278A Chr.12: 112405259-112405259 on Build GRCh38
6412155 c.750G>T p.Q250H Chr.12: 112405341-112405341 on Build GRCh38
6378709 c.854A>T p.K285I Chr.12: 112405237-112405237 on Build GRCh38
6412154 c.750G>T p.Q250H Chr.12: 112405341-112405341 on Build GRCh38
3710685 c.723G>C p.E241D Chr.12: 112405368-112405368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982131965 Chr.12: 112405462-112405462 on Build GRCh38
rs770203911 Chr.12: 112405285-112405285 on Build GRCh38
rs1054414986 Chr.12: 112405491-112405491 on Build GRCh38
rs199644294 Chr.12: 112405258-112405258 on Build GRCh38
rs755018845 Chr.12: 112405265-112405265 on Build GRCh38
rs763013604 Chr.12: 112405223-112405223 on Build GRCh38
rs959289336 Chr.12: 112405482-112405482 on Build GRCh38
rs939781226 Chr.12: 112405149-112405150 on Build GRCh38
rs369723671 Chr.12: 112405371-112405371 on Build GRCh38
rs878876444 Chr.12: 112405456-112405456 on Build GRCh38
rs753454038 Chr.12: 112405372-112405372 on Build GRCh38
rs574412503 Chr.12: 112405149-112405149 on Build GRCh38
rs755596305 Chr.12: 112405412-112405412 on Build GRCh38
rs748363567 Chr.12: 112405277-112405277 on Build GRCh38
rs530435248 Chr.12: 112405572-112405572 on Build GRCh38
rs372506857 Chr.12: 112405386-112405386 on Build GRCh38
rs766956794 Chr.12: 112405356-112405356 on Build GRCh38
rs770656516 Chr.12: 112405304-112405304 on Build GRCh38
rs997839730 Chr.12: 112405157-112405157 on Build GRCh38
rs926596835 Chr.12: 112405478-112405478 on Build GRCh38
rs138678598 Chr.12: 112405363-112405363 on Build GRCh38
rs759039631 Chr.12: 112405182-112405182 on Build GRCh38
rs756681985 Chr.12: 112405489-112405489 on Build GRCh38
rs754262209 Chr.12: 112405349-112405349 on Build GRCh38
rs1021073203 Chr.12: 112405389-112405389 on Build GRCh38
rs781434101 Chr.12: 112405269-112405269 on Build GRCh38
rs779525048 Chr.12: 112405257-112405257 on Build GRCh38
rs939834444 Chr.12: 112405535-112405535 on Build GRCh38
rs781413585 Chr.12: 112405414-112405414 on Build GRCh38
rs889396739 Chr.12: 112405190-112405191 on Build GRCh38
rs766656932 Chr.12: 112405408-112405408 on Build GRCh38
rs541588591 Chr.12: 112405164-112405164 on Build GRCh38
rs762792357 Chr.12: 112405177-112405177 on Build GRCh38
rs375788971 Chr.12: 112405410-112405410 on Build GRCh38
rs77546309 Chr.12: 112405534-112405534 on Build GRCh38
rs973688075 Chr.12: 112405271-112405271 on Build GRCh38
rs190878762 Chr.12: 112405445-112405445 on Build GRCh38
rs2301723 Chr.12: 112405559-112405559 on Build GRCh38
rs1026994605 Chr.12: 112405276-112405276 on Build GRCh38
rs755238468 Chr.12: 112405536-112405536 on Build GRCh38
rs984031303 Chr.12: 112405503-112405503 on Build GRCh38
rs1050035510 Chr.12: 112405570-112405570 on Build GRCh38
rs1015160692 Chr.12: 112405243-112405243 on Build GRCh38
rs765573513 Chr.12: 112405227-112405227 on Build GRCh38
rs970800067 Chr.12: 112405439-112405439 on Build GRCh38
rs755777373 Chr.12: 112405210-112405210 on Build GRCh38
rs144975352 Chr.12: 112405345-112405345 on Build GRCh38
rs747500436 Chr.12: 112405263-112405263 on Build GRCh38
rs933730683 Chr.12: 112405584-112405584 on Build GRCh38
rs376364700 Chr.12: 112405327-112405327 on Build GRCh38
rs900840704 Chr.12: 112405154-112405154 on Build GRCh38
rs11066285 Chr.12: 112405589-112405589 on Build GRCh38
rs1036742291 Chr.12: 112405150-112405150 on Build GRCh38
rs532509520 Chr.12: 112405452-112405452 on Build GRCh38
rs767049060 Chr.12: 112405185-112405185 on Build GRCh38
rs995892446 Chr.12: 112405140-112405140 on Build GRCh38
rs922394265 Chr.12: 112405562-112405562 on Build GRCh38
rs1007738616 Chr.12: 112405237-112405237 on Build GRCh38
rs766377263 Chr.12: 112405179-112405179 on Build GRCh38
rs751401290 Chr.12: 112405463-112405463 on Build GRCh38
rs759400397 Chr.12: 112405354-112405354 on Build GRCh38
rs773107304 Chr.12: 112405286-112405286 on Build GRCh38
rs141161168 Chr.12: 112405301-112405301 on Build GRCh38
rs116602545 Chr.12: 112405148-112405148 on Build GRCh38
rs1032944421 Chr.12: 112405284-112405284 on Build GRCh38
rs1008758677 Chr.12: 112405194-112405194 on Build GRCh38
rs370747206 Chr.12: 112405362-112405362 on Build GRCh38
rs773715273 Chr.12: 112405181-112405181 on Build GRCh38
rs752283937 Chr.12: 112405186-112405186 on Build GRCh38
rs748275719 Chr.12: 112405426-112405426 on Build GRCh38

View Full Product Details