Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12260615-12261111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS13D
Entrez Gene ID
55187
Gene Name
vacuolar protein sorting 13 homolog D
Gene Aliases
-
UniGene
Hs.439381
Gene Chromosome Location
Chr.1: 12230039-12512047 on Build GRCh38
Transcripts
NM_015378.3 NM_018156.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4522124 c.1147G>A p.E383K Chr.1: 12260729-12260729 on Build GRCh38
894803 c.1124G>A p.R375Q Chr.1: 12260706-12260706 on Build GRCh38
5804149 c.1133A>T p.E378V Chr.1: 12260715-12260715 on Build GRCh38
894802 c.1123C>T p.R375W Chr.1: 12260705-12260705 on Build GRCh38
1185320 c.1325G>A p.G442D Chr.1: 12261060-12261060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200482221 Chr.1: 12260695-12260695 on Build GRCh38
rs776648168 Chr.1: 12261042-12261042 on Build GRCh38
rs752221445 Chr.1: 12260981-12260981 on Build GRCh38
rs6657877 Chr.1: 12260892-12260892 on Build GRCh38
rs199641752 Chr.1: 12261068-12261068 on Build GRCh38
rs768737006 Chr.1: 12260747-12260747 on Build GRCh38
rs780534218 Chr.1: 12260998-12260998 on Build GRCh38
rs759086538 Chr.1: 12261051-12261051 on Build GRCh38
rs752015762 Chr.1: 12260784-12260784 on Build GRCh38
rs775228221 Chr.1: 12261067-12261067 on Build GRCh38
rs555141309 Chr.1: 12260886-12260886 on Build GRCh38
rs750321685 Chr.1: 12260766-12260766 on Build GRCh38
rs747979971 Chr.1: 12260906-12260906 on Build GRCh38
rs763703295 Chr.1: 12261071-12261071 on Build GRCh38
rs769795678 Chr.1: 12260714-12260714 on Build GRCh38
rs757502708 Chr.1: 12260787-12260787 on Build GRCh38
rs750007705 Chr.1: 12260649-12260649 on Build GRCh38
rs765941549 Chr.1: 12260774-12260774 on Build GRCh38
rs76782506 Chr.1: 12260696-12260696 on Build GRCh38
rs779859519 Chr.1: 12260654-12260654 on Build GRCh38
rs112286021 Chr.1: 12260964-12260964 on Build GRCh38
rs760920787 Chr.1: 12260843-12260843 on Build GRCh38
rs910434071 Chr.1: 12260639-12260639 on Build GRCh38
rs138147813 Chr.1: 12260737-12260737 on Build GRCh38
rs774887046 Chr.1: 12260646-12260648 on Build GRCh38
rs752359125 Chr.1: 12260765-12260765 on Build GRCh38
rs756456366 Chr.1: 12261078-12261078 on Build GRCh38
rs12033054 Chr.1: 12260807-12260807 on Build GRCh38
rs199791415 Chr.1: 12260650-12260650 on Build GRCh38
rs773306147 Chr.1: 12260944-12260944 on Build GRCh38
rs149554788 Chr.1: 12260748-12260748 on Build GRCh38
rs781723691 Chr.1: 12260802-12260802 on Build GRCh38
rs775717158 Chr.1: 12260736-12260736 on Build GRCh38
rs905420307 Chr.1: 12260790-12260790 on Build GRCh38
rs765851388 Chr.1: 12261079-12261079 on Build GRCh38
rs1003976998 Chr.1: 12260808-12260808 on Build GRCh38
rs200409077 Chr.1: 12261002-12261002 on Build GRCh38
rs140957286 Chr.1: 12261083-12261083 on Build GRCh38
rs755627616 Chr.1: 12260651-12260651 on Build GRCh38
rs758960081 Chr.1: 12260973-12260973 on Build GRCh38
rs778430207 Chr.1: 12260783-12260783 on Build GRCh38
rs769883435 Chr.1: 12260835-12260835 on Build GRCh38
rs557581730 Chr.1: 12261027-12261027 on Build GRCh38
rs772466193 Chr.1: 12260660-12260663 on Build GRCh38
rs535567591 Chr.1: 12260836-12260836 on Build GRCh38
rs772318879 Chr.1: 12261024-12261024 on Build GRCh38
rs375840170 Chr.1: 12260951-12260951 on Build GRCh38
rs1017669888 Chr.1: 12260729-12260729 on Build GRCh38
rs928936472 Chr.1: 12261064-12261064 on Build GRCh38
rs772266311 Chr.1: 12260701-12260701 on Build GRCh38
rs996685089 Chr.1: 12260756-12260761 on Build GRCh38
rs779228981 Chr.1: 12261005-12261005 on Build GRCh38
rs771538100 Chr.1: 12260699-12260699 on Build GRCh38
rs756652458 Chr.1: 12260990-12260990 on Build GRCh38
rs753502351 Chr.1: 12260667-12260667 on Build GRCh38
rs766391071 Chr.1: 12260955-12260955 on Build GRCh38
rs985951184 Chr.1: 12260927-12260927 on Build GRCh38
rs776442029 Chr.1: 12260920-12260920 on Build GRCh38
rs774379629 Chr.1: 12260898-12260898 on Build GRCh38
rs767101690 Chr.1: 12260719-12260719 on Build GRCh38
rs757669477 Chr.1: 12260988-12260988 on Build GRCh38
rs973759719 Chr.1: 12261012-12261012 on Build GRCh38
rs201330814 Chr.1: 12260709-12260709 on Build GRCh38
rs146488112 Chr.1: 12261077-12261077 on Build GRCh38
rs149663017 Chr.1: 12261025-12261025 on Build GRCh38
rs894328398 Chr.1: 12261055-12261055 on Build GRCh38
rs150950475 Chr.1: 12260989-12260989 on Build GRCh38
rs375214360 Chr.1: 12260923-12260923 on Build GRCh38
rs767326123 Chr.1: 12260646-12260646 on Build GRCh38
rs760294849 Chr.1: 12260769-12260769 on Build GRCh38
rs749334750 Chr.1: 12260838-12260838 on Build GRCh38
rs764952329 Chr.1: 12260974-12260974 on Build GRCh38
rs781533659 Chr.1: 12260706-12260706 on Build GRCh38
rs145656125 Chr.1: 12261028-12261028 on Build GRCh38
rs764609068 Chr.1: 12261065-12261065 on Build GRCh38
rs767503767 Chr.1: 12260756-12260756 on Build GRCh38
rs766877176 Chr.1: 12261080-12261080 on Build GRCh38
rs1022348390 Chr.1: 12260887-12260887 on Build GRCh38
rs758859544 Chr.1: 12260685-12260685 on Build GRCh38
rs141005030 Chr.1: 12260954-12260954 on Build GRCh38
rs773501645 Chr.1: 12260939-12260939 on Build GRCh38
rs370704795 Chr.1: 12260778-12260778 on Build GRCh38
rs755110629 Chr.1: 12261088-12261088 on Build GRCh38
rs750359917 Chr.1: 12261004-12261004 on Build GRCh38
rs762148690 Chr.1: 12260651-12260656 on Build GRCh38

View Full Product Details