Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35081285-35081809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM177A1
Entrez Gene ID
283635
Gene Name
family with sequence similarity 177 member A1
Gene Aliases
C14orf24
UniGene
Hs.446357
Gene Chromosome Location
Chr.14: 35044907-35083383 on Build GRCh38
Transcripts
NM_001079519.1 NM_001289022.1 NM_173607.3 NR_110415.1

Gene Symbol
LOC101927178
Entrez Gene ID
101927178
Gene Name
uncharacterized LOC101927178
Gene Aliases
-
UniGene
Hs.617139
Gene Chromosome Location
Chr.14: 35081081-35113130 on Build GRCh38
Transcripts
NM_001079519.1 NM_001289022.1 NM_173607.3 NR_110415.1

SNP Detail

SNP ID Chromosome Location
rs913479731 Chr.14: 35081472-35081473 on Build GRCh38
rs758431524 Chr.14: 35081513-35081513 on Build GRCh38
rs1054039470 Chr.14: 35081749-35081749 on Build GRCh38
rs952624593 Chr.14: 35081564-35081564 on Build GRCh38
rs537120671 Chr.14: 35081311-35081311 on Build GRCh38
rs779655982 Chr.14: 35081767-35081767 on Build GRCh38
rs957414459 Chr.14: 35081705-35081705 on Build GRCh38
rs1040653729 Chr.14: 35081741-35081741 on Build GRCh38
rs910805644 Chr.14: 35081691-35081691 on Build GRCh38
rs559661562 Chr.14: 35081716-35081716 on Build GRCh38
rs1041020820 Chr.14: 35081490-35081490 on Build GRCh38
rs995825002 Chr.14: 35081519-35081519 on Build GRCh38
rs922154428 Chr.14: 35081763-35081763 on Build GRCh38
rs961174312 Chr.14: 35081602-35081602 on Build GRCh38
rs79788636 Chr.14: 35081494-35081494 on Build GRCh38
rs1013536416 Chr.14: 35081609-35081609 on Build GRCh38
rs887496275 Chr.14: 35081556-35081556 on Build GRCh38
rs543530405 Chr.14: 35081738-35081738 on Build GRCh38
rs570496776 Chr.14: 35081340-35081340 on Build GRCh38
rs1024712714 Chr.14: 35081392-35081392 on Build GRCh38
rs1034015501 Chr.14: 35081522-35081522 on Build GRCh38
rs960817273 Chr.14: 35081604-35081604 on Build GRCh38
rs936943062 Chr.14: 35081699-35081699 on Build GRCh38
rs926018507 Chr.14: 35081379-35081379 on Build GRCh38
rs892631056 Chr.14: 35081335-35081335 on Build GRCh38
rs972149340 Chr.14: 35081615-35081615 on Build GRCh38
rs537801469 Chr.14: 35081318-35081318 on Build GRCh38
rs371523743 Chr.14: 35081689-35081689 on Build GRCh38
rs983995426 Chr.14: 35081576-35081576 on Build GRCh38
rs900195361 Chr.14: 35081516-35081516 on Build GRCh38
rs112571405 Chr.14: 35081480-35081480 on Build GRCh38
rs926049662 Chr.14: 35081673-35081676 on Build GRCh38
rs1053926403 Chr.14: 35081325-35081325 on Build GRCh38
rs182873307 Chr.14: 35081779-35081779 on Build GRCh38
rs376880814 Chr.14: 35081720-35081720 on Build GRCh38
rs191784433 Chr.14: 35081628-35081628 on Build GRCh38
rs543523630 Chr.14: 35081574-35081575 on Build GRCh38
rs145922727 Chr.14: 35081781-35081781 on Build GRCh38
rs144052189 Chr.14: 35081353-35081353 on Build GRCh38
rs936188753 Chr.14: 35081414-35081414 on Build GRCh38
rs980045766 Chr.14: 35081667-35081667 on Build GRCh38
rs184025379 Chr.14: 35081322-35081322 on Build GRCh38
rs945018197 Chr.14: 35081476-35081476 on Build GRCh38
rs555644005 Chr.14: 35081663-35081663 on Build GRCh38
rs573408624 Chr.14: 35081515-35081515 on Build GRCh38
rs1050411868 Chr.14: 35081679-35081679 on Build GRCh38
rs913435916 Chr.14: 35081716-35081716 on Build GRCh38
rs908420788 Chr.14: 35081580-35081580 on Build GRCh38
rs757826176 Chr.14: 35081649-35081649 on Build GRCh38
rs1053183416 Chr.14: 35081460-35081460 on Build GRCh38
rs187338760 Chr.14: 35081621-35081621 on Build GRCh38

View Full Product Details