Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 36684658-36685156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A21
Entrez Gene ID
89874
Gene Name
solute carrier family 25 member 21
Gene Aliases
ODC, ODC1
UniGene
Hs.730857
Gene Chromosome Location
Chr.14: 36677921-37172660 on Build GRCh38
Transcripts
NM_001171170.1 NM_030631.3 XM_011537287.2 XM_011537288.2 XM_011537289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
955581 c.692C>T p.A231V Chr.14: 36684837-36684837 on Build GRCh38
4595768 c.665C>T p.S222L Chr.14: 36684864-36684864 on Build GRCh38
4394397 c.778G>A p.E260K Chr.14: 36684751-36684751 on Build GRCh38
6780573 c.758C>A p.T253K Chr.14: 36684771-36684771 on Build GRCh38
6015500 c.724C>T p.P242S Chr.14: 36684805-36684805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033903243 Chr.14: 36684876-36684876 on Build GRCh38
rs1036996141 Chr.14: 36684687-36684687 on Build GRCh38
rs146678499 Chr.14: 36684865-36684865 on Build GRCh38
rs779354786 Chr.14: 36684907-36684907 on Build GRCh38
rs778946635 Chr.14: 36684738-36684738 on Build GRCh38
rs368483580 Chr.14: 36684919-36684919 on Build GRCh38
rs763959858 Chr.14: 36684715-36684715 on Build GRCh38
rs894971161 Chr.14: 36685073-36685073 on Build GRCh38
rs759035550 Chr.14: 36684698-36684698 on Build GRCh38
rs749664916 Chr.14: 36684897-36684897 on Build GRCh38
rs148530715 Chr.14: 36684943-36684943 on Build GRCh38
rs777717548 Chr.14: 36685108-36685108 on Build GRCh38
rs911364768 Chr.14: 36684962-36684962 on Build GRCh38
rs755022802 Chr.14: 36684756-36684756 on Build GRCh38
rs373715236 Chr.14: 36684700-36684700 on Build GRCh38
rs762912767 Chr.14: 36684953-36684953 on Build GRCh38
rs746373141 Chr.14: 36684908-36684908 on Build GRCh38
rs372358161 Chr.14: 36684789-36684789 on Build GRCh38
rs746470725 Chr.14: 36684927-36684927 on Build GRCh38
rs757292423 Chr.14: 36684735-36684735 on Build GRCh38
rs569175370 Chr.14: 36684791-36684791 on Build GRCh38
rs528200258 Chr.14: 36685066-36685066 on Build GRCh38
rs375416783 Chr.14: 36684886-36684886 on Build GRCh38
rs1011625059 Chr.14: 36685051-36685051 on Build GRCh38
rs141855997 Chr.14: 36685049-36685049 on Build GRCh38
rs769069480 Chr.14: 36684709-36684709 on Build GRCh38
rs750373497 Chr.14: 36684873-36684873 on Build GRCh38
rs774784455 Chr.14: 36684713-36684713 on Build GRCh38
rs766184073 Chr.14: 36684963-36684963 on Build GRCh38
rs539795840 Chr.14: 36684838-36684838 on Build GRCh38
rs1006669783 Chr.14: 36684753-36684753 on Build GRCh38
rs1017928170 Chr.14: 36684811-36684811 on Build GRCh38
rs184683668 Chr.14: 36684990-36684990 on Build GRCh38
rs747918370 Chr.14: 36684863-36684864 on Build GRCh38
rs771523078 Chr.14: 36684917-36684917 on Build GRCh38
rs781339634 Chr.14: 36684758-36684758 on Build GRCh38
rs748392929 Chr.14: 36684766-36684766 on Build GRCh38
rs537051049 Chr.14: 36685123-36685123 on Build GRCh38
rs201299512 Chr.14: 36684906-36684906 on Build GRCh38
rs267603985 Chr.14: 36684797-36684797 on Build GRCh38
rs755075132 Chr.14: 36684878-36684878 on Build GRCh38
rs774862375 Chr.14: 36684823-36684823 on Build GRCh38
rs771447285 Chr.14: 36684802-36684802 on Build GRCh38
rs981237719 Chr.14: 36684869-36684869 on Build GRCh38
rs752793158 Chr.14: 36684964-36684964 on Build GRCh38
rs943532243 Chr.14: 36685034-36685034 on Build GRCh38
rs760372757 Chr.14: 36684845-36684845 on Build GRCh38
rs958767958 Chr.14: 36684893-36684893 on Build GRCh38
rs267603986 Chr.14: 36684851-36684851 on Build GRCh38
rs752898382 Chr.14: 36684879-36684879 on Build GRCh38
rs773077020 Chr.14: 36684949-36684949 on Build GRCh38
rs765071286 Chr.14: 36684946-36684946 on Build GRCh38
rs140386930 Chr.14: 36684896-36684896 on Build GRCh38
rs751117253 Chr.14: 36684954-36684955 on Build GRCh38
rs147474937 Chr.14: 36684940-36684940 on Build GRCh38
rs749611395 Chr.14: 36684780-36684780 on Build GRCh38
rs930218909 Chr.14: 36685022-36685022 on Build GRCh38
rs898077038 Chr.14: 36684688-36684688 on Build GRCh38
rs763800167 Chr.14: 36684857-36684857 on Build GRCh38
rs989786105 Chr.14: 36684882-36684882 on Build GRCh38
rs761644645 Chr.14: 36684868-36684868 on Build GRCh38
rs150614998 Chr.14: 36685050-36685050 on Build GRCh38
rs761769431 Chr.14: 36684944-36684944 on Build GRCh38
rs568022532 Chr.14: 36685121-36685121 on Build GRCh38
rs201104006 Chr.14: 36684712-36684712 on Build GRCh38
rs941577145 Chr.14: 36684686-36684686 on Build GRCh38
rs1011260360 Chr.14: 36684832-36684832 on Build GRCh38
rs765925319 Chr.14: 36684928-36684932 on Build GRCh38
rs1010776523 Chr.14: 36685076-36685076 on Build GRCh38
rs770189774 Chr.14: 36684767-36684767 on Build GRCh38
rs889317907 Chr.14: 36685027-36685027 on Build GRCh38
rs753867574 Chr.14: 36684732-36684732 on Build GRCh38
rs778076388 Chr.14: 36684769-36684769 on Build GRCh38
rs548324146 Chr.14: 36685071-36685071 on Build GRCh38
rs765093427 Chr.14: 36684871-36684871 on Build GRCh38
rs771605649 Chr.14: 36684837-36684837 on Build GRCh38
rs758972415 Chr.14: 36684831-36684831 on Build GRCh38
rs751607788 Chr.14: 36684751-36684751 on Build GRCh38

View Full Product Details