Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 49598583-49599086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100288910
Entrez Gene ID
100288910
Gene Name
uncharacterized LOC100288910
Gene Aliases
-
UniGene
Hs.156367
Gene Chromosome Location
Chr.14: 49597660-49599158 on Build GRCh38
Transcripts
NM_152329.3 NM_203467.1 NR_037792.1 NR_037793.1

Gene Symbol
LRR1
Entrez Gene ID
122769
Gene Name
leucine rich repeat protein 1
Gene Aliases
4-1BBLRR, LRR-1, PPIL5
UniGene
Hs.451090
Gene Chromosome Location
Chr.14: 49598697-49614672 on Build GRCh38
Transcripts
NM_152329.3 NM_203467.1 NR_037792.1 NR_037793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4150794 c.159C>A p.S53R Chr.14: 49598704-49598704 on Build GRCh38
6353609 c.222G>C p.A74A Chr.14: 49598641-49598641 on Build GRCh38
3765761 c.137C>T p.S46F Chr.14: 49598726-49598726 on Build GRCh38
5946660 c.7C>T p.L3L Chr.14: 49599027-49599027 on Build GRCh38
5764161 c.140C>T p.P47L Chr.14: 49598723-49598723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769891684 Chr.14: 49598870-49598870 on Build GRCh38
rs534500128 Chr.14: 49598789-49598789 on Build GRCh38
rs193128280 Chr.14: 49598902-49598902 on Build GRCh38
rs905115050 Chr.14: 49598874-49598874 on Build GRCh38
rs376829507 Chr.14: 49598742-49598742 on Build GRCh38
rs781223690 Chr.14: 49598865-49598865 on Build GRCh38
rs774827042 Chr.14: 49599035-49599035 on Build GRCh38
rs552048643 Chr.14: 49598630-49598630 on Build GRCh38
rs756092770 Chr.14: 49598719-49598719 on Build GRCh38
rs2281837 Chr.14: 49598641-49598641 on Build GRCh38
rs763651696 Chr.14: 49599058-49599058 on Build GRCh38
rs542085969 Chr.14: 49598627-49598627 on Build GRCh38
rs760413305 Chr.14: 49599050-49599050 on Build GRCh38
rs45489097 Chr.14: 49598614-49598614 on Build GRCh38
rs1051730630 Chr.14: 49598697-49598697 on Build GRCh38
rs56203410 Chr.14: 49598983-49598983 on Build GRCh38
rs1009312180 Chr.14: 49598688-49598688 on Build GRCh38
rs967127117 Chr.14: 49598766-49598766 on Build GRCh38
rs371940926 Chr.14: 49598992-49598992 on Build GRCh38
rs950764576 Chr.14: 49598960-49598960 on Build GRCh38
rs954857643 Chr.14: 49598718-49598718 on Build GRCh38
rs1053899297 Chr.14: 49598889-49598889 on Build GRCh38
rs1011433911 Chr.14: 49598741-49598741 on Build GRCh38
rs116106140 Chr.14: 49598875-49598875 on Build GRCh38
rs555664038 Chr.14: 49598725-49598725 on Build GRCh38
rs973665915 Chr.14: 49598948-49598948 on Build GRCh38
rs949214994 Chr.14: 49598803-49598803 on Build GRCh38
rs111254895 Chr.14: 49598897-49598897 on Build GRCh38
rs1031968956 Chr.14: 49598684-49598684 on Build GRCh38
rs187516116 Chr.14: 49598727-49598727 on Build GRCh38
rs919426750 Chr.14: 49598878-49598878 on Build GRCh38
rs367587395 Chr.14: 49599013-49599013 on Build GRCh38
rs1009755834 Chr.14: 49598893-49598893 on Build GRCh38
rs12884574 Chr.14: 49598726-49598726 on Build GRCh38
rs889124636 Chr.14: 49598712-49598712 on Build GRCh38
rs1001879240 Chr.14: 49598679-49598679 on Build GRCh38
rs1002568628 Chr.14: 49598974-49598974 on Build GRCh38
rs747342738 Chr.14: 49598871-49598871 on Build GRCh38
rs747205989 Chr.14: 49598605-49598605 on Build GRCh38
rs964964238 Chr.14: 49598934-49598934 on Build GRCh38
rs1020714876 Chr.14: 49598895-49598895 on Build GRCh38
rs759329796 Chr.14: 49598685-49598685 on Build GRCh38
rs1035315061 Chr.14: 49598799-49598799 on Build GRCh38
rs141675587 Chr.14: 49598868-49598868 on Build GRCh38
rs1039623703 Chr.14: 49598951-49598951 on Build GRCh38
rs530985688 Chr.14: 49598615-49598615 on Build GRCh38
rs1021569675 Chr.14: 49598705-49598705 on Build GRCh38
rs756160890 Chr.14: 49599011-49599011 on Build GRCh38
rs775229347 Chr.14: 49599048-49599048 on Build GRCh38
rs530615974 Chr.14: 49598916-49598916 on Build GRCh38
rs1009021236 Chr.14: 49598908-49598908 on Build GRCh38
rs886930634 Chr.14: 49598635-49598635 on Build GRCh38
rs943387045 Chr.14: 49598613-49598613 on Build GRCh38
rs767321209 Chr.14: 49598975-49598975 on Build GRCh38
rs1047371009 Chr.14: 49598631-49598631 on Build GRCh38
rs752306798 Chr.14: 49598706-49598706 on Build GRCh38
rs556814580 Chr.14: 49598892-49598892 on Build GRCh38
rs933352229 Chr.14: 49598886-49598886 on Build GRCh38
rs567614258 Chr.14: 49598686-49598686 on Build GRCh38
rs867277673 Chr.14: 49598733-49598733 on Build GRCh38
rs542050485 Chr.14: 49598655-49598655 on Build GRCh38
rs577785049 Chr.14: 49598811-49598811 on Build GRCh38
rs776059298 Chr.14: 49599065-49599065 on Build GRCh38
rs889113436 Chr.14: 49598924-49598924 on Build GRCh38
rs757022709 Chr.14: 49598767-49598767 on Build GRCh38
rs747223711 Chr.14: 49598791-49598791 on Build GRCh38
rs772591431 Chr.14: 49599039-49599039 on Build GRCh38
rs773416950 Chr.14: 49598642-49598642 on Build GRCh38
rs200242739 Chr.14: 49598698-49598698 on Build GRCh38
rs768920344 Chr.14: 49598793-49598793 on Build GRCh38
rs371268573 Chr.14: 49599020-49599020 on Build GRCh38
rs903020512 Chr.14: 49598973-49598973 on Build GRCh38
rs770406526 Chr.14: 49598991-49598991 on Build GRCh38
rs771323197 Chr.14: 49598887-49598887 on Build GRCh38
rs1011924332 Chr.14: 49598957-49598957 on Build GRCh38
rs2281836 Chr.14: 49598999-49598999 on Build GRCh38
rs763336832 Chr.14: 49598646-49598646 on Build GRCh38
rs145978609 Chr.14: 49598987-49598987 on Build GRCh38
rs753738685 Chr.14: 49598990-49598990 on Build GRCh38
rs920677984 Chr.14: 49598626-49598626 on Build GRCh38
rs983830782 Chr.14: 49598962-49598962 on Build GRCh38
rs751157330 Chr.14: 49598662-49598662 on Build GRCh38
rs981983141 Chr.14: 49598994-49598994 on Build GRCh38
rs762291690 Chr.14: 49598632-49598632 on Build GRCh38
rs897885554 Chr.14: 49598680-49598680 on Build GRCh38
rs557488129 Chr.14: 49598800-49598800 on Build GRCh38
rs748391102 Chr.14: 49599010-49599010 on Build GRCh38
rs981167164 Chr.14: 49598796-49598796 on Build GRCh38
rs892948027 Chr.14: 49598740-49598740 on Build GRCh38
rs958397577 Chr.14: 49598804-49598804 on Build GRCh38
rs766675526 Chr.14: 49598657-49598657 on Build GRCh38
rs991685749 Chr.14: 49598818-49598818 on Build GRCh38
rs781696623 Chr.14: 49598953-49598953 on Build GRCh38
rs147992630 Chr.14: 49598755-49598755 on Build GRCh38
rs1017724809 Chr.14: 49598926-49598926 on Build GRCh38
rs746197235 Chr.14: 49599036-49599036 on Build GRCh38
rs750265840 Chr.14: 49598995-49598995 on Build GRCh38
rs771392301 Chr.14: 49599033-49599033 on Build GRCh38
rs909488825 Chr.14: 49598825-49598825 on Build GRCh38
rs973850023 Chr.14: 49598744-49598744 on Build GRCh38
rs763529661 Chr.14: 49598989-49598989 on Build GRCh38
rs375782086 Chr.14: 49599010-49599011 on Build GRCh38
rs893691600 Chr.14: 49599052-49599052 on Build GRCh38
rs1046232517 Chr.14: 49598819-49598819 on Build GRCh38

View Full Product Details