Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 47761006-47761519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA6D
Entrez Gene ID
80031
Gene Name
semaphorin 6D
Gene Aliases
-
UniGene
Hs.511265
Gene Chromosome Location
Chr.15: 47184101-47774223 on Build GRCh38
Transcripts
NM_001198999.1 NM_020858.1 NM_024966.2 NM_153616.1 NM_153617.1 NM_153618.1 NM_153619.1 XM_005254685.3 XM_005254686.2 XM_005254687.2 XM_005254689.3 XM_011522075.2 XM_011522076.2 XM_011522077.2 XM_011522078.2 XM_011522079.2 XM_011522080.2 XM_011522081.2 XM_017022617.1 XM_017022618.1 XM_017022619.1 XM_017022620.1 XM_017022621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6046098 c.359A>T p.N120I Chr.15: 47761343-47761343 on Build GRCh38
6142427 c.336C>A p.G112G Chr.15: 47761211-47761211 on Build GRCh38
6217018 c.285A>G p.K95K Chr.15: 47761160-47761160 on Build GRCh38
1587896 c.442T>G p.Y148D Chr.15: 47761426-47761426 on Build GRCh38
6194963 c.296G>T p.R99L Chr.15: 47761171-47761171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746895205 Chr.15: 47761083-47761083 on Build GRCh38
rs181632562 Chr.15: 47761462-47761462 on Build GRCh38
rs371683860 Chr.15: 47761295-47761295 on Build GRCh38
rs758178406 Chr.15: 47761076-47761076 on Build GRCh38
rs599111 Chr.15: 47761137-47761137 on Build GRCh38
rs540805949 Chr.15: 47761105-47761105 on Build GRCh38
rs576171821 Chr.15: 47761489-47761489 on Build GRCh38
rs142054743 Chr.15: 47761254-47761254 on Build GRCh38
rs779013254 Chr.15: 47761443-47761443 on Build GRCh38
rs944491428 Chr.15: 47761381-47761381 on Build GRCh38
rs546355749 Chr.15: 47761378-47761378 on Build GRCh38
rs562385125 Chr.15: 47761051-47761051 on Build GRCh38
rs757331801 Chr.15: 47761249-47761249 on Build GRCh38
rs777244903 Chr.15: 47761302-47761302 on Build GRCh38
rs774631553 Chr.15: 47761109-47761109 on Build GRCh38
rs773332083 Chr.15: 47761279-47761280 on Build GRCh38
rs990180902 Chr.15: 47761099-47761099 on Build GRCh38
rs767343230 Chr.15: 47761401-47761401 on Build GRCh38
rs1040924468 Chr.15: 47761415-47761415 on Build GRCh38
rs374736689 Chr.15: 47761031-47761031 on Build GRCh38
rs780417997 Chr.15: 47761452-47761452 on Build GRCh38
rs763151924 Chr.15: 47761320-47761323 on Build GRCh38
rs368693350 Chr.15: 47761070-47761070 on Build GRCh38
rs778910646 Chr.15: 47761251-47761251 on Build GRCh38
rs758561359 Chr.15: 47761450-47761450 on Build GRCh38
rs770204651 Chr.15: 47761468-47761468 on Build GRCh38
rs377217662 Chr.15: 47761119-47761119 on Build GRCh38
rs747364196 Chr.15: 47761288-47761288 on Build GRCh38
rs764927722 Chr.15: 47761054-47761054 on Build GRCh38
rs370119933 Chr.15: 47761166-47761166 on Build GRCh38
rs78293687 Chr.15: 47761171-47761171 on Build GRCh38
rs780507902 Chr.15: 47761262-47761262 on Build GRCh38
rs769296658 Chr.15: 47761344-47761344 on Build GRCh38
rs147097507 Chr.15: 47761255-47761255 on Build GRCh38
rs971621739 Chr.15: 47761485-47761485 on Build GRCh38
rs766658429 Chr.15: 47761466-47761468 on Build GRCh38
rs202186547 Chr.15: 47761363-47761363 on Build GRCh38
rs960829788 Chr.15: 47761207-47761207 on Build GRCh38
rs752609201 Chr.15: 47761404-47761404 on Build GRCh38
rs748644228 Chr.15: 47761467-47761467 on Build GRCh38
rs763650475 Chr.15: 47761049-47761049 on Build GRCh38
rs147211657 Chr.15: 47761177-47761177 on Build GRCh38
rs747274277 Chr.15: 47761456-47761456 on Build GRCh38
rs550852135 Chr.15: 47761474-47761474 on Build GRCh38
rs189191536 Chr.15: 47761458-47761458 on Build GRCh38
rs781098399 Chr.15: 47761189-47761189 on Build GRCh38
rs140523651 Chr.15: 47761374-47761374 on Build GRCh38
rs762273367 Chr.15: 47761305-47761305 on Build GRCh38
rs114660817 Chr.15: 47761278-47761278 on Build GRCh38
rs575200196 Chr.15: 47761047-47761047 on Build GRCh38
rs981317299 Chr.15: 47761488-47761488 on Build GRCh38
rs993790174 Chr.15: 47761113-47761113 on Build GRCh38
rs562027378 Chr.15: 47761377-47761377 on Build GRCh38
rs772580647 Chr.15: 47761215-47761215 on Build GRCh38
rs376339849 Chr.15: 47761084-47761084 on Build GRCh38
rs772461994 Chr.15: 47761122-47761122 on Build GRCh38
rs752701003 Chr.15: 47761214-47761214 on Build GRCh38
rs368939087 Chr.15: 47761371-47761371 on Build GRCh38
rs373094795 Chr.15: 47761085-47761085 on Build GRCh38
rs769119796 Chr.15: 47761457-47761457 on Build GRCh38
rs772624008 Chr.15: 47761356-47761356 on Build GRCh38
rs756028175 Chr.15: 47761435-47761435 on Build GRCh38
rs777334923 Chr.15: 47761143-47761143 on Build GRCh38
rs754855116 Chr.15: 47761179-47761179 on Build GRCh38
rs761401878 Chr.15: 47761151-47761151 on Build GRCh38
rs373474008 Chr.15: 47761052-47761052 on Build GRCh38
rs761225388 Chr.15: 47761131-47761131 on Build GRCh38
rs779760382 Chr.15: 47761078-47761078 on Build GRCh38
rs977335758 Chr.15: 47761285-47761285 on Build GRCh38
rs750475520 Chr.15: 47761446-47761446 on Build GRCh38
rs539110815 Chr.15: 47761051-47761051 on Build GRCh38
rs772717029 Chr.15: 47761471-47761471 on Build GRCh38
rs501916 Chr.15: 47761032-47761032 on Build GRCh38
rs200866177 Chr.15: 47761170-47761170 on Build GRCh38
rs573006671 Chr.15: 47761088-47761088 on Build GRCh38
rs764692263 Chr.15: 47761163-47761163 on Build GRCh38

View Full Product Details