Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52046183-52046728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK6
Entrez Gene ID
5597
Gene Name
mitogen-activated protein kinase 6
Gene Aliases
ERK3, HsT17250, PRKM6, p97MAPK
UniGene
Hs.411847
Gene Chromosome Location
Chr.15: 52004265-52066265 on Build GRCh38
Transcripts
NM_002748.3 XM_005254537.2 XM_005254538.2 XM_005254539.3 XM_011521782.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7151065 c.34C>T p.H12Y Chr.15: 52046494-52046494 on Build GRCh38
6331259 c.138A>G p.V46V Chr.15: 52046598-52046598 on Build GRCh38
2217209 c.189A>C p.L63L Chr.15: 52046649-52046649 on Build GRCh38
6301710 c.66C>T p.D22D Chr.15: 52046526-52046526 on Build GRCh38
2217208 c.162T>C p.T54T Chr.15: 52046622-52046622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140290030 Chr.15: 52046674-52046674 on Build GRCh38
rs750413811 Chr.15: 52046543-52046543 on Build GRCh38
rs767991240 Chr.15: 52046702-52046702 on Build GRCh38
rs145258990 Chr.15: 52046424-52046424 on Build GRCh38
rs945659106 Chr.15: 52046342-52046342 on Build GRCh38
rs965383981 Chr.15: 52046556-52046557 on Build GRCh38
rs989525229 Chr.15: 52046309-52046309 on Build GRCh38
rs779209290 Chr.15: 52046672-52046672 on Build GRCh38
rs937692765 Chr.15: 52046281-52046281 on Build GRCh38
rs753239878 Chr.15: 52046355-52046355 on Build GRCh38
rs536761280 Chr.15: 52046254-52046254 on Build GRCh38
rs536294389 Chr.15: 52046621-52046621 on Build GRCh38
rs768348426 Chr.15: 52046478-52046478 on Build GRCh38
rs373321122 Chr.15: 52046418-52046418 on Build GRCh38
rs551807024 Chr.15: 52046317-52046317 on Build GRCh38
rs371245995 Chr.15: 52046689-52046689 on Build GRCh38
rs901390417 Chr.15: 52046396-52046396 on Build GRCh38
rs779407633 Chr.15: 52046443-52046443 on Build GRCh38
rs889829521 Chr.15: 52046583-52046583 on Build GRCh38
rs753951278 Chr.15: 52046649-52046649 on Build GRCh38
rs771602277 Chr.15: 52046449-52046449 on Build GRCh38
rs774958224 Chr.15: 52046451-52046451 on Build GRCh38
rs756361678 Chr.15: 52046633-52046633 on Build GRCh38
rs776077924 Chr.15: 52046484-52046484 on Build GRCh38
rs757641589 Chr.15: 52046668-52046668 on Build GRCh38
rs767758720 Chr.15: 52046598-52046598 on Build GRCh38
rs746326607 Chr.15: 52046445-52046445 on Build GRCh38
rs113845594 Chr.15: 52046380-52046380 on Build GRCh38
rs867935778 Chr.15: 52046622-52046622 on Build GRCh38
rs147149863 Chr.15: 52046536-52046536 on Build GRCh38
rs767994752 Chr.15: 52046293-52046293 on Build GRCh38
rs746532599 Chr.15: 52046696-52046696 on Build GRCh38
rs762948597 Chr.15: 52046559-52046559 on Build GRCh38
rs1038749513 Chr.15: 52046703-52046703 on Build GRCh38
rs185311664 Chr.15: 52046407-52046407 on Build GRCh38
rs765071980 Chr.15: 52046540-52046540 on Build GRCh38
rs571528310 Chr.15: 52046367-52046367 on Build GRCh38
rs761678467 Chr.15: 52046521-52046521 on Build GRCh38
rs190224255 Chr.15: 52046426-52046426 on Build GRCh38
rs981891707 Chr.15: 52046212-52046212 on Build GRCh38
rs926331013 Chr.15: 52046228-52046228 on Build GRCh38
rs760301718 Chr.15: 52046470-52046470 on Build GRCh38
rs779496309 Chr.15: 52046691-52046691 on Build GRCh38

View Full Product Details