Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68164541-68165049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIAS1
Entrez Gene ID
8554
Gene Name
protein inhibitor of activated STAT 1
Gene Aliases
DDXBP1, GBP, GU/RH-II, ZMIZ3
UniGene
Hs.162458
Gene Chromosome Location
Chr.15: 68054179-68191464 on Build GRCh38
Transcripts
NM_001320687.1 NM_016166.2 XM_011522126.2 XM_011522127.2 XM_017022688.1 XM_017022689.1 XM_017022690.1 XM_017022691.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
470996 c.964A>G p.S322G Chr.15: 68164760-68164760 on Build GRCh38
6025613 c.1001T>C p.L334P Chr.15: 68164797-68164797 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs919695948 Chr.15: 68164789-68164789 on Build GRCh38
rs765747175 Chr.15: 68164798-68164798 on Build GRCh38
rs529106497 Chr.15: 68164856-68164856 on Build GRCh38
rs758939080 Chr.15: 68164838-68164838 on Build GRCh38
rs571300306 Chr.15: 68164783-68164783 on Build GRCh38
rs749693423 Chr.15: 68164796-68164796 on Build GRCh38
rs1048376377 Chr.15: 68164999-68164999 on Build GRCh38
rs750949287 Chr.15: 68164826-68164826 on Build GRCh38
rs550984039 Chr.15: 68164992-68164992 on Build GRCh38
rs772952348 Chr.15: 68164716-68164716 on Build GRCh38
rs772801338 Chr.15: 68164582-68164582 on Build GRCh38
rs887807453 Chr.15: 68164573-68164573 on Build GRCh38
rs565445089 Chr.15: 68164927-68164927 on Build GRCh38
rs748187644 Chr.15: 68164699-68164699 on Build GRCh38
rs534369062 Chr.15: 68164960-68164960 on Build GRCh38
rs758770172 Chr.15: 68164812-68164813 on Build GRCh38
rs912237180 Chr.15: 68164654-68164654 on Build GRCh38
rs975174444 Chr.15: 68164668-68164668 on Build GRCh38
rs949120981 Chr.15: 68164914-68164914 on Build GRCh38
rs774970714 Chr.15: 68164891-68164891 on Build GRCh38
rs1046241467 Chr.15: 68164932-68164932 on Build GRCh38
rs769582247 Chr.15: 68164674-68164674 on Build GRCh38
rs955092405 Chr.15: 68164968-68164968 on Build GRCh38
rs200837613 Chr.15: 68164686-68164686 on Build GRCh38
rs771167413 Chr.15: 68164807-68164807 on Build GRCh38
rs749125598 Chr.15: 68164719-68164719 on Build GRCh38
rs775005054 Chr.15: 68164795-68164795 on Build GRCh38
rs774195055 Chr.15: 68164761-68164761 on Build GRCh38
rs574594878 Chr.15: 68164633-68164633 on Build GRCh38
rs1046347694 Chr.15: 68164864-68164864 on Build GRCh38
rs759943939 Chr.15: 68164955-68164955 on Build GRCh38
rs893920054 Chr.15: 68164910-68164910 on Build GRCh38
rs908450661 Chr.15: 68164835-68164835 on Build GRCh38
rs769879843 Chr.15: 68164715-68164715 on Build GRCh38
rs750654471 Chr.15: 68164692-68164693 on Build GRCh38
rs766691827 Chr.15: 68164848-68164848 on Build GRCh38
rs962398882 Chr.15: 68164669-68164669 on Build GRCh38
rs1021611850 Chr.15: 68164594-68164594 on Build GRCh38
rs986464926 Chr.15: 68164650-68164650 on Build GRCh38
rs376205918 Chr.15: 68164853-68164853 on Build GRCh38
rs540153202 Chr.15: 68164863-68164866 on Build GRCh38
rs549151518 Chr.15: 68164909-68164909 on Build GRCh38
rs905214468 Chr.15: 68164866-68164866 on Build GRCh38
rs780342916 Chr.15: 68164822-68164822 on Build GRCh38
rs1010142751 Chr.15: 68164911-68164911 on Build GRCh38
rs955018944 Chr.15: 68164607-68164607 on Build GRCh38
rs543577040 Chr.15: 68164952-68164952 on Build GRCh38
rs767136670 Chr.15: 68164793-68164793 on Build GRCh38
rs191030763 Chr.15: 68164610-68164610 on Build GRCh38
rs549772742 Chr.15: 68164657-68164657 on Build GRCh38
rs770693174 Chr.15: 68164750-68164750 on Build GRCh38
rs888362956 Chr.15: 68165021-68165021 on Build GRCh38
rs796088090 Chr.15: 68164907-68164907 on Build GRCh38

View Full Product Details