Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 94345019-94345530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCTP2
Entrez Gene ID
55784
Gene Name
multiple C2 and transmembrane domain containing 2
Gene Aliases
-
UniGene
Hs.33368
Gene Chromosome Location
Chr.15: 94231184-94483952 on Build GRCh38
Transcripts
NM_001159643.1 NM_018349.3 XM_005254955.3 XM_006720603.2 XM_011521770.1 XM_011521771.2 XM_011521772.2 XM_011521773.2 XM_011521774.2 XM_017022403.1 XM_017022404.1 XM_017022405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
84312 c.970C>T p.R324C Chr.15: 94345129-94345129 on Build GRCh38
3936903 c.988C>T p.R330* Chr.15: 94345147-94345147 on Build GRCh38
6435061 c.1001G>A p.S334N Chr.15: 94345160-94345160 on Build GRCh38
6416651 c.973T>C p.W325R Chr.15: 94345132-94345132 on Build GRCh38
3887832 c.1005+1G>A p.? Chr.15: 94345165-94345165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009044768 Chr.15: 94345174-94345174 on Build GRCh38
rs376931832 Chr.15: 94345083-94345083 on Build GRCh38
rs771613991 Chr.15: 94345110-94345110 on Build GRCh38
rs926173674 Chr.15: 94345300-94345300 on Build GRCh38
rs751903241 Chr.15: 94345289-94345289 on Build GRCh38
rs917495739 Chr.15: 94345056-94345056 on Build GRCh38
rs940825874 Chr.15: 94345297-94345297 on Build GRCh38
rs989419014 Chr.15: 94345234-94345234 on Build GRCh38
rs1046485775 Chr.15: 94345061-94345061 on Build GRCh38
rs760689185 Chr.15: 94345196-94345196 on Build GRCh38
rs1009139754 Chr.15: 94345365-94345365 on Build GRCh38
rs150833683 Chr.15: 94345345-94345345 on Build GRCh38
rs764926630 Chr.15: 94345173-94345174 on Build GRCh38
rs774184065 Chr.15: 94345121-94345121 on Build GRCh38
rs1014607156 Chr.15: 94345221-94345221 on Build GRCh38
rs931930501 Chr.15: 94345448-94345448 on Build GRCh38
rs761710431 Chr.15: 94345122-94345122 on Build GRCh38
rs866423564 Chr.15: 94345169-94345170 on Build GRCh38
rs1049438345 Chr.15: 94345496-94345496 on Build GRCh38
rs1013195728 Chr.15: 94345273-94345273 on Build GRCh38
rs376471203 Chr.15: 94345176-94345176 on Build GRCh38
rs371516871 Chr.15: 94345112-94345112 on Build GRCh38
rs17712773 Chr.15: 94345070-94345070 on Build GRCh38
rs762642870 Chr.15: 94345174-94345175 on Build GRCh38
rs763052907 Chr.15: 94345216-94345216 on Build GRCh38
rs150589799 Chr.15: 94345130-94345130 on Build GRCh38
rs763751455 Chr.15: 94345120-94345120 on Build GRCh38
rs760284458 Chr.15: 94345090-94345090 on Build GRCh38
rs768021642 Chr.15: 94345094-94345095 on Build GRCh38
rs755191179 Chr.15: 94345079-94345079 on Build GRCh38
rs747417238 Chr.15: 94345156-94345156 on Build GRCh38
rs960398930 Chr.15: 94345241-94345241 on Build GRCh38
rs201413238 Chr.15: 94345184-94345184 on Build GRCh38
rs368031205 Chr.15: 94345100-94345100 on Build GRCh38
rs576551221 Chr.15: 94345372-94345372 on Build GRCh38
rs113569491 Chr.15: 94345119-94345119 on Build GRCh38
rs746383091 Chr.15: 94345160-94345160 on Build GRCh38
rs762767171 Chr.15: 94345415-94345415 on Build GRCh38
rs913312423 Chr.15: 94345380-94345380 on Build GRCh38
rs900429047 Chr.15: 94345131-94345131 on Build GRCh38
rs921903889 Chr.15: 94345425-94345425 on Build GRCh38
rs538171651 Chr.15: 94345186-94345186 on Build GRCh38
rs766387715 Chr.15: 94345206-94345211 on Build GRCh38
rs187855263 Chr.15: 94345147-94345147 on Build GRCh38
rs1023246517 Chr.15: 94345495-94345495 on Build GRCh38
rs369967192 Chr.15: 94345129-94345129 on Build GRCh38
rs748413093 Chr.15: 94345084-94345084 on Build GRCh38
rs199956375 Chr.15: 94345111-94345111 on Build GRCh38
rs927835736 Chr.15: 94345503-94345503 on Build GRCh38
rs867738947 Chr.15: 94345106-94345106 on Build GRCh38
rs769348862 Chr.15: 94345157-94345157 on Build GRCh38
rs776242042 Chr.15: 94345149-94345150 on Build GRCh38
rs957932833 Chr.15: 94345217-94345217 on Build GRCh38
rs774938937 Chr.15: 94345118-94345118 on Build GRCh38
rs533754857 Chr.15: 94345311-94345311 on Build GRCh38
rs1005119011 Chr.15: 94345507-94345507 on Build GRCh38
rs139457907 Chr.15: 94345148-94345148 on Build GRCh38
rs903345959 Chr.15: 94345391-94345391 on Build GRCh38
rs539111166 Chr.15: 94345395-94345395 on Build GRCh38
rs887639244 Chr.15: 94345501-94345501 on Build GRCh38
rs765177265 Chr.15: 94345212-94345212 on Build GRCh38
rs764921694 Chr.15: 94345322-94345322 on Build GRCh38
rs747468517 Chr.15: 94345109-94345109 on Build GRCh38
rs527872734 Chr.15: 94345113-94345113 on Build GRCh38
rs776276978 Chr.15: 94345195-94345195 on Build GRCh38
rs761581370 Chr.15: 94345151-94345154 on Build GRCh38
rs768399324 Chr.15: 94345178-94345178 on Build GRCh38
rs112905390 Chr.15: 94345282-94345282 on Build GRCh38
rs548186862 Chr.15: 94345286-94345286 on Build GRCh38
rs781742639 Chr.15: 94345159-94345159 on Build GRCh38
rs373530587 Chr.15: 94345151-94345151 on Build GRCh38
rs989078756 Chr.15: 94345460-94345460 on Build GRCh38
rs773063931 Chr.15: 94345194-94345194 on Build GRCh38
rs889342384 Chr.15: 94345351-94345351 on Build GRCh38
rs113959656 Chr.15: 94345404-94345404 on Build GRCh38
rs778230616 Chr.15: 94345102-94345102 on Build GRCh38
rs202137443 Chr.15: 94345141-94345141 on Build GRCh38
rs957747373 Chr.15: 94345411-94345411 on Build GRCh38
rs767310630 Chr.15: 94345190-94345190 on Build GRCh38
rs1023221492 Chr.15: 94345275-94345275 on Build GRCh38
rs750309440 Chr.15: 94345175-94345175 on Build GRCh38
rs568285043 Chr.15: 94345296-94345296 on Build GRCh38
rs1036102717 Chr.15: 94345508-94345508 on Build GRCh38
rs747535720 Chr.15: 94345097-94345097 on Build GRCh38
rs769147908 Chr.15: 94345165-94345165 on Build GRCh38

View Full Product Details