Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24548757-24549304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP6
Entrez Gene ID
5930
Gene Name
RB binding protein 6, ubiquitin ligase
Gene Aliases
MY038, P2P-R, PACT, RBQ-1, SNAMA
UniGene
Hs.188553
Gene Chromosome Location
Chr.16: 24539587-24572863 on Build GRCh38
Transcripts
NM_006910.4 NM_018703.3 NM_032626.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
968940 c.283G>T p.A95S Chr.16: 24548961-24548961 on Build GRCh38
76184 c.272G>A p.R91Q Chr.16: 24548950-24548950 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs570115601 Chr.16: 24549092-24549092 on Build GRCh38
rs749481890 Chr.16: 24549008-24549008 on Build GRCh38
rs72783902 Chr.16: 24548784-24548784 on Build GRCh38
rs754677047 Chr.16: 24548902-24548902 on Build GRCh38
rs933921826 Chr.16: 24548876-24548876 on Build GRCh38
rs748988481 Chr.16: 24548949-24548949 on Build GRCh38
rs566425324 Chr.16: 24549151-24549151 on Build GRCh38
rs953410337 Chr.16: 24548957-24548957 on Build GRCh38
rs992849028 Chr.16: 24549141-24549141 on Build GRCh38
rs780670947 Chr.16: 24548908-24548908 on Build GRCh38
rs1045440864 Chr.16: 24549274-24549274 on Build GRCh38
rs903040566 Chr.16: 24548871-24548871 on Build GRCh38
rs116753683 Chr.16: 24549163-24549163 on Build GRCh38
rs746954580 Chr.16: 24549047-24549048 on Build GRCh38
rs1054770210 Chr.16: 24548885-24548885 on Build GRCh38
rs751929377 Chr.16: 24549077-24549077 on Build GRCh38
rs949778362 Chr.16: 24549211-24549211 on Build GRCh38
rs775576986 Chr.16: 24548939-24548939 on Build GRCh38
rs949809984 Chr.16: 24548833-24548833 on Build GRCh38
rs904015622 Chr.16: 24549011-24549011 on Build GRCh38
rs962191652 Chr.16: 24549056-24549056 on Build GRCh38
rs539005354 Chr.16: 24549113-24549113 on Build GRCh38
rs548507486 Chr.16: 24548920-24548925 on Build GRCh38
rs530727501 Chr.16: 24549065-24549065 on Build GRCh38
rs745415572 Chr.16: 24548909-24548910 on Build GRCh38
rs774392196 Chr.16: 24549196-24549196 on Build GRCh38
rs761988126 Chr.16: 24549038-24549038 on Build GRCh38
rs747726563 Chr.16: 24548909-24548909 on Build GRCh38
rs751253519 Chr.16: 24548901-24548901 on Build GRCh38
rs201348556 Chr.16: 24549070-24549070 on Build GRCh38
rs940784120 Chr.16: 24549215-24549215 on Build GRCh38
rs770135687 Chr.16: 24548873-24548873 on Build GRCh38
rs897040056 Chr.16: 24548840-24548840 on Build GRCh38
rs1011324331 Chr.16: 24548910-24548910 on Build GRCh38
rs1008999000 Chr.16: 24549006-24549007 on Build GRCh38
rs1023615393 Chr.16: 24549162-24549162 on Build GRCh38
rs769056475 Chr.16: 24549035-24549035 on Build GRCh38
rs373345269 Chr.16: 24548914-24548914 on Build GRCh38
rs1045509672 Chr.16: 24548859-24548859 on Build GRCh38
rs145572792 Chr.16: 24548962-24548962 on Build GRCh38
rs766732912 Chr.16: 24549075-24549075 on Build GRCh38
rs909264502 Chr.16: 24549201-24549201 on Build GRCh38
rs750958664 Chr.16: 24549069-24549069 on Build GRCh38
rs999653915 Chr.16: 24549055-24549055 on Build GRCh38
rs893088796 Chr.16: 24548890-24548890 on Build GRCh38
rs148048189 Chr.16: 24548963-24548963 on Build GRCh38
rs771886841 Chr.16: 24548910-24548911 on Build GRCh38
rs952331893 Chr.16: 24549107-24549107 on Build GRCh38
rs760955205 Chr.16: 24549017-24549017 on Build GRCh38
rs776846062 Chr.16: 24549036-24549036 on Build GRCh38
rs202138834 Chr.16: 24548912-24548912 on Build GRCh38
rs546714672 Chr.16: 24549150-24549150 on Build GRCh38

View Full Product Details