Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24559391-24559907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP6
Entrez Gene ID
5930
Gene Name
RB binding protein 6, ubiquitin ligase
Gene Aliases
MY038, P2P-R, PACT, RBQ-1, SNAMA
UniGene
Hs.188553
Gene Chromosome Location
Chr.16: 24539587-24572863 on Build GRCh38
Transcripts
NM_006910.4 NM_018703.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4962756 c.836A>G p.Y279C Chr.16: 24559666-24559666 on Build GRCh38
6763523 c.755C>A p.P252H Chr.16: 24559585-24559585 on Build GRCh38
4881084 c.826G>T p.G276* Chr.16: 24559656-24559656 on Build GRCh38
5012991 c.790G>A p.D264N Chr.16: 24559620-24559620 on Build GRCh38
6573477 c.714C>G p.F238L Chr.16: 24559544-24559544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748768320 Chr.16: 24559630-24559630 on Build GRCh38
rs767560774 Chr.16: 24559706-24559706 on Build GRCh38
rs754475421 Chr.16: 24559458-24559458 on Build GRCh38
rs1010162051 Chr.16: 24559646-24559646 on Build GRCh38
rs1027544328 Chr.16: 24559769-24559769 on Build GRCh38
rs530026638 Chr.16: 24559548-24559548 on Build GRCh38
rs758492115 Chr.16: 24559493-24559493 on Build GRCh38
rs757469000 Chr.16: 24559484-24559484 on Build GRCh38
rs770534799 Chr.16: 24559661-24559661 on Build GRCh38
rs373161623 Chr.16: 24559485-24559485 on Build GRCh38
rs73560576 Chr.16: 24559851-24559851 on Build GRCh38
rs549890813 Chr.16: 24559692-24559692 on Build GRCh38
rs45591839 Chr.16: 24559523-24559523 on Build GRCh38
rs1002948600 Chr.16: 24559703-24559703 on Build GRCh38
rs1017028071 Chr.16: 24559452-24559452 on Build GRCh38
rs975897853 Chr.16: 24559809-24559809 on Build GRCh38
rs540977089 Chr.16: 24559795-24559795 on Build GRCh38
rs907567796 Chr.16: 24559878-24559878 on Build GRCh38
rs921359507 Chr.16: 24559821-24559821 on Build GRCh38
rs755557428 Chr.16: 24559538-24559538 on Build GRCh38
rs778349718 Chr.16: 24559735-24559735 on Build GRCh38
rs907286180 Chr.16: 24559698-24559698 on Build GRCh38
rs893384211 Chr.16: 24559610-24559610 on Build GRCh38
rs940256459 Chr.16: 24559840-24559840 on Build GRCh38
rs977397929 Chr.16: 24559729-24559729 on Build GRCh38
rs1051217075 Chr.16: 24559826-24559826 on Build GRCh38
rs758942550 Chr.16: 24559719-24559719 on Build GRCh38
rs759381718 Chr.16: 24559702-24559702 on Build GRCh38
rs545565114 Chr.16: 24559455-24559455 on Build GRCh38
rs1054673596 Chr.16: 24559577-24559577 on Build GRCh38
rs934124976 Chr.16: 24559824-24559824 on Build GRCh38
rs898146915 Chr.16: 24559872-24559873 on Build GRCh38
rs769516225 Chr.16: 24559563-24559563 on Build GRCh38
rs3815906 Chr.16: 24559436-24559436 on Build GRCh38
rs866270306 Chr.16: 24559835-24559835 on Build GRCh38
rs887335106 Chr.16: 24559830-24559830 on Build GRCh38
rs1003804579 Chr.16: 24559450-24559450 on Build GRCh38
rs750441922 Chr.16: 24559487-24559487 on Build GRCh38
rs965777150 Chr.16: 24559884-24559884 on Build GRCh38
rs936046624 Chr.16: 24559794-24559794 on Build GRCh38
rs369086557 Chr.16: 24559476-24559476 on Build GRCh38
rs765391137 Chr.16: 24559728-24559728 on Build GRCh38
rs934197010 Chr.16: 24559432-24559432 on Build GRCh38
rs951961272 Chr.16: 24559822-24559822 on Build GRCh38
rs775077836 Chr.16: 24559707-24559707 on Build GRCh38
rs557954841 Chr.16: 24559869-24559869 on Build GRCh38
rs569799648 Chr.16: 24559753-24559753 on Build GRCh38
rs769599576 Chr.16: 24559697-24559697 on Build GRCh38
rs773023307 Chr.16: 24559599-24559599 on Build GRCh38
rs780495188 Chr.16: 24559511-24559511 on Build GRCh38
rs200693893 Chr.16: 24559459-24559459 on Build GRCh38
rs377317418 Chr.16: 24559562-24559562 on Build GRCh38
rs990792682 Chr.16: 24559424-24559424 on Build GRCh38
rs199982748 Chr.16: 24559474-24559474 on Build GRCh38
rs576620769 Chr.16: 24559788-24559788 on Build GRCh38
rs145489419 Chr.16: 24559446-24559446 on Build GRCh38
rs567331195 Chr.16: 24559541-24559541 on Build GRCh38
rs747366243 Chr.16: 24559814-24559814 on Build GRCh38

View Full Product Details