Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 5517724-5518154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRP1
Entrez Gene ID
22861
Gene Name
NLR family pyrin domain containing 1
Gene Aliases
CARD7, CIDED, CLR17.1, DEFCAP, DEFCAP-L/S, NAC, NALP1, PP1044, SLEV1, VAMAS1
UniGene
Hs.652273
Gene Chromosome Location
Chr.17: 5501399-5584512 on Build GRCh38
Transcripts
NM_001033053.2 NM_014922.4 NM_033004.3 NM_033006.3 NM_033007.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6861593 c.4040A>T p.E1347V Chr.17: 5517763-5517763 on Build GRCh38
4707199 c.3963G>A p.S1321S Chr.17: 5517840-5517840 on Build GRCh38
6346566 c.3946C>A p.R1316R Chr.17: 5517869-5517869 on Build GRCh38
4707196 c.3973G>A p.V1325I Chr.17: 5517830-5517830 on Build GRCh38
4780128 c.4052A>G p.E1351G Chr.17: 5517763-5517763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749521716 Chr.17: 5517807-5517807 on Build GRCh38
rs199667505 Chr.17: 5517930-5517930 on Build GRCh38
rs7223605 Chr.17: 5518109-5518109 on Build GRCh38
rs567062857 Chr.17: 5518114-5518114 on Build GRCh38
rs144711602 Chr.17: 5517830-5517830 on Build GRCh38
rs953855584 Chr.17: 5517831-5517831 on Build GRCh38
rs201583715 Chr.17: 5517761-5517761 on Build GRCh38
rs965832435 Chr.17: 5517976-5517976 on Build GRCh38
rs756567141 Chr.17: 5517777-5517777 on Build GRCh38
rs757672601 Chr.17: 5517926-5517926 on Build GRCh38
rs530799056 Chr.17: 5517972-5517972 on Build GRCh38
rs771346182 Chr.17: 5517825-5517825 on Build GRCh38
rs748646281 Chr.17: 5517774-5517774 on Build GRCh38
rs765031318 Chr.17: 5517857-5517857 on Build GRCh38
rs147885770 Chr.17: 5517832-5517832 on Build GRCh38
rs551457068 Chr.17: 5517909-5517909 on Build GRCh38
rs761691866 Chr.17: 5517852-5517852 on Build GRCh38
rs1006611498 Chr.17: 5518129-5518129 on Build GRCh38
rs937466788 Chr.17: 5517975-5517975 on Build GRCh38
rs199808820 Chr.17: 5517861-5517861 on Build GRCh38
rs536450921 Chr.17: 5518083-5518083 on Build GRCh38
rs765377790 Chr.17: 5517868-5517868 on Build GRCh38
rs766081243 Chr.17: 5517866-5517866 on Build GRCh38
rs143021829 Chr.17: 5517962-5517962 on Build GRCh38
rs1012797266 Chr.17: 5517816-5517816 on Build GRCh38
rs201563922 Chr.17: 5517915-5517915 on Build GRCh38
rs1054851902 Chr.17: 5517977-5517977 on Build GRCh38
rs1026829338 Chr.17: 5517910-5517910 on Build GRCh38
rs998012544 Chr.17: 5518074-5518074 on Build GRCh38
rs567922162 Chr.17: 5518021-5518021 on Build GRCh38
rs924043737 Chr.17: 5518012-5518012 on Build GRCh38
rs902440753 Chr.17: 5518040-5518040 on Build GRCh38
rs367868204 Chr.17: 5518084-5518084 on Build GRCh38
rs552024507 Chr.17: 5518064-5518064 on Build GRCh38
rs141522655 Chr.17: 5517873-5517873 on Build GRCh38
rs778222806 Chr.17: 5517792-5517792 on Build GRCh38
rs774689496 Chr.17: 5517823-5517823 on Build GRCh38
rs766791576 Chr.17: 5517898-5517898 on Build GRCh38
rs1045630706 Chr.17: 5518131-5518131 on Build GRCh38
rs201385462 Chr.17: 5517858-5517858 on Build GRCh38
rs1020897098 Chr.17: 5517804-5517804 on Build GRCh38
rs7223443 Chr.17: 5517997-5517997 on Build GRCh38
rs902324390 Chr.17: 5517821-5517821 on Build GRCh38
rs1053796752 Chr.17: 5518069-5518069 on Build GRCh38
rs772159137 Chr.17: 5517836-5517836 on Build GRCh38
rs1010853454 Chr.17: 5517785-5517785 on Build GRCh38
rs754489472 Chr.17: 5517869-5517869 on Build GRCh38
rs1054019727 Chr.17: 5517784-5517784 on Build GRCh38
rs749727136 Chr.17: 5517916-5517916 on Build GRCh38
rs893108367 Chr.17: 5517982-5517982 on Build GRCh38
rs199687052 Chr.17: 5517829-5517829 on Build GRCh38
rs62072728 Chr.17: 5517896-5517896 on Build GRCh38
rs776467398 Chr.17: 5517840-5517840 on Build GRCh38
rs200713360 Chr.17: 5517881-5517881 on Build GRCh38

View Full Product Details