Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33299610-33300106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF362
Entrez Gene ID
149076
Gene Name
zinc finger protein 362
Gene Aliases
RN, lin-29
UniGene
Hs.743358
Gene Chromosome Location
Chr.1: 33256307-33300719 on Build GRCh38
Transcripts
NM_152493.2 XM_005270507.2 XM_005270508.4 XM_017000415.1

SNP Detail

SNP ID Chromosome Location
rs939935788 Chr.1: 33299836-33299836 on Build GRCh38
rs1048822832 Chr.1: 33299654-33299654 on Build GRCh38
rs886207111 Chr.1: 33299670-33299670 on Build GRCh38
rs1021225916 Chr.1: 33300021-33300021 on Build GRCh38
rs1004259383 Chr.1: 33299632-33299632 on Build GRCh38
rs976798990 Chr.1: 33299774-33299774 on Build GRCh38
rs958327797 Chr.1: 33299734-33299734 on Build GRCh38
rs542098129 Chr.1: 33299677-33299677 on Build GRCh38
rs932710802 Chr.1: 33300047-33300047 on Build GRCh38
rs1000674927 Chr.1: 33299730-33299730 on Build GRCh38
rs1048240303 Chr.1: 33299812-33299812 on Build GRCh38
rs796890820 Chr.1: 33299692-33299692 on Build GRCh38
rs905638265 Chr.1: 33299926-33299926 on Build GRCh38
rs980908685 Chr.1: 33299695-33299695 on Build GRCh38
rs932452205 Chr.1: 33299810-33299810 on Build GRCh38
rs1023537698 Chr.1: 33299696-33299696 on Build GRCh38
rs76058787 Chr.1: 33299803-33299803 on Build GRCh38
rs532719470 Chr.1: 33299744-33299744 on Build GRCh38
rs975647134 Chr.1: 33299857-33299857 on Build GRCh38
rs375130127 Chr.1: 33299729-33299729 on Build GRCh38
rs920081155 Chr.1: 33299630-33299630 on Build GRCh38
rs958487961 Chr.1: 33300004-33300004 on Build GRCh38
rs34615319 Chr.1: 33299767-33299768 on Build GRCh38
rs1012158201 Chr.1: 33299681-33299681 on Build GRCh38
rs1056651098 Chr.1: 33300022-33300022 on Build GRCh38
rs1132789 Chr.1: 33299676-33299676 on Build GRCh38
rs988404108 Chr.1: 33300019-33300019 on Build GRCh38
rs888308281 Chr.1: 33299833-33299833 on Build GRCh38
rs548954053 Chr.1: 33299811-33299811 on Build GRCh38
rs920197306 Chr.1: 33299870-33299870 on Build GRCh38
rs1022627767 Chr.1: 33299915-33299915 on Build GRCh38
rs1037394504 Chr.1: 33299855-33299855 on Build GRCh38
rs896140796 Chr.1: 33300053-33300053 on Build GRCh38
rs982960861 Chr.1: 33299950-33299950 on Build GRCh38
rs571395609 Chr.1: 33300035-33300035 on Build GRCh38
rs1132790 Chr.1: 33299671-33299671 on Build GRCh38
rs931496720 Chr.1: 33299651-33299651 on Build GRCh38
rs566564831 Chr.1: 33299799-33299799 on Build GRCh38
rs556398707 Chr.1: 33299986-33299986 on Build GRCh38
rs921322956 Chr.1: 33300032-33300032 on Build GRCh38
rs537718393 Chr.1: 33299926-33299926 on Build GRCh38
rs1013881841 Chr.1: 33299902-33299902 on Build GRCh38
rs987168783 Chr.1: 33299773-33299773 on Build GRCh38
rs568494914 Chr.1: 33299752-33299752 on Build GRCh38
rs567575557 Chr.1: 33299835-33299835 on Build GRCh38
rs984637794 Chr.1: 33300052-33300052 on Build GRCh38
rs931444413 Chr.1: 33299928-33299928 on Build GRCh38
rs1032497223 Chr.1: 33299999-33299999 on Build GRCh38
rs913902286 Chr.1: 33299758-33299758 on Build GRCh38
rs907582242 Chr.1: 33299979-33299979 on Build GRCh38

View Full Product Details