Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58272656-58273161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPO
Entrez Gene ID
4353
Gene Name
myeloperoxidase
Gene Aliases
-
UniGene
Hs.458272
Gene Chromosome Location
Chr.17: 58269856-58280935 on Build GRCh38
Transcripts
NM_000250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
417431 c.1744G>C p.D582H Chr.17: 58272796-58272796 on Build GRCh38
4796356 c.1885C>T p.P629S Chr.17: 58272751-58272751 on Build GRCh38
3195273 c.1880G>T p.G627V Chr.17: 58272756-58272756 on Build GRCh38
4068118 c.1735C>T p.L579F Chr.17: 58272901-58272901 on Build GRCh38
7140383 c.1671G>T p.L557L Chr.17: 58272869-58272869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948122932 Chr.17: 58272698-58272698 on Build GRCh38
rs749111097 Chr.17: 58272921-58272921 on Build GRCh38
rs979180081 Chr.17: 58272678-58272678 on Build GRCh38
rs781130824 Chr.17: 58272818-58272818 on Build GRCh38
rs776822724 Chr.17: 58272952-58272952 on Build GRCh38
rs533996917 Chr.17: 58273047-58273047 on Build GRCh38
rs119469012 Chr.17: 58272825-58272825 on Build GRCh38
rs554451056 Chr.17: 58272998-58272998 on Build GRCh38
rs1038102422 Chr.17: 58273114-58273114 on Build GRCh38
rs767608651 Chr.17: 58273141-58273141 on Build GRCh38
rs35400724 Chr.17: 58273136-58273136 on Build GRCh38
rs745917861 Chr.17: 58272931-58272931 on Build GRCh38
rs543855522 Chr.17: 58272681-58272681 on Build GRCh38
rs773353816 Chr.17: 58272726-58272726 on Build GRCh38
rs777372931 Chr.17: 58272912-58272912 on Build GRCh38
rs537308789 Chr.17: 58273111-58273113 on Build GRCh38
rs768084345 Chr.17: 58272843-58272843 on Build GRCh38
rs768139578 Chr.17: 58272764-58272764 on Build GRCh38
rs754533715 Chr.17: 58272883-58272883 on Build GRCh38
rs779296131 Chr.17: 58272926-58272926 on Build GRCh38
rs562564787 Chr.17: 58272689-58272689 on Build GRCh38
rs759716007 Chr.17: 58272836-58272836 on Build GRCh38
rs763057805 Chr.17: 58272731-58272731 on Build GRCh38
rs200188979 Chr.17: 58272915-58272915 on Build GRCh38
rs770010897 Chr.17: 58272702-58272702 on Build GRCh38
rs775905828 Chr.17: 58272845-58272845 on Build GRCh38
rs759041226 Chr.17: 58272962-58272962 on Build GRCh38
rs119468010 Chr.17: 58272835-58272835 on Build GRCh38
rs918073785 Chr.17: 58273083-58273083 on Build GRCh38
rs774984207 Chr.17: 58272757-58272757 on Build GRCh38
rs938146845 Chr.17: 58272714-58272714 on Build GRCh38
rs368720980 Chr.17: 58272922-58272922 on Build GRCh38
rs577253275 Chr.17: 58273130-58273130 on Build GRCh38
rs771235998 Chr.17: 58272925-58272925 on Build GRCh38
rs992399931 Chr.17: 58273100-58273100 on Build GRCh38
rs959112827 Chr.17: 58273043-58273043 on Build GRCh38
rs949631117 Chr.17: 58272809-58272809 on Build GRCh38
rs762528347 Chr.17: 58272865-58272865 on Build GRCh38
rs1024923921 Chr.17: 58273102-58273102 on Build GRCh38
rs764655809 Chr.17: 58272703-58272703 on Build GRCh38
rs746527352 Chr.17: 58272811-58272811 on Build GRCh38
rs142082076 Chr.17: 58272772-58272772 on Build GRCh38
rs769213195 Chr.17: 58272949-58272949 on Build GRCh38
rs750114243 Chr.17: 58272779-58272779 on Build GRCh38
rs751022705 Chr.17: 58272881-58272881 on Build GRCh38
rs765994933 Chr.17: 58272954-58272954 on Build GRCh38
rs772348166 Chr.17: 58272939-58272939 on Build GRCh38
rs969457593 Chr.17: 58273109-58273109 on Build GRCh38
rs546861952 Chr.17: 58272886-58272886 on Build GRCh38
rs34471705 Chr.17: 58273015-58273015 on Build GRCh38
rs773478778 Chr.17: 58272829-58272829 on Build GRCh38
rs764426896 Chr.17: 58272864-58272864 on Build GRCh38
rs144371238 Chr.17: 58272897-58272897 on Build GRCh38
rs138764204 Chr.17: 58272758-58272758 on Build GRCh38
rs779484560 Chr.17: 58272804-58272804 on Build GRCh38
rs200618562 Chr.17: 58272904-58272904 on Build GRCh38
rs751254636 Chr.17: 58272957-58272957 on Build GRCh38
rs888814317 Chr.17: 58272892-58272892 on Build GRCh38
rs769040237 Chr.17: 58272707-58272708 on Build GRCh38
rs558852664 Chr.17: 58273072-58273072 on Build GRCh38
rs747927146 Chr.17: 58272697-58272697 on Build GRCh38
rs549292089 Chr.17: 58272820-58272820 on Build GRCh38
rs529845670 Chr.17: 58272748-58272748 on Build GRCh38
rs769515999 Chr.17: 58272828-58272828 on Build GRCh38
rs996299337 Chr.17: 58272895-58272895 on Build GRCh38
rs370125571 Chr.17: 58272953-58272953 on Build GRCh38
rs761032711 Chr.17: 58272948-58272948 on Build GRCh38
rs148802625 Chr.17: 58272898-58272898 on Build GRCh38
rs151004081 Chr.17: 58272943-58272943 on Build GRCh38
rs990564759 Chr.17: 58273063-58273063 on Build GRCh38
rs754871401 Chr.17: 58272817-58272817 on Build GRCh38
rs766512647 Chr.17: 58273077-58273077 on Build GRCh38
rs146363806 Chr.17: 58272830-58272830 on Build GRCh38
rs571611976 Chr.17: 58272889-58272889 on Build GRCh38
rs956935061 Chr.17: 58273096-58273096 on Build GRCh38
rs753062060 Chr.17: 58272771-58272771 on Build GRCh38
rs751679813 Chr.17: 58273131-58273131 on Build GRCh38
rs766054225 Chr.17: 58272879-58272879 on Build GRCh38
rs139720381 Chr.17: 58272834-58272834 on Build GRCh38
rs1051409286 Chr.17: 58272860-58272860 on Build GRCh38
rs968874158 Chr.17: 58272985-58272985 on Build GRCh38
rs761101621 Chr.17: 58272854-58272854 on Build GRCh38
rs909038610 Chr.17: 58272821-58272821 on Build GRCh38
rs757914812 Chr.17: 58272802-58272802 on Build GRCh38

View Full Product Details