Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78120562-78121056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMC6
Entrez Gene ID
11322
Gene Name
transmembrane channel like 6
Gene Aliases
EV1, EVER1, EVIN1, LAK-4P
UniGene
Hs.632227
Gene Chromosome Location
Chr.17: 78108775-78132427 on Build GRCh38
Transcripts
NM_001127198.2 NM_001321185.1 NM_007267.7 XM_005256995.1 XM_011524255.1 XM_011524256.1 XM_011524257.2 XM_011524258.1 XM_017024107.1 XM_017024108.1 XM_017024109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7188804 c.1552C>A p.L518M Chr.17: 78120816-78120816 on Build GRCh38
3421944 c.1592G>A p.R531H Chr.17: 78120776-78120776 on Build GRCh38
437650 c.1671C>T p.F557F Chr.17: 78120697-78120697 on Build GRCh38
4447329 c.1715+3G>A p.? Chr.17: 78120650-78120650 on Build GRCh38
6279057 c.824C>T p.P275L Chr.17: 78121043-78121043 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546888861 Chr.17: 78121003-78121003 on Build GRCh38
rs957525623 Chr.17: 78120584-78120584 on Build GRCh38
rs749031381 Chr.17: 78120990-78120990 on Build GRCh38
rs763923856 Chr.17: 78120616-78120616 on Build GRCh38
rs139049167 Chr.17: 78120777-78120777 on Build GRCh38
rs748258392 Chr.17: 78120968-78120968 on Build GRCh38
rs766212767 Chr.17: 78120865-78120865 on Build GRCh38
rs759497226 Chr.17: 78121002-78121002 on Build GRCh38
rs754592361 Chr.17: 78120815-78120815 on Build GRCh38
rs763643231 Chr.17: 78121007-78121007 on Build GRCh38
rs780536424 Chr.17: 78120949-78120949 on Build GRCh38
rs751047936 Chr.17: 78120807-78120807 on Build GRCh38
rs767047782 Chr.17: 78120811-78120811 on Build GRCh38
rs749657326 Chr.17: 78120754-78120754 on Build GRCh38
rs765210208 Chr.17: 78120697-78120697 on Build GRCh38
rs141464477 Chr.17: 78120760-78120760 on Build GRCh38
rs779481795 Chr.17: 78120717-78120717 on Build GRCh38
rs752435878 Chr.17: 78120823-78120823 on Build GRCh38
rs757672687 Chr.17: 78120830-78120830 on Build GRCh38
rs746682799 Chr.17: 78120649-78120649 on Build GRCh38
rs766782993 Chr.17: 78121021-78121021 on Build GRCh38
rs368590817 Chr.17: 78120669-78120669 on Build GRCh38
rs748914621 Chr.17: 78120953-78120954 on Build GRCh38
rs755793681 Chr.17: 78120608-78120609 on Build GRCh38
rs370943130 Chr.17: 78120860-78120860 on Build GRCh38
rs752700395 Chr.17: 78120614-78120614 on Build GRCh38
rs750398210 Chr.17: 78120712-78120712 on Build GRCh38
rs772844733 Chr.17: 78120862-78120862 on Build GRCh38
rs537200296 Chr.17: 78120716-78120716 on Build GRCh38
rs780986633 Chr.17: 78120821-78120821 on Build GRCh38
rs750353199 Chr.17: 78120778-78120778 on Build GRCh38
rs762812716 Chr.17: 78120864-78120864 on Build GRCh38
rs369378117 Chr.17: 78120886-78120886 on Build GRCh38
rs117553598 Chr.17: 78120836-78120836 on Build GRCh38
rs759547569 Chr.17: 78120610-78120610 on Build GRCh38
rs35860746 Chr.17: 78120893-78120893 on Build GRCh38
rs142835344 Chr.17: 78120844-78120844 on Build GRCh38
rs201780305 Chr.17: 78120837-78120837 on Build GRCh38
rs370791917 Chr.17: 78120979-78120979 on Build GRCh38
rs111907805 Chr.17: 78120920-78120920 on Build GRCh38
rs754637431 Chr.17: 78120751-78120751 on Build GRCh38
rs371220608 Chr.17: 78120617-78120617 on Build GRCh38
rs778830451 Chr.17: 78120635-78120635 on Build GRCh38
rs755678139 Chr.17: 78120937-78120937 on Build GRCh38
rs915001402 Chr.17: 78120895-78120895 on Build GRCh38
rs775756280 Chr.17: 78120677-78120677 on Build GRCh38
rs768811941 Chr.17: 78120770-78120770 on Build GRCh38
rs752479813 Chr.17: 78120607-78120608 on Build GRCh38
rs376404481 Chr.17: 78121018-78121018 on Build GRCh38
rs746650669 Chr.17: 78120834-78120834 on Build GRCh38
rs778235026 Chr.17: 78120658-78120658 on Build GRCh38
rs146998467 Chr.17: 78120776-78120776 on Build GRCh38
rs779755857 Chr.17: 78120647-78120647 on Build GRCh38
rs764327372 Chr.17: 78120688-78120688 on Build GRCh38
rs759085535 Chr.17: 78120921-78120921 on Build GRCh38
rs780908976 Chr.17: 78120602-78120608 on Build GRCh38
rs757818144 Chr.17: 78120670-78120670 on Build GRCh38
rs948686171 Chr.17: 78120957-78120957 on Build GRCh38
rs759655248 Chr.17: 78120674-78120674 on Build GRCh38
rs781004703 Chr.17: 78120752-78120752 on Build GRCh38
rs757130256 Chr.17: 78120895-78120896 on Build GRCh38
rs747559259 Chr.17: 78120950-78120950 on Build GRCh38
rs71931149 Chr.17: 78120595-78120607 on Build GRCh38
rs376804045 Chr.17: 78120602-78120602 on Build GRCh38
rs373929867 Chr.17: 78121004-78121004 on Build GRCh38
rs751102312 Chr.17: 78120745-78120745 on Build GRCh38
rs767184349 Chr.17: 78120922-78120922 on Build GRCh38
rs951048399 Chr.17: 78120854-78120854 on Build GRCh38
rs201479637 Chr.17: 78120763-78120763 on Build GRCh38
rs774407923 Chr.17: 78120998-78120998 on Build GRCh38
rs964766376 Chr.17: 78120872-78120872 on Build GRCh38
rs755904758 Chr.17: 78120615-78120615 on Build GRCh38
rs962971490 Chr.17: 78120934-78120934 on Build GRCh38
rs368155695 Chr.17: 78120870-78120870 on Build GRCh38
rs552965413 Chr.17: 78121027-78121027 on Build GRCh38
rs1030046325 Chr.17: 78120889-78120889 on Build GRCh38
rs777395340 Chr.17: 78120765-78120765 on Build GRCh38
rs776900537 Chr.17: 78120772-78120772 on Build GRCh38
rs758858152 Chr.17: 78120946-78120946 on Build GRCh38
rs202229397 Chr.17: 78120597-78120597 on Build GRCh38
rs760833588 Chr.17: 78120681-78120681 on Build GRCh38
rs753596652 Chr.17: 78121011-78121011 on Build GRCh38
rs781200935 Chr.17: 78120952-78120952 on Build GRCh38
rs766371395 Chr.17: 78120789-78120789 on Build GRCh38
rs749702427 Chr.17: 78120667-78120667 on Build GRCh38
rs146977134 Chr.17: 78120907-78120907 on Build GRCh38
rs963338971 Chr.17: 78120844-78120844 on Build GRCh38
rs759084047 Chr.17: 78120589-78120589 on Build GRCh38
rs111815200 Chr.17: 78120585-78120597 on Build GRCh38
rs770798041 Chr.17: 78120997-78120997 on Build GRCh38
rs748419745 Chr.17: 78120849-78120849 on Build GRCh38
rs752928727 Chr.17: 78120612-78120612 on Build GRCh38
rs775388787 Chr.17: 78120766-78120766 on Build GRCh38
rs758762407 Chr.17: 78120842-78120842 on Build GRCh38
rs755438439 Chr.17: 78121028-78121028 on Build GRCh38
rs772083222 Chr.17: 78120676-78120676 on Build GRCh38
rs908757659 Chr.17: 78120695-78120695 on Build GRCh38
rs767765175 Chr.17: 78120611-78120611 on Build GRCh38
rs770263060 Chr.17: 78120652-78120652 on Build GRCh38
rs989804477 Chr.17: 78120596-78120596 on Build GRCh38
rs773499019 Chr.17: 78120984-78120984 on Build GRCh38
rs564306887 Chr.17: 78120940-78120940 on Build GRCh38
rs375082957 Chr.17: 78120867-78120867 on Build GRCh38
rs371778724 Chr.17: 78120845-78120845 on Build GRCh38
rs367991714 Chr.17: 78120843-78120843 on Build GRCh38
rs183447230 Chr.17: 78120951-78120951 on Build GRCh38
rs757906231 Chr.17: 78120644-78120644 on Build GRCh38
rs777356090 Chr.17: 78120644-78120644 on Build GRCh38
rs757142728 Chr.17: 78120630-78120630 on Build GRCh38
rs199724291 Chr.17: 78120696-78120696 on Build GRCh38
rs754941049 Chr.17: 78120601-78120608 on Build GRCh38
rs753566703 Chr.17: 78120692-78120692 on Build GRCh38
rs375990147 Chr.17: 78120636-78120636 on Build GRCh38

View Full Product Details