Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36097508-36098002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL8A2
Entrez Gene ID
1296
Gene Name
collagen type VIII alpha 2
Gene Aliases
FECD, FECD1, PPCD, PPCD2
UniGene
Hs.353001
Gene Chromosome Location
Chr.1: 36095236-36126207 on Build GRCh38
Transcripts
NM_001294347.1 NM_005202.3 XM_005270477.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5120554 c.2063C>T p.T688M Chr.1: 36097618-36097618 on Build GRCh38
5745612 c.2033C>T p.P678L Chr.1: 36097648-36097648 on Build GRCh38
4476564 c.2078C>T p.S693F Chr.1: 36097603-36097603 on Build GRCh38
128690 c.1682_1683insA p.P562fs*34 Chr.1: 36097998-36097999 on Build GRCh38
908827 c.2069A>G p.Y690C Chr.1: 36097612-36097612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029952617 Chr.1: 36097736-36097736 on Build GRCh38
rs374892980 Chr.1: 36097737-36097737 on Build GRCh38
rs772492757 Chr.1: 36097713-36097713 on Build GRCh38
rs769252088 Chr.1: 36097856-36097856 on Build GRCh38
rs778792447 Chr.1: 36097607-36097607 on Build GRCh38
rs755678047 Chr.1: 36097956-36097956 on Build GRCh38
rs768247177 Chr.1: 36097827-36097827 on Build GRCh38
rs758485853 Chr.1: 36097937-36097937 on Build GRCh38
rs201587529 Chr.1: 36097679-36097679 on Build GRCh38
rs756900371 Chr.1: 36097556-36097557 on Build GRCh38
rs534995976 Chr.1: 36097761-36097761 on Build GRCh38
rs891447843 Chr.1: 36097533-36097533 on Build GRCh38
rs761981458 Chr.1: 36097932-36097932 on Build GRCh38
rs745843219 Chr.1: 36097980-36097980 on Build GRCh38
rs776891661 Chr.1: 36097931-36097931 on Build GRCh38
rs764312183 Chr.1: 36097864-36097864 on Build GRCh38
rs200767854 Chr.1: 36097747-36097747 on Build GRCh38
rs774743866 Chr.1: 36097644-36097644 on Build GRCh38
rs768122602 Chr.1: 36097746-36097746 on Build GRCh38
rs780073352 Chr.1: 36097543-36097543 on Build GRCh38
rs755677174 Chr.1: 36097566-36097566 on Build GRCh38
rs138158024 Chr.1: 36097786-36097786 on Build GRCh38
rs755299937 Chr.1: 36097881-36097881 on Build GRCh38
rs867531302 Chr.1: 36097910-36097910 on Build GRCh38
rs777087120 Chr.1: 36097740-36097740 on Build GRCh38
rs766331675 Chr.1: 36097757-36097757 on Build GRCh38
rs774419881 Chr.1: 36097979-36097979 on Build GRCh38
rs771423414 Chr.1: 36097695-36097695 on Build GRCh38
rs780889462 Chr.1: 36097884-36097884 on Build GRCh38
rs374316118 Chr.1: 36097894-36097894 on Build GRCh38
rs770404305 Chr.1: 36097561-36097561 on Build GRCh38
rs148241498 Chr.1: 36097944-36097944 on Build GRCh38
rs980314273 Chr.1: 36097965-36097965 on Build GRCh38
rs928574991 Chr.1: 36097586-36097586 on Build GRCh38
rs767141059 Chr.1: 36097647-36097647 on Build GRCh38
rs749820012 Chr.1: 36097632-36097632 on Build GRCh38
rs146876759 Chr.1: 36097948-36097948 on Build GRCh38
rs751354492 Chr.1: 36097860-36097860 on Build GRCh38
rs749361746 Chr.1: 36097692-36097692 on Build GRCh38
rs149251273 Chr.1: 36097866-36097866 on Build GRCh38
rs200185433 Chr.1: 36097799-36097799 on Build GRCh38
rs758861286 Chr.1: 36097877-36097877 on Build GRCh38
rs374239481 Chr.1: 36097617-36097617 on Build GRCh38
rs974128746 Chr.1: 36097722-36097722 on Build GRCh38
rs142307403 Chr.1: 36097982-36097982 on Build GRCh38
rs753778442 Chr.1: 36097581-36097581 on Build GRCh38
rs190071119 Chr.1: 36097953-36097953 on Build GRCh38
rs765117541 Chr.1: 36097672-36097672 on Build GRCh38
rs561961321 Chr.1: 36097704-36097704 on Build GRCh38
rs778043170 Chr.1: 36097691-36097691 on Build GRCh38
rs140015576 Chr.1: 36097917-36097917 on Build GRCh38
rs776159463 Chr.1: 36097830-36097830 on Build GRCh38
rs775189078 Chr.1: 36097648-36097648 on Build GRCh38
rs146313416 Chr.1: 36097671-36097671 on Build GRCh38
rs752124675 Chr.1: 36097945-36097945 on Build GRCh38
rs991834696 Chr.1: 36097829-36097829 on Build GRCh38
rs373062066 Chr.1: 36097535-36097535 on Build GRCh38
rs756944002 Chr.1: 36097600-36097600 on Build GRCh38
rs761249667 Chr.1: 36097750-36097750 on Build GRCh38
rs373128364 Chr.1: 36097947-36097947 on Build GRCh38
rs371369693 Chr.1: 36097689-36097689 on Build GRCh38
rs914457455 Chr.1: 36097843-36097843 on Build GRCh38
rs3738360 Chr.1: 36097557-36097557 on Build GRCh38
rs753428477 Chr.1: 36097556-36097556 on Build GRCh38
rs981464774 Chr.1: 36097803-36097803 on Build GRCh38
rs145553904 Chr.1: 36097957-36097957 on Build GRCh38
rs752940687 Chr.1: 36097758-36097758 on Build GRCh38
rs997174521 Chr.1: 36097709-36097709 on Build GRCh38
rs989021812 Chr.1: 36097981-36097981 on Build GRCh38
rs866697723 Chr.1: 36097670-36097670 on Build GRCh38
rs761318308 Chr.1: 36097840-36097840 on Build GRCh38
rs773184010 Chr.1: 36097858-36097858 on Build GRCh38
rs35366412 Chr.1: 36097697-36097698 on Build GRCh38
rs780560468 Chr.1: 36097879-36097879 on Build GRCh38
rs770069044 Chr.1: 36097628-36097628 on Build GRCh38
rs747161067 Chr.1: 36097609-36097609 on Build GRCh38
rs759864363 Chr.1: 36097645-36097645 on Build GRCh38
rs765161350 Chr.1: 36097752-36097752 on Build GRCh38
rs199954041 Chr.1: 36097662-36097662 on Build GRCh38
rs1011646582 Chr.1: 36097844-36097844 on Build GRCh38
rs763029689 Chr.1: 36097859-36097859 on Build GRCh38
rs752179559 Chr.1: 36097565-36097565 on Build GRCh38
rs560249177 Chr.1: 36097608-36097608 on Build GRCh38
rs1042743559 Chr.1: 36097845-36097845 on Build GRCh38
rs149090218 Chr.1: 36097935-36097935 on Build GRCh38
rs772560697 Chr.1: 36097908-36097908 on Build GRCh38
rs1041453380 Chr.1: 36097677-36097677 on Build GRCh38
rs147063250 Chr.1: 36097933-36097933 on Build GRCh38
rs938886783 Chr.1: 36097792-36097792 on Build GRCh38
rs775111340 Chr.1: 36097731-36097731 on Build GRCh38
rs201409639 Chr.1: 36097800-36097800 on Build GRCh38
rs141682102 Chr.1: 36097678-36097678 on Build GRCh38
rs550694898 Chr.1: 36097753-36097753 on Build GRCh38
rs762026636 Chr.1: 36097558-36097558 on Build GRCh38
rs35841184 Chr.1: 36097923-36097923 on Build GRCh38
rs149180733 Chr.1: 36097564-36097564 on Build GRCh38
rs567011408 Chr.1: 36097821-36097821 on Build GRCh38
rs753899490 Chr.1: 36097788-36097788 on Build GRCh38
rs776780878 Chr.1: 36097785-36097785 on Build GRCh38
rs766775820 Chr.1: 36097563-36097563 on Build GRCh38
rs774436601 Chr.1: 36097918-36097918 on Build GRCh38
rs928614322 Chr.1: 36097805-36097805 on Build GRCh38
rs769321257 Chr.1: 36097907-36097907 on Build GRCh38
rs920141328 Chr.1: 36097734-36097734 on Build GRCh38
rs150169820 Chr.1: 36097826-36097826 on Build GRCh38
rs771901419 Chr.1: 36097611-36097611 on Build GRCh38
rs770635015 Chr.1: 36097967-36097967 on Build GRCh38
rs772403294 Chr.1: 36097809-36097809 on Build GRCh38
rs745640221 Chr.1: 36097780-36097781 on Build GRCh38
rs938833154 Chr.1: 36097977-36097977 on Build GRCh38
rs760110124 Chr.1: 36097649-36097649 on Build GRCh38
rs762834432 Chr.1: 36097755-36097755 on Build GRCh38
rs754076571 Chr.1: 36097865-36097865 on Build GRCh38
rs747313752 Chr.1: 36097880-36097880 on Build GRCh38
rs770824891 Chr.1: 36097537-36097537 on Build GRCh38
rs201628588 Chr.1: 36097555-36097556 on Build GRCh38
rs375533913 Chr.1: 36097791-36097791 on Build GRCh38
rs764013903 Chr.1: 36097578-36097578 on Build GRCh38
rs763607342 Chr.1: 36097661-36097661 on Build GRCh38
rs142904531 Chr.1: 36097914-36097914 on Build GRCh38
rs139081156 Chr.1: 36097872-36097872 on Build GRCh38
rs748743889 Chr.1: 36097966-36097966 on Build GRCh38
rs759403098 Chr.1: 36097922-36097922 on Build GRCh38
rs747269126 Chr.1: 36097546-36097546 on Build GRCh38
rs748443606 Chr.1: 36097688-36097688 on Build GRCh38
rs759421533 Chr.1: 36097863-36097863 on Build GRCh38
rs750898608 Chr.1: 36097873-36097873 on Build GRCh38
rs763678155 Chr.1: 36097558-36097559 on Build GRCh38
rs746179003 Chr.1: 36097707-36097707 on Build GRCh38
rs961674103 Chr.1: 36097711-36097711 on Build GRCh38

View Full Product Details