Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81651523-81652025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE6G
Entrez Gene ID
5148
Gene Name
phosphodiesterase 6G
Gene Aliases
PDEG, RP57
UniGene
Hs.654482
Gene Chromosome Location
Chr.17: 81650459-81663418 on Build GRCh38
Transcripts
NM_002602.3 NR_026872.1 XM_006721942.3 XM_017024734.1 XM_017024735.1 XM_017024736.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1236263 c.172G>C p.E58Q Chr.17: 81651660-81651660 on Build GRCh38
7168813 c.157G>A p.D53N Chr.17: 81651675-81651675 on Build GRCh38
5768287 c.187+5G>C p.? Chr.17: 81651640-81651640 on Build GRCh38
4910629 c.174A>G p.E58E Chr.17: 81651658-81651658 on Build GRCh38
2805364 c.147-1G>T p.? Chr.17: 81651686-81651686 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773145285 Chr.17: 81651679-81651679 on Build GRCh38
rs961635324 Chr.17: 81651677-81651677 on Build GRCh38
rs182193334 Chr.17: 81651869-81651869 on Build GRCh38
rs754032199 Chr.17: 81651622-81651622 on Build GRCh38
rs11871837 Chr.17: 81651952-81651952 on Build GRCh38
rs183900269 Chr.17: 81651548-81651548 on Build GRCh38
rs12603868 Chr.17: 81651804-81651804 on Build GRCh38
rs760726888 Chr.17: 81651598-81651599 on Build GRCh38
rs773923808 Chr.17: 81651669-81651669 on Build GRCh38
rs749474428 Chr.17: 81651637-81651637 on Build GRCh38
rs775603283 Chr.17: 81651939-81651939 on Build GRCh38
rs911488036 Chr.17: 81651963-81651963 on Build GRCh38
rs757861494 Chr.17: 81651549-81651549 on Build GRCh38
rs1054874378 Chr.17: 81651860-81651860 on Build GRCh38
rs945150360 Chr.17: 81651993-81651993 on Build GRCh38
rs766494400 Chr.17: 81651694-81651694 on Build GRCh38
rs765454894 Chr.17: 81651715-81651715 on Build GRCh38
rs922822167 Chr.17: 81651720-81651720 on Build GRCh38
rs753710064 Chr.17: 81651701-81651703 on Build GRCh38
rs978391146 Chr.17: 81651877-81651877 on Build GRCh38
rs566473438 Chr.17: 81651550-81651550 on Build GRCh38
rs752889943 Chr.17: 81651716-81651716 on Build GRCh38
rs757755619 Chr.17: 81651737-81651737 on Build GRCh38
rs1044261015 Chr.17: 81651944-81651944 on Build GRCh38
rs759743449 Chr.17: 81651713-81651713 on Build GRCh38
rs906440005 Chr.17: 81651842-81651842 on Build GRCh38
rs189399954 Chr.17: 81651593-81651593 on Build GRCh38
rs775431906 Chr.17: 81651599-81651599 on Build GRCh38
rs749465692 Chr.17: 81651929-81651929 on Build GRCh38
rs1003184074 Chr.17: 81651946-81651946 on Build GRCh38
rs775250199 Chr.17: 81651717-81651717 on Build GRCh38
rs763919364 Chr.17: 81651613-81651614 on Build GRCh38
rs747473734 Chr.17: 81651645-81651645 on Build GRCh38
rs985746764 Chr.17: 81651957-81651957 on Build GRCh38
rs533764417 Chr.17: 81651557-81651557 on Build GRCh38
rs372592115 Chr.17: 81651657-81651657 on Build GRCh38
rs893498688 Chr.17: 81651576-81651576 on Build GRCh38
rs769489406 Chr.17: 81651830-81651830 on Build GRCh38
rs558623992 Chr.17: 81651579-81651579 on Build GRCh38
rs1005386682 Chr.17: 81651888-81651888 on Build GRCh38
rs117011954 Chr.17: 81651676-81651676 on Build GRCh38
rs914325806 Chr.17: 81651905-81651905 on Build GRCh38
rs377261644 Chr.17: 81651717-81651717 on Build GRCh38
rs1054171586 Chr.17: 81651901-81651901 on Build GRCh38
rs754875390 Chr.17: 81651793-81651793 on Build GRCh38
rs1053456529 Chr.17: 81651560-81651560 on Build GRCh38
rs772282729 Chr.17: 81651595-81651596 on Build GRCh38
rs577479782 Chr.17: 81651928-81651928 on Build GRCh38
rs144191975 Chr.17: 81651675-81651675 on Build GRCh38
rs769669512 Chr.17: 81651551-81651551 on Build GRCh38
rs146449791 Chr.17: 81651813-81651813 on Build GRCh38
rs902877500 Chr.17: 81651626-81651626 on Build GRCh38
rs751976719 Chr.17: 81651724-81651724 on Build GRCh38
rs368835071 Chr.17: 81651620-81651620 on Build GRCh38
rs201820039 Chr.17: 81651596-81651596 on Build GRCh38
rs945022160 Chr.17: 81651820-81651820 on Build GRCh38
rs185585153 Chr.17: 81651968-81651968 on Build GRCh38
rs1016280914 Chr.17: 81651965-81651965 on Build GRCh38
rs753086874 Chr.17: 81651636-81651636 on Build GRCh38
rs1006182899 Chr.17: 81651956-81651956 on Build GRCh38
rs564094328 Chr.17: 81651781-81651781 on Build GRCh38
rs1042065218 Chr.17: 81651823-81651823 on Build GRCh38
rs780279422 Chr.17: 81651596-81651605 on Build GRCh38
rs570430187 Chr.17: 81651978-81651978 on Build GRCh38
rs924270419 Chr.17: 81651882-81651882 on Build GRCh38
rs779537639 Chr.17: 81651553-81651553 on Build GRCh38
rs994156852 Chr.17: 81651945-81651945 on Build GRCh38
rs1002845011 Chr.17: 81651810-81651810 on Build GRCh38
rs911419005 Chr.17: 81651778-81651778 on Build GRCh38
rs764376029 Chr.17: 81651721-81651721 on Build GRCh38
rs867563610 Chr.17: 81651962-81651962 on Build GRCh38
rs375443020 Chr.17: 81651600-81651600 on Build GRCh38
rs542089493 Chr.17: 81651629-81651629 on Build GRCh38
rs527467431 Chr.17: 81651584-81651584 on Build GRCh38
rs115998357 Chr.17: 81651814-81651814 on Build GRCh38
rs750359525 Chr.17: 81651603-81651603 on Build GRCh38
rs368718418 Chr.17: 81651968-81651969 on Build GRCh38
rs1026372897 Chr.17: 81651558-81651558 on Build GRCh38
rs990354092 Chr.17: 81651969-81651969 on Build GRCh38
rs924224099 Chr.17: 81651979-81651979 on Build GRCh38
rs760677354 Chr.17: 81651692-81651692 on Build GRCh38
rs774995002 Chr.17: 81651750-81651750 on Build GRCh38
rs934378648 Chr.17: 81651895-81651895 on Build GRCh38
rs139487034 Chr.17: 81651665-81651665 on Build GRCh38
rs200581539 Chr.17: 81651638-81651638 on Build GRCh38
rs760771444 Chr.17: 81651611-81651611 on Build GRCh38
rs529877143 Chr.17: 81651843-81651843 on Build GRCh38
rs1029692458 Chr.17: 81651949-81651949 on Build GRCh38
rs776823448 Chr.17: 81651708-81651708 on Build GRCh38
rs549090507 Chr.17: 81651951-81651951 on Build GRCh38
rs1017410266 Chr.17: 81651890-81651890 on Build GRCh38
rs779525290 Chr.17: 81651656-81651656 on Build GRCh38
rs572488905 Chr.17: 81651652-81651652 on Build GRCh38

View Full Product Details