Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 23462818-23463344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIOK3
Entrez Gene ID
8780
Gene Name
RIO kinase 3
Gene Aliases
SUDD
UniGene
Hs.445511
Gene Chromosome Location
Chr.18: 23452823-23483140 on Build GRCh38
Transcripts
NM_003831.3 XM_011526242.1 XM_011526243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1388006 c.179+1G>A p.? Chr.18: 23463080-23463080 on Build GRCh38
987044 c.85A>C p.N29H Chr.18: 23462985-23462985 on Build GRCh38
5986015 c.161C>G p.A54G Chr.18: 23463061-23463061 on Build GRCh38
5981357 c.91A>G p.I31V Chr.18: 23462991-23462991 on Build GRCh38
5158452 c.97T>A p.C33S Chr.18: 23462997-23462997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957892990 Chr.18: 23463147-23463147 on Build GRCh38
rs1019100449 Chr.18: 23463207-23463207 on Build GRCh38
rs373422920 Chr.18: 23463080-23463080 on Build GRCh38
rs751938573 Chr.18: 23462918-23462918 on Build GRCh38
rs900146192 Chr.18: 23463105-23463105 on Build GRCh38
rs762569874 Chr.18: 23463027-23463027 on Build GRCh38
rs114614448 Chr.18: 23463231-23463231 on Build GRCh38
rs112166210 Chr.18: 23463039-23463039 on Build GRCh38
rs952298019 Chr.18: 23463289-23463289 on Build GRCh38
rs755606702 Chr.18: 23463294-23463294 on Build GRCh38
rs141535428 Chr.18: 23463026-23463026 on Build GRCh38
rs773807572 Chr.18: 23463048-23463048 on Build GRCh38
rs971497747 Chr.18: 23463303-23463303 on Build GRCh38
rs991934758 Chr.18: 23463292-23463292 on Build GRCh38
rs200387789 Chr.18: 23463079-23463079 on Build GRCh38
rs770658209 Chr.18: 23462991-23462991 on Build GRCh38
rs72884533 Chr.18: 23463142-23463142 on Build GRCh38
rs766415242 Chr.18: 23463191-23463191 on Build GRCh38
rs761229779 Chr.18: 23463064-23463064 on Build GRCh38
rs749509029 Chr.18: 23462957-23462957 on Build GRCh38
rs755217285 Chr.18: 23462963-23462963 on Build GRCh38
rs574279502 Chr.18: 23463006-23463006 on Build GRCh38
rs757075368 Chr.18: 23463108-23463108 on Build GRCh38
rs987607300 Chr.18: 23462869-23462869 on Build GRCh38
rs780238098 Chr.18: 23462953-23462953 on Build GRCh38
rs376925840 Chr.18: 23462893-23462893 on Build GRCh38
rs996402967 Chr.18: 23462888-23462888 on Build GRCh38
rs755847831 Chr.18: 23463276-23463276 on Build GRCh38
rs964940156 Chr.18: 23462851-23462851 on Build GRCh38
rs1045635112 Chr.18: 23463062-23463062 on Build GRCh38
rs768143230 Chr.18: 23462927-23462927 on Build GRCh38
rs927327534 Chr.18: 23463036-23463036 on Build GRCh38
rs780934701 Chr.18: 23463119-23463119 on Build GRCh38
rs11082517 Chr.18: 23463236-23463236 on Build GRCh38
rs769335281 Chr.18: 23463012-23463012 on Build GRCh38
rs367644955 Chr.18: 23462979-23462979 on Build GRCh38
rs971495108 Chr.18: 23463166-23463166 on Build GRCh38
rs73390218 Chr.18: 23463156-23463156 on Build GRCh38
rs1028325944 Chr.18: 23463259-23463259 on Build GRCh38
rs958657176 Chr.18: 23463255-23463255 on Build GRCh38
rs565592958 Chr.18: 23462929-23462929 on Build GRCh38
rs886187341 Chr.18: 23463085-23463085 on Build GRCh38
rs754922698 Chr.18: 23463047-23463049 on Build GRCh38
rs767291749 Chr.18: 23463268-23463269 on Build GRCh38
rs573989960 Chr.18: 23463103-23463106 on Build GRCh38
rs981508968 Chr.18: 23463312-23463312 on Build GRCh38
rs745857103 Chr.18: 23463123-23463123 on Build GRCh38
rs751540802 Chr.18: 23462945-23462946 on Build GRCh38
rs534714634 Chr.18: 23462941-23462941 on Build GRCh38
rs201554501 Chr.18: 23462969-23462969 on Build GRCh38
rs996918957 Chr.18: 23463121-23463121 on Build GRCh38
rs974050017 Chr.18: 23463284-23463284 on Build GRCh38
rs201383568 Chr.18: 23463055-23463055 on Build GRCh38
rs762251854 Chr.18: 23463054-23463054 on Build GRCh38
rs554510760 Chr.18: 23462942-23462942 on Build GRCh38
rs754948562 Chr.18: 23463096-23463096 on Build GRCh38
rs138447722 Chr.18: 23463063-23463063 on Build GRCh38
rs117180203 Chr.18: 23463103-23463103 on Build GRCh38
rs760773665 Chr.18: 23463154-23463154 on Build GRCh38
rs148323869 Chr.18: 23463003-23463003 on Build GRCh38
rs777499107 Chr.18: 23462976-23462976 on Build GRCh38

View Full Product Details