Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 67511153-67511651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSEL
Entrez Gene ID
92126
Gene Name
dermatan sulfate epimerase-like
Gene Aliases
C18orf4
UniGene
Hs.124673
Gene Chromosome Location
Chr.18: 67506582-67516730 on Build GRCh38
Transcripts
NM_032160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
989657 c.3289G>T p.E1097* Chr.18: 67511350-67511350 on Build GRCh38
6149144 c.3485C>T p.A1162V Chr.18: 67511154-67511154 on Build GRCh38
6649554 c.3332A>G p.H1111R Chr.18: 67511307-67511307 on Build GRCh38
7097888 c.3308C>T p.S1103L Chr.18: 67511331-67511331 on Build GRCh38
989661 c.3099G>A p.S1033S Chr.18: 67511540-67511540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768050443 Chr.18: 67511391-67511391 on Build GRCh38
rs749689370 Chr.18: 67511209-67511209 on Build GRCh38
rs201124385 Chr.18: 67511450-67511450 on Build GRCh38
rs778238479 Chr.18: 67511504-67511504 on Build GRCh38
rs72954698 Chr.18: 67511270-67511270 on Build GRCh38
rs769985638 Chr.18: 67511573-67511573 on Build GRCh38
rs754957416 Chr.18: 67511491-67511491 on Build GRCh38
rs137895691 Chr.18: 67511206-67511206 on Build GRCh38
rs1017870994 Chr.18: 67511556-67511556 on Build GRCh38
rs755546753 Chr.18: 67511215-67511215 on Build GRCh38
rs199711651 Chr.18: 67511542-67511542 on Build GRCh38
rs558529844 Chr.18: 67511350-67511350 on Build GRCh38
rs772984422 Chr.18: 67511540-67511540 on Build GRCh38
rs879917373 Chr.18: 67511596-67511596 on Build GRCh38
rs773918044 Chr.18: 67511393-67511393 on Build GRCh38
rs963693612 Chr.18: 67511557-67511557 on Build GRCh38
rs530463345 Chr.18: 67511613-67511613 on Build GRCh38
rs777836420 Chr.18: 67511251-67511251 on Build GRCh38
rs142161259 Chr.18: 67511417-67511417 on Build GRCh38
rs779079443 Chr.18: 67511216-67511216 on Build GRCh38
rs369391532 Chr.18: 67511371-67511371 on Build GRCh38
rs766784460 Chr.18: 67511336-67511336 on Build GRCh38
rs1012663323 Chr.18: 67511492-67511492 on Build GRCh38
rs750643328 Chr.18: 67511614-67511614 on Build GRCh38
rs201645988 Chr.18: 67511414-67511414 on Build GRCh38
rs764960987 Chr.18: 67511458-67511460 on Build GRCh38
rs1056932022 Chr.18: 67511367-67511367 on Build GRCh38
rs749309181 Chr.18: 67511385-67511385 on Build GRCh38
rs760072414 Chr.18: 67511347-67511347 on Build GRCh38
rs973397286 Chr.18: 67511561-67511561 on Build GRCh38
rs373010660 Chr.18: 67511409-67511409 on Build GRCh38
rs144741940 Chr.18: 67511597-67511597 on Build GRCh38
rs764067720 Chr.18: 67511561-67511561 on Build GRCh38
rs754271088 Chr.18: 67511346-67511346 on Build GRCh38
rs758544654 Chr.18: 67511240-67511240 on Build GRCh38
rs376015920 Chr.18: 67511356-67511356 on Build GRCh38
rs146176907 Chr.18: 67511189-67511189 on Build GRCh38
rs755945574 Chr.18: 67511378-67511378 on Build GRCh38
rs768266389 Chr.18: 67511321-67511321 on Build GRCh38
rs774758577 Chr.18: 67511605-67511605 on Build GRCh38
rs745744933 Chr.18: 67511283-67511283 on Build GRCh38
rs774943954 Chr.18: 67511296-67511296 on Build GRCh38
rs747692842 Chr.18: 67511508-67511508 on Build GRCh38
rs779970286 Chr.18: 67511481-67511481 on Build GRCh38
rs763315186 Chr.18: 67511580-67511580 on Build GRCh38
rs764017830 Chr.18: 67511428-67511428 on Build GRCh38
rs764299854 Chr.18: 67511361-67511361 on Build GRCh38
rs1009731167 Chr.18: 67511488-67511488 on Build GRCh38
rs771755291 Chr.18: 67511524-67511524 on Build GRCh38
rs762694026 Chr.18: 67511560-67511561 on Build GRCh38
rs75559029 Chr.18: 67511446-67511446 on Build GRCh38
rs770705002 Chr.18: 67511262-67511262 on Build GRCh38
rs773152317 Chr.18: 67511489-67511489 on Build GRCh38
rs540858968 Chr.18: 67511415-67511415 on Build GRCh38
rs751662919 Chr.18: 67511364-67511364 on Build GRCh38
rs753295138 Chr.18: 67511354-67511354 on Build GRCh38
rs372969112 Chr.18: 67511615-67511615 on Build GRCh38
rs771875702 Chr.18: 67511406-67511406 on Build GRCh38
rs148652078 Chr.18: 67511328-67511328 on Build GRCh38
rs758394930 Chr.18: 67511360-67511360 on Build GRCh38
rs551079213 Chr.18: 67511468-67511468 on Build GRCh38
rs779836111 Chr.18: 67511384-67511384 on Build GRCh38
rs750696157 Chr.18: 67511466-67511466 on Build GRCh38
rs139752903 Chr.18: 67511579-67511579 on Build GRCh38
rs757909793 Chr.18: 67511527-67511527 on Build GRCh38
rs369348785 Chr.18: 67511513-67511513 on Build GRCh38
rs557149581 Chr.18: 67511190-67511190 on Build GRCh38
rs919336823 Chr.18: 67511566-67511566 on Build GRCh38
rs374210715 Chr.18: 67511297-67511297 on Build GRCh38
rs757479420 Chr.18: 67511366-67511366 on Build GRCh38
rs546180436 Chr.18: 67511325-67511325 on Build GRCh38
rs769742441 Chr.18: 67511287-67511287 on Build GRCh38
rs776198401 Chr.18: 67511412-67511413 on Build GRCh38
rs751679229 Chr.18: 67511441-67511441 on Build GRCh38
rs766030986 Chr.18: 67511622-67511622 on Build GRCh38
rs752742986 Chr.18: 67511229-67511229 on Build GRCh38
rs147867124 Chr.18: 67511606-67511606 on Build GRCh38
rs376328875 Chr.18: 67511461-67511461 on Build GRCh38
rs751505302 Chr.18: 67511602-67511603 on Build GRCh38
rs775293820 Chr.18: 67511546-67511546 on Build GRCh38
rs755894252 Chr.18: 67511479-67511479 on Build GRCh38
rs749114153 Chr.18: 67511482-67511482 on Build GRCh38
rs773042057 Chr.18: 67511414-67511417 on Build GRCh38
rs747313884 Chr.18: 67511254-67511254 on Build GRCh38
rs745691476 Chr.18: 67511372-67511372 on Build GRCh38

View Full Product Details