Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 75199339-75199849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZADH2
Entrez Gene ID
284273
Gene Name
zinc binding alcohol dehydrogenase domain containing 2
Gene Aliases
-
UniGene
Hs.465433
Gene Chromosome Location
Chr.18: 75195108-75209348 on Build GRCh38
Transcripts
NM_001306093.1 NM_175907.5

SNP Detail

SNP ID Chromosome Location
rs530025402 Chr.18: 75199513-75199513 on Build GRCh38
rs1021443994 Chr.18: 75199567-75199567 on Build GRCh38
rs959797566 Chr.18: 75199549-75199549 on Build GRCh38
rs906555147 Chr.18: 75199517-75199517 on Build GRCh38
rs1018199156 Chr.18: 75199799-75199799 on Build GRCh38
rs369464103 Chr.18: 75199770-75199770 on Build GRCh38
rs889996574 Chr.18: 75199689-75199689 on Build GRCh38
rs888645512 Chr.18: 75199674-75199674 on Build GRCh38
rs959661059 Chr.18: 75199733-75199733 on Build GRCh38
rs201462712 Chr.18: 75199653-75199653 on Build GRCh38
rs1049871264 Chr.18: 75199824-75199824 on Build GRCh38
rs1045357090 Chr.18: 75199696-75199696 on Build GRCh38
rs538929727 Chr.18: 75199805-75199805 on Build GRCh38
rs201951915 Chr.18: 75199652-75199652 on Build GRCh38
rs188904945 Chr.18: 75199719-75199719 on Build GRCh38
rs552399341 Chr.18: 75199687-75199687 on Build GRCh38
rs17056649 Chr.18: 75199655-75199655 on Build GRCh38
rs1048506057 Chr.18: 75199454-75199454 on Build GRCh38
rs17056651 Chr.18: 75199724-75199724 on Build GRCh38
rs977477148 Chr.18: 75199543-75199543 on Build GRCh38
rs1007128895 Chr.18: 75199759-75199759 on Build GRCh38
rs147268507 Chr.18: 75199749-75199749 on Build GRCh38
rs749445315 Chr.18: 75199753-75199753 on Build GRCh38
rs939946106 Chr.18: 75199766-75199766 on Build GRCh38
rs548227973 Chr.18: 75199725-75199725 on Build GRCh38
rs930153977 Chr.18: 75199435-75199435 on Build GRCh38
rs956741290 Chr.18: 75199560-75199560 on Build GRCh38
rs976791181 Chr.18: 75199624-75199624 on Build GRCh38
rs78389534 Chr.18: 75199461-75199461 on Build GRCh38
rs1031117451 Chr.18: 75199546-75199546 on Build GRCh38
rs868619932 Chr.18: 75199656-75199656 on Build GRCh38
rs561888412 Chr.18: 75199510-75199510 on Build GRCh38
rs906768354 Chr.18: 75199713-75199713 on Build GRCh38
rs1003949014 Chr.18: 75199545-75199545 on Build GRCh38
rs976464690 Chr.18: 75199818-75199818 on Build GRCh38
rs34616479 Chr.18: 75199648-75199649 on Build GRCh38
rs764698586 Chr.18: 75199431-75199431 on Build GRCh38
rs766058261 Chr.18: 75199363-75199363 on Build GRCh38
rs752163686 Chr.18: 75199564-75199564 on Build GRCh38
rs998247077 Chr.18: 75199386-75199386 on Build GRCh38
rs73478089 Chr.18: 75199408-75199408 on Build GRCh38
rs572990860 Chr.18: 75199409-75199409 on Build GRCh38
rs184868869 Chr.18: 75199714-75199714 on Build GRCh38
rs1010966556 Chr.18: 75199737-75199737 on Build GRCh38
rs541830389 Chr.18: 75199423-75199423 on Build GRCh38
rs1022571787 Chr.18: 75199711-75199711 on Build GRCh38
rs1048341532 Chr.18: 75199640-75199640 on Build GRCh38
rs193233384 Chr.18: 75199413-75199413 on Build GRCh38
rs571121481 Chr.18: 75199817-75199817 on Build GRCh38
rs933976399 Chr.18: 75199820-75199820 on Build GRCh38
rs35407927 Chr.18: 75199656-75199656 on Build GRCh38
rs1012427398 Chr.18: 75199470-75199470 on Build GRCh38
rs632157 Chr.18: 75199593-75199593 on Build GRCh38
rs922599408 Chr.18: 75199815-75199815 on Build GRCh38
rs767956979 Chr.18: 75199605-75199605 on Build GRCh38
rs370167746 Chr.18: 75199550-75199553 on Build GRCh38
rs909693424 Chr.18: 75199763-75199763 on Build GRCh38
rs532379171 Chr.18: 75199643-75199643 on Build GRCh38
rs142822241 Chr.18: 75199614-75199614 on Build GRCh38
rs537031565 Chr.18: 75199754-75199754 on Build GRCh38
rs929705719 Chr.18: 75199653-75199653 on Build GRCh38
rs943752937 Chr.18: 75199622-75199622 on Build GRCh38

View Full Product Details