Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43731059-43731549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMG9
Entrez Gene ID
56006
Gene Name
SMG9, nonsense mediated mRNA decay factor
Gene Aliases
C19orf61, F17127_1, HBMS
UniGene
Hs.466875
Gene Chromosome Location
Chr.19: 43727992-43754990 on Build GRCh38
Transcripts
XM_005259057.2 XM_011527113.1 XM_011527114.1 XM_011527115.1 XM_011527116.1 XM_017026988.1

SNP Detail

SNP ID Chromosome Location
rs936611747 Chr.19: 43731376-43731376 on Build GRCh38
rs113829853 Chr.19: 43731163-43731163 on Build GRCh38
rs576685510 Chr.19: 43731299-43731299 on Build GRCh38
rs974028689 Chr.19: 43731253-43731253 on Build GRCh38
rs980369448 Chr.19: 43731360-43731360 on Build GRCh38
rs917981127 Chr.19: 43731518-43731518 on Build GRCh38
rs1044321 Chr.19: 43731459-43731459 on Build GRCh38
rs539341775 Chr.19: 43731300-43731300 on Build GRCh38
rs1036157450 Chr.19: 43731083-43731083 on Build GRCh38
rs918628390 Chr.19: 43731258-43731258 on Build GRCh38
rs780542469 Chr.19: 43731238-43731239 on Build GRCh38
rs1044334 Chr.19: 43731321-43731321 on Build GRCh38
rs549189844 Chr.19: 43731152-43731152 on Build GRCh38
rs754580497 Chr.19: 43731392-43731392 on Build GRCh38
rs947856900 Chr.19: 43731340-43731340 on Build GRCh38
rs897650400 Chr.19: 43731092-43731092 on Build GRCh38
rs202057162 Chr.19: 43731395-43731396 on Build GRCh38
rs535892284 Chr.19: 43731143-43731143 on Build GRCh38
rs1001528701 Chr.19: 43731287-43731287 on Build GRCh38
rs1006870840 Chr.19: 43731231-43731231 on Build GRCh38
rs1044933343 Chr.19: 43731341-43731341 on Build GRCh38
rs1009565602 Chr.19: 43731499-43731499 on Build GRCh38
rs566745179 Chr.19: 43731122-43731122 on Build GRCh38
rs375571880 Chr.19: 43731428-43731428 on Build GRCh38
rs891236917 Chr.19: 43731224-43731224 on Build GRCh38
rs888886257 Chr.19: 43731410-43731410 on Build GRCh38
rs12669 Chr.19: 43731383-43731383 on Build GRCh38
rs1000910834 Chr.19: 43731517-43731517 on Build GRCh38
rs756083054 Chr.19: 43731236-43731236 on Build GRCh38
rs572726861 Chr.19: 43731332-43731332 on Build GRCh38
rs917649467 Chr.19: 43731359-43731359 on Build GRCh38
rs190072355 Chr.19: 43731303-43731303 on Build GRCh38
rs918012057 Chr.19: 43731301-43731301 on Build GRCh38
rs9612 Chr.19: 43731365-43731365 on Build GRCh38
rs556737642 Chr.19: 43731252-43731252 on Build GRCh38
rs941739234 Chr.19: 43731370-43731370 on Build GRCh38
rs1044336 Chr.19: 43731240-43731240 on Build GRCh38
rs564681236 Chr.19: 43731442-43731442 on Build GRCh38
rs992191813 Chr.19: 43731503-43731503 on Build GRCh38
rs546977704 Chr.19: 43731502-43731502 on Build GRCh38
rs536819893 Chr.19: 43731233-43731233 on Build GRCh38
rs569023219 Chr.19: 43731212-43731212 on Build GRCh38
rs1051620599 Chr.19: 43731428-43731428 on Build GRCh38
rs992368594 Chr.19: 43731319-43731319 on Build GRCh38
rs575433529 Chr.19: 43731408-43731408 on Build GRCh38
rs997298996 Chr.19: 43731119-43731119 on Build GRCh38
rs761992761 Chr.19: 43731297-43731297 on Build GRCh38
rs974330875 Chr.19: 43731466-43731466 on Build GRCh38
rs962932383 Chr.19: 43731465-43731465 on Build GRCh38
rs892100621 Chr.19: 43731384-43731384 on Build GRCh38
rs1006988799 Chr.19: 43731452-43731452 on Build GRCh38
rs533617818 Chr.19: 43731456-43731456 on Build GRCh38

View Full Product Details