Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 33514929-33515428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASGRP3
Entrez Gene ID
25780
Gene Name
RAS guanyl releasing protein 3
Gene Aliases
GRP3
UniGene
Hs.143674
Gene Chromosome Location
Chr.2: 33436348-33564731 on Build GRCh38
Transcripts
NM_001139488.1 NM_015376.2 NM_170672.2 XM_005264246.3 XM_011532746.2 XM_011532747.2 XM_011532748.2 XM_017003759.1 XM_017003760.1 XM_017003761.1 XM_017003762.1 XM_017003763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
404709 c.70G>T p.D24Y Chr.2: 33515206-33515206 on Build GRCh38
1236516 c.57C>A p.C19* Chr.2: 33515193-33515193 on Build GRCh38
2943115 c.24A>C p.K8N Chr.2: 33515160-33515160 on Build GRCh38
4312968 c.44T>C p.L15P Chr.2: 33515180-33515180 on Build GRCh38
5949612 c.70+2T>C p.? Chr.2: 33515208-33515208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021890114 Chr.2: 33515287-33515287 on Build GRCh38
rs938727181 Chr.2: 33515091-33515091 on Build GRCh38
rs911316694 Chr.2: 33515245-33515245 on Build GRCh38
rs997471570 Chr.2: 33515223-33515223 on Build GRCh38
rs769234948 Chr.2: 33515076-33515076 on Build GRCh38
rs531417643 Chr.2: 33515338-33515338 on Build GRCh38
rs377201321 Chr.2: 33515124-33515124 on Build GRCh38
rs757835759 Chr.2: 33515238-33515238 on Build GRCh38
rs936458593 Chr.2: 33515336-33515336 on Build GRCh38
rs781662268 Chr.2: 33515157-33515157 on Build GRCh38
rs774482287 Chr.2: 33515195-33515195 on Build GRCh38
rs745352044 Chr.2: 33515145-33515145 on Build GRCh38
rs1030753010 Chr.2: 33515158-33515158 on Build GRCh38
rs774517357 Chr.2: 33515104-33515104 on Build GRCh38
rs556078226 Chr.2: 33515038-33515038 on Build GRCh38
rs868297673 Chr.2: 33515203-33515204 on Build GRCh38
rs955907738 Chr.2: 33514969-33514969 on Build GRCh38
rs529397346 Chr.2: 33515161-33515161 on Build GRCh38
rs774446401 Chr.2: 33515204-33515205 on Build GRCh38
rs370175209 Chr.2: 33515115-33515115 on Build GRCh38
rs779405453 Chr.2: 33515384-33515384 on Build GRCh38
rs753700031 Chr.2: 33515228-33515228 on Build GRCh38
rs779551774 Chr.2: 33515246-33515246 on Build GRCh38
rs765298936 Chr.2: 33515067-33515067 on Build GRCh38
rs771149612 Chr.2: 33515184-33515184 on Build GRCh38
rs868221360 Chr.2: 33515207-33515207 on Build GRCh38
rs151310793 Chr.2: 33515057-33515057 on Build GRCh38
rs183877175 Chr.2: 33515259-33515259 on Build GRCh38
rs76096788 Chr.2: 33515322-33515322 on Build GRCh38
rs575745227 Chr.2: 33515281-33515281 on Build GRCh38
rs79274432 Chr.2: 33515058-33515058 on Build GRCh38
rs1057111829 Chr.2: 33515089-33515089 on Build GRCh38
rs1056742504 Chr.2: 33515302-33515302 on Build GRCh38
rs990423884 Chr.2: 33514996-33514996 on Build GRCh38
rs759570048 Chr.2: 33515110-33515110 on Build GRCh38
rs144716086 Chr.2: 33515032-33515032 on Build GRCh38
rs985834345 Chr.2: 33515240-33515240 on Build GRCh38
rs763933440 Chr.2: 33515219-33515219 on Build GRCh38
rs749428928 Chr.2: 33515174-33515174 on Build GRCh38
rs969976611 Chr.2: 33515289-33515289 on Build GRCh38
rs535963940 Chr.2: 33515343-33515343 on Build GRCh38
rs150489838 Chr.2: 33515351-33515351 on Build GRCh38
rs942929355 Chr.2: 33515393-33515393 on Build GRCh38
rs375302546 Chr.2: 33515135-33515135 on Build GRCh38
rs368115160 Chr.2: 33515269-33515269 on Build GRCh38
rs2305578 Chr.2: 33515122-33515122 on Build GRCh38
rs756957301 Chr.2: 33515136-33515136 on Build GRCh38
rs368361129 Chr.2: 33515143-33515143 on Build GRCh38
rs201561963 Chr.2: 33515211-33515211 on Build GRCh38
rs947760279 Chr.2: 33515308-33515308 on Build GRCh38
rs930117706 Chr.2: 33515286-33515286 on Build GRCh38
rs752636179 Chr.2: 33515074-33515074 on Build GRCh38
rs757953919 Chr.2: 33515153-33515153 on Build GRCh38
rs878959744 Chr.2: 33515005-33515005 on Build GRCh38
rs113370628 Chr.2: 33515125-33515125 on Build GRCh38
rs551803972 Chr.2: 33515261-33515261 on Build GRCh38
rs896183301 Chr.2: 33515101-33515101 on Build GRCh38
rs558780048 Chr.2: 33515114-33515114 on Build GRCh38
rs1048529572 Chr.2: 33515364-33515364 on Build GRCh38
rs941521944 Chr.2: 33515033-33515033 on Build GRCh38
rs371725125 Chr.2: 33515230-33515230 on Build GRCh38
rs903615936 Chr.2: 33515328-33515328 on Build GRCh38
rs551511091 Chr.2: 33514998-33514999 on Build GRCh38
rs567000061 Chr.2: 33515011-33515011 on Build GRCh38
rs918140470 Chr.2: 33515303-33515303 on Build GRCh38
rs115337971 Chr.2: 33515388-33515388 on Build GRCh38
rs985585306 Chr.2: 33515024-33515024 on Build GRCh38
rs760704497 Chr.2: 33515215-33515215 on Build GRCh38
rs527318101 Chr.2: 33515321-33515321 on Build GRCh38
rs764921508 Chr.2: 33515235-33515235 on Build GRCh38
rs202227874 Chr.2: 33515160-33515160 on Build GRCh38
rs562942258 Chr.2: 33515258-33515258 on Build GRCh38
rs759651278 Chr.2: 33515210-33515210 on Build GRCh38
rs554329413 Chr.2: 33515280-33515280 on Build GRCh38
rs376168602 Chr.2: 33515213-33515213 on Build GRCh38

View Full Product Details