Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130342999-130343475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMP4
Entrez Gene ID
92856
Gene Name
IMP4 homolog, U3 small nucleolar ribonucleoprotein
Gene Aliases
BXDC4
UniGene
Hs.91579
Gene Chromosome Location
Chr.2: 130342225-130347961 on Build GRCh38
Transcripts
NM_001320304.1 NM_001320305.1 NM_001320306.1 NM_001320307.1 NM_001320309.1 NM_001320310.1 NM_001320311.1 NM_033416.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6340761 c.4-6C>G p.? Chr.2: 130343080-130343080 on Build GRCh38
5354260 c.9C>A p.R3R Chr.2: 130343091-130343091 on Build GRCh38
4648439 c.67C>T p.R23C Chr.2: 130343149-130343149 on Build GRCh38
6340850 c.81G>T p.E27D Chr.2: 130343163-130343163 on Build GRCh38
39121 c.13G>A p.E5K Chr.2: 130343095-130343095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761281200 Chr.2: 130343338-130343338 on Build GRCh38
rs774522477 Chr.2: 130343069-130343069 on Build GRCh38
rs547873913 Chr.2: 130343169-130343169 on Build GRCh38
rs774036320 Chr.2: 130343229-130343229 on Build GRCh38
rs11542411 Chr.2: 130343161-130343161 on Build GRCh38
rs762130210 Chr.2: 130343299-130343299 on Build GRCh38
rs899949086 Chr.2: 130343423-130343423 on Build GRCh38
rs1036573197 Chr.2: 130343036-130343036 on Build GRCh38
rs188123387 Chr.2: 130343422-130343422 on Build GRCh38
rs867739597 Chr.2: 130343351-130343351 on Build GRCh38
rs760152080 Chr.2: 130343073-130343073 on Build GRCh38
rs73960310 Chr.2: 130343263-130343263 on Build GRCh38
rs769479224 Chr.2: 130343125-130343125 on Build GRCh38
rs768140565 Chr.2: 130343194-130343194 on Build GRCh38
rs749543790 Chr.2: 130343134-130343134 on Build GRCh38
rs981806405 Chr.2: 130343302-130343302 on Build GRCh38
rs771148162 Chr.2: 130343066-130343066 on Build GRCh38
rs768537819 Chr.2: 130343042-130343042 on Build GRCh38
rs903256641 Chr.2: 130343433-130343433 on Build GRCh38
rs761004311 Chr.2: 130343089-130343089 on Build GRCh38
rs559100058 Chr.2: 130343150-130343150 on Build GRCh38
rs930233828 Chr.2: 130343330-130343330 on Build GRCh38
rs781046293 Chr.2: 130343117-130343117 on Build GRCh38
rs372488819 Chr.2: 130343438-130343438 on Build GRCh38
rs750051393 Chr.2: 130343099-130343099 on Build GRCh38
rs772181665 Chr.2: 130343185-130343185 on Build GRCh38
rs1047247974 Chr.2: 130343361-130343361 on Build GRCh38
rs748108748 Chr.2: 130343119-130343119 on Build GRCh38
rs773372090 Chr.2: 130343061-130343061 on Build GRCh38
rs890361711 Chr.2: 130343441-130343441 on Build GRCh38
rs781149016 Chr.2: 130343045-130343045 on Build GRCh38
rs61744776 Chr.2: 130343026-130343026 on Build GRCh38
rs754457920 Chr.2: 130343116-130343116 on Build GRCh38
rs746966091 Chr.2: 130343038-130343038 on Build GRCh38
rs569656506 Chr.2: 130343184-130343184 on Build GRCh38
rs766064767 Chr.2: 130343300-130343300 on Build GRCh38
rs373818681 Chr.2: 130343350-130343350 on Build GRCh38
rs747940085 Chr.2: 130343046-130343046 on Build GRCh38
rs751197615 Chr.2: 130343304-130343304 on Build GRCh38
rs761179766 Chr.2: 130343237-130343237 on Build GRCh38
rs754373846 Chr.2: 130343379-130343379 on Build GRCh38
rs771688194 Chr.2: 130343350-130343353 on Build GRCh38
rs928198458 Chr.2: 130343306-130343306 on Build GRCh38
rs998640773 Chr.2: 130343214-130343214 on Build GRCh38
rs1043085989 Chr.2: 130343193-130343193 on Build GRCh38
rs908290279 Chr.2: 130343357-130343357 on Build GRCh38
rs957912006 Chr.2: 130343176-130343176 on Build GRCh38
rs547440763 Chr.2: 130343109-130343109 on Build GRCh38
rs768179974 Chr.2: 130343079-130343079 on Build GRCh38
rs1035551887 Chr.2: 130343250-130343250 on Build GRCh38
rs778061864 Chr.2: 130343308-130343308 on Build GRCh38
rs74268761 Chr.2: 130343424-130343424 on Build GRCh38
rs1006077835 Chr.2: 130343048-130343048 on Build GRCh38
rs753227794 Chr.2: 130343081-130343081 on Build GRCh38
rs536934514 Chr.2: 130343236-130343236 on Build GRCh38
rs756029734 Chr.2: 130343381-130343381 on Build GRCh38
rs1051070040 Chr.2: 130343430-130343430 on Build GRCh38
rs557379910 Chr.2: 130343448-130343448 on Build GRCh38
rs961141050 Chr.2: 130343358-130343358 on Build GRCh38
rs764518364 Chr.2: 130343093-130343093 on Build GRCh38
rs796859459 Chr.2: 130343227-130343228 on Build GRCh38
rs769615550 Chr.2: 130343055-130343055 on Build GRCh38
rs763167644 Chr.2: 130343065-130343065 on Build GRCh38
rs764382053 Chr.2: 130343245-130343245 on Build GRCh38
rs79014886 Chr.2: 130343343-130343343 on Build GRCh38
rs972214432 Chr.2: 130343359-130343359 on Build GRCh38
rs534824876 Chr.2: 130343383-130343383 on Build GRCh38
rs774683967 Chr.2: 130343166-130343166 on Build GRCh38
rs779644628 Chr.2: 130343128-130343128 on Build GRCh38
rs1012806128 Chr.2: 130343311-130343311 on Build GRCh38
rs376438230 Chr.2: 130343390-130343390 on Build GRCh38
rs769619902 Chr.2: 130343126-130343126 on Build GRCh38
rs916333335 Chr.2: 130343205-130343205 on Build GRCh38
rs779566149 Chr.2: 130343113-130343113 on Build GRCh38
rs1050662518 Chr.2: 130343400-130343400 on Build GRCh38
rs930120685 Chr.2: 130343021-130343021 on Build GRCh38
rs752587772 Chr.2: 130343370-130343370 on Build GRCh38
rs940832292 Chr.2: 130343035-130343035 on Build GRCh38

View Full Product Details