Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 174566992-174567494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WIPF1
Entrez Gene ID
7456
Gene Name
WAS/WASL interacting protein family member 1
Gene Aliases
PRPL-2, WAS2, WASPIP, WIP
UniGene
Hs.128067
Gene Chromosome Location
Chr.2: 174559574-174682899 on Build GRCh38
Transcripts
NM_001077269.1 NM_003387.4 XM_011511780.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4525748 c.1350G>A p.W450* Chr.2: 174567176-174567176 on Build GRCh38
3425480 c.1358G>T p.R453I Chr.2: 174567168-174567168 on Build GRCh38
3425479 c.1358G>T p.R453I Chr.2: 174567168-174567168 on Build GRCh38
6043429 c.1394C>T p.P465L Chr.2: 174567132-174567132 on Build GRCh38
341270 c.1373C>G p.P458R Chr.2: 174567153-174567153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991985029 Chr.2: 174567304-174567304 on Build GRCh38
rs3217351 Chr.2: 174567270-174567273 on Build GRCh38
rs746587628 Chr.2: 174567064-174567064 on Build GRCh38
rs761674906 Chr.2: 174567074-174567074 on Build GRCh38
rs761503886 Chr.2: 174567145-174567145 on Build GRCh38
rs757711719 Chr.2: 174567152-174567152 on Build GRCh38
rs183514108 Chr.2: 174567368-174567368 on Build GRCh38
rs887294523 Chr.2: 174567434-174567434 on Build GRCh38
rs547689816 Chr.2: 174567404-174567404 on Build GRCh38
rs866520694 Chr.2: 174567100-174567100 on Build GRCh38
rs768372129 Chr.2: 174567219-174567219 on Build GRCh38
rs766679061 Chr.2: 174567241-174567241 on Build GRCh38
rs962702774 Chr.2: 174567256-174567256 on Build GRCh38
rs529408738 Chr.2: 174567393-174567393 on Build GRCh38
rs769628861 Chr.2: 174567230-174567230 on Build GRCh38
rs925903392 Chr.2: 174567329-174567329 on Build GRCh38
rs1004384210 Chr.2: 174567456-174567456 on Build GRCh38
rs373572516 Chr.2: 174567226-174567226 on Build GRCh38
rs916162933 Chr.2: 174567305-174567305 on Build GRCh38
rs768251974 Chr.2: 174567070-174567070 on Build GRCh38
rs113179543 Chr.2: 174567112-174567112 on Build GRCh38
rs779896185 Chr.2: 174567203-174567203 on Build GRCh38
rs1039819305 Chr.2: 174567130-174567130 on Build GRCh38
rs767550471 Chr.2: 174567394-174567394 on Build GRCh38
rs1004886221 Chr.2: 174567197-174567197 on Build GRCh38
rs369668974 Chr.2: 174567131-174567131 on Build GRCh38
rs1033380200 Chr.2: 174567288-174567288 on Build GRCh38
rs149092210 Chr.2: 174567113-174567113 on Build GRCh38
rs34258448 Chr.2: 174567269-174567272 on Build GRCh38
rs764135214 Chr.2: 174567143-174567143 on Build GRCh38
rs116757787 Chr.2: 174567080-174567080 on Build GRCh38
rs746640640 Chr.2: 174567216-174567216 on Build GRCh38
rs747624048 Chr.2: 174567229-174567229 on Build GRCh38
rs752556801 Chr.2: 174567432-174567432 on Build GRCh38
rs376965787 Chr.2: 174567188-174567188 on Build GRCh38
rs779418182 Chr.2: 174567161-174567161 on Build GRCh38
rs201208878 Chr.2: 174567105-174567105 on Build GRCh38
rs746177198 Chr.2: 174567024-174567024 on Build GRCh38
rs1041727019 Chr.2: 174567073-174567073 on Build GRCh38
rs143689509 Chr.2: 174567263-174567266 on Build GRCh38
rs1043656127 Chr.2: 174567034-174567034 on Build GRCh38
rs1051246656 Chr.2: 174567410-174567410 on Build GRCh38
rs753915750 Chr.2: 174567146-174567146 on Build GRCh38
rs3815969 Chr.2: 174567370-174567370 on Build GRCh38
rs2303891 Chr.2: 174567328-174567328 on Build GRCh38
rs775272660 Chr.2: 174567036-174567036 on Build GRCh38
rs752911584 Chr.2: 174567114-174567114 on Build GRCh38
rs935980569 Chr.2: 174567343-174567343 on Build GRCh38
rs960775758 Chr.2: 174567289-174567289 on Build GRCh38
rs546167717 Chr.2: 174567245-174567245 on Build GRCh38
rs564455904 Chr.2: 174567325-174567325 on Build GRCh38
rs1001873494 Chr.2: 174567286-174567286 on Build GRCh38
rs973744147 Chr.2: 174567270-174567270 on Build GRCh38
rs776761827 Chr.2: 174567072-174567072 on Build GRCh38
rs746276409 Chr.2: 174567162-174567162 on Build GRCh38
rs1015222378 Chr.2: 174567246-174567246 on Build GRCh38
rs934098478 Chr.2: 174567379-174567379 on Build GRCh38
rs200493942 Chr.2: 174567035-174567035 on Build GRCh38

View Full Product Details