Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 189755448-189755985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKAR
Entrez Gene ID
150709
Gene Name
ankyrin and armadillo repeat containing
Gene Aliases
-
UniGene
Hs.60772
Gene Chromosome Location
Chr.2: 189674292-189763059 on Build GRCh38
Transcripts
XM_011510676.2 XM_011510677.2 XM_011510681.1 XM_011510682.2 NM_022353.2 XM_005246766.3 XM_006712685.1 XM_006712686.1 XM_011511631.1 XM_017004676.1 XM_017004677.1 XM_017004678.1 XM_017004679.1 XM_017004680.1 XM_017004681.1 XM_017004682.1 XM_017004683.1 XM_017004684.1 XM_017004685.1

Gene Symbol
OSGEPL1
Entrez Gene ID
64172
Gene Name
O-sialoglycoprotein endopeptidase-like 1
Gene Aliases
Qri7
UniGene
Hs.60772
Gene Chromosome Location
Chr.2: 189746660-189763198 on Build GRCh38
Transcripts
XM_011510676.2 XM_011510677.2 XM_011510681.1 XM_011510682.2 NM_022353.2 XM_005246766.3 XM_006712685.1 XM_006712686.1 XM_011511631.1 XM_017004676.1 XM_017004677.1 XM_017004678.1 XM_017004679.1 XM_017004680.1 XM_017004681.1 XM_017004682.1 XM_017004683.1 XM_017004684.1 XM_017004685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5586101 c.299C>T p.S100F Chr.2: 189755483-189755483 on Build GRCh38
1014064 c.278G>T p.R93L Chr.2: 189755504-189755504 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757571698 Chr.2: 189755575-189755575 on Build GRCh38
rs538730876 Chr.2: 189755892-189755892 on Build GRCh38
rs773068198 Chr.2: 189755502-189755502 on Build GRCh38
rs1046697435 Chr.2: 189755789-189755789 on Build GRCh38
rs746752481 Chr.2: 189755790-189755790 on Build GRCh38
rs536822486 Chr.2: 189755484-189755484 on Build GRCh38
rs570164694 Chr.2: 189755472-189755472 on Build GRCh38
rs570672168 Chr.2: 189755597-189755597 on Build GRCh38
rs778898112 Chr.2: 189755730-189755730 on Build GRCh38
rs1013767870 Chr.2: 189755523-189755523 on Build GRCh38
rs2244190 Chr.2: 189755766-189755766 on Build GRCh38
rs778494248 Chr.2: 189755486-189755486 on Build GRCh38
rs954877846 Chr.2: 189755959-189755959 on Build GRCh38
rs771838736 Chr.2: 189755496-189755496 on Build GRCh38
rs1051823293 Chr.2: 189755539-189755539 on Build GRCh38
rs531402192 Chr.2: 189755670-189755670 on Build GRCh38
rs559659052 Chr.2: 189755839-189755839 on Build GRCh38
rs377465918 Chr.2: 189755504-189755504 on Build GRCh38
rs904493222 Chr.2: 189755908-189755908 on Build GRCh38
rs1001810857 Chr.2: 189755933-189755933 on Build GRCh38
rs941973202 Chr.2: 189755807-189755807 on Build GRCh38
rs764292642 Chr.2: 189755563-189755563 on Build GRCh38
rs776381876 Chr.2: 189755528-189755528 on Build GRCh38
rs574484862 Chr.2: 189755640-189755640 on Build GRCh38
rs898113854 Chr.2: 189755481-189755481 on Build GRCh38
rs775557271 Chr.2: 189755552-189755552 on Build GRCh38
rs765162906 Chr.2: 189755536-189755536 on Build GRCh38
rs767581066 Chr.2: 189755725-189755728 on Build GRCh38
rs986628968 Chr.2: 189755666-189755666 on Build GRCh38
rs191653364 Chr.2: 189755630-189755630 on Build GRCh38
rs371254771 Chr.2: 189755569-189755569 on Build GRCh38
rs1024756905 Chr.2: 189755763-189755763 on Build GRCh38
rs1030299148 Chr.2: 189755956-189755956 on Build GRCh38
rs774331405 Chr.2: 189755562-189755563 on Build GRCh38
rs377276256 Chr.2: 189755579-189755579 on Build GRCh38
rs530025918 Chr.2: 189755856-189755856 on Build GRCh38
rs941936646 Chr.2: 189755495-189755495 on Build GRCh38
rs755187413 Chr.2: 189755713-189755713 on Build GRCh38
rs777756479 Chr.2: 189755788-189755788 on Build GRCh38
rs540478958 Chr.2: 189755732-189755732 on Build GRCh38
rs956627633 Chr.2: 189755897-189755897 on Build GRCh38
rs182721041 Chr.2: 189755710-189755710 on Build GRCh38
rs763075595 Chr.2: 189755557-189755557 on Build GRCh38
rs116782323 Chr.2: 189755795-189755795 on Build GRCh38
rs16831964 Chr.2: 189755634-189755634 on Build GRCh38
rs541851689 Chr.2: 189755688-189755688 on Build GRCh38
rs138161711 Chr.2: 189755898-189755898 on Build GRCh38
rs2278591 Chr.2: 189755778-189755778 on Build GRCh38
rs752251874 Chr.2: 189755599-189755599 on Build GRCh38
rs766489407 Chr.2: 189755620-189755620 on Build GRCh38
rs75321854 Chr.2: 189755490-189755490 on Build GRCh38
rs746412119 Chr.2: 189755476-189755476 on Build GRCh38
rs532577623 Chr.2: 189755791-189755791 on Build GRCh38
rs756662240 Chr.2: 189755584-189755584 on Build GRCh38
rs933180933 Chr.2: 189755852-189755852 on Build GRCh38
rs946097226 Chr.2: 189755881-189755881 on Build GRCh38
rs187290885 Chr.2: 189755904-189755904 on Build GRCh38
rs904987107 Chr.2: 189755818-189755818 on Build GRCh38
rs577825925 Chr.2: 189755505-189755505 on Build GRCh38
rs1023336318 Chr.2: 189755817-189755817 on Build GRCh38
rs35057357 Chr.2: 189755887-189755888 on Build GRCh38
rs1013295761 Chr.2: 189755805-189755805 on Build GRCh38
rs376142185 Chr.2: 189755577-189755577 on Build GRCh38
rs759548788 Chr.2: 189755529-189755529 on Build GRCh38
rs781517462 Chr.2: 189755473-189755473 on Build GRCh38
rs933392424 Chr.2: 189755663-189755663 on Build GRCh38
rs923284562 Chr.2: 189755657-189755657 on Build GRCh38
rs921855814 Chr.2: 189755824-189755824 on Build GRCh38

View Full Product Details