Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33039256-33039755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFB6
Entrez Gene ID
128859
Gene Name
BPI fold containing family B member 6
Gene Aliases
BPIL3, LPLUNC6
UniGene
Hs.375090
Gene Chromosome Location
Chr.20: 33031648-33044108 on Build GRCh38
Transcripts
NM_174897.2 XM_017027663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1632237 c.1054T>C p.S352P Chr.20: 33039500-33039500 on Build GRCh38
7357668 c.1024G>A p.A342T Chr.20: 33039470-33039470 on Build GRCh38
5656496 c.1012C>A p.L338M Chr.20: 33039458-33039458 on Build GRCh38
6394936 c.1041C>A p.S347R Chr.20: 33039487-33039487 on Build GRCh38
4001877 c.1039A>G p.S347G Chr.20: 33039485-33039485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545174792 Chr.20: 33039704-33039704 on Build GRCh38
rs140306506 Chr.20: 33039313-33039313 on Build GRCh38
rs773409886 Chr.20: 33039515-33039515 on Build GRCh38
rs953360646 Chr.20: 33039347-33039347 on Build GRCh38
rs139462993 Chr.20: 33039381-33039381 on Build GRCh38
rs144493645 Chr.20: 33039542-33039542 on Build GRCh38
rs765273988 Chr.20: 33039478-33039478 on Build GRCh38
rs186277546 Chr.20: 33039680-33039680 on Build GRCh38
rs751907499 Chr.20: 33039593-33039593 on Build GRCh38
rs376078530 Chr.20: 33039544-33039544 on Build GRCh38
rs746548517 Chr.20: 33039423-33039423 on Build GRCh38
rs780819648 Chr.20: 33039488-33039488 on Build GRCh38
rs369417019 Chr.20: 33039525-33039525 on Build GRCh38
rs766503313 Chr.20: 33039476-33039476 on Build GRCh38
rs778835815 Chr.20: 33039404-33039404 on Build GRCh38
rs527723448 Chr.20: 33039382-33039382 on Build GRCh38
rs549365003 Chr.20: 33039416-33039416 on Build GRCh38
rs772474393 Chr.20: 33039502-33039502 on Build GRCh38
rs993110722 Chr.20: 33039613-33039613 on Build GRCh38
rs919508695 Chr.20: 33039396-33039396 on Build GRCh38
rs191277611 Chr.20: 33039691-33039691 on Build GRCh38
rs768894788 Chr.20: 33039437-33039437 on Build GRCh38
rs769231359 Chr.20: 33039492-33039492 on Build GRCh38
rs45495506 Chr.20: 33039470-33039470 on Build GRCh38
rs370022331 Chr.20: 33039375-33039375 on Build GRCh38
rs777369840 Chr.20: 33039419-33039419 on Build GRCh38
rs370448245 Chr.20: 33039571-33039571 on Build GRCh38
rs377574610 Chr.20: 33039497-33039497 on Build GRCh38
rs550355845 Chr.20: 33039482-33039482 on Build GRCh38
rs766902113 Chr.20: 33039540-33039541 on Build GRCh38
rs1014001796 Chr.20: 33039621-33039621 on Build GRCh38
rs775084528 Chr.20: 33039483-33039483 on Build GRCh38
rs945697619 Chr.20: 33039490-33039490 on Build GRCh38
rs375412873 Chr.20: 33039321-33039321 on Build GRCh38
rs751714465 Chr.20: 33039336-33039336 on Build GRCh38
rs58463355 Chr.20: 33039533-33039533 on Build GRCh38
rs549045100 Chr.20: 33039640-33039640 on Build GRCh38
rs751250158 Chr.20: 33039565-33039565 on Build GRCh38
rs762138075 Chr.20: 33039501-33039501 on Build GRCh38
rs1026525251 Chr.20: 33039599-33039599 on Build GRCh38
rs573607888 Chr.20: 33039641-33039641 on Build GRCh38
rs560786246 Chr.20: 33039668-33039668 on Build GRCh38
rs558937473 Chr.20: 33039285-33039286 on Build GRCh38
rs752557415 Chr.20: 33039479-33039479 on Build GRCh38
rs371753435 Chr.20: 33039407-33039407 on Build GRCh38
rs560399603 Chr.20: 33039731-33039731 on Build GRCh38
rs770676947 Chr.20: 33039395-33039395 on Build GRCh38
rs577793162 Chr.20: 33039699-33039699 on Build GRCh38
rs916136422 Chr.20: 33039639-33039639 on Build GRCh38
rs144850349 Chr.20: 33039491-33039491 on Build GRCh38
rs750671898 Chr.20: 33039673-33039673 on Build GRCh38
rs971258901 Chr.20: 33039445-33039445 on Build GRCh38
rs202181196 Chr.20: 33039469-33039469 on Build GRCh38
rs756008911 Chr.20: 33039379-33039379 on Build GRCh38
rs139555218 Chr.20: 33039487-33039487 on Build GRCh38
rs776516578 Chr.20: 33039434-33039434 on Build GRCh38
rs181019748 Chr.20: 33039580-33039580 on Build GRCh38
rs770602260 Chr.20: 33039426-33039426 on Build GRCh38
rs972216918 Chr.20: 33039385-33039385 on Build GRCh38
rs766304727 Chr.20: 33039530-33039530 on Build GRCh38
rs374270182 Chr.20: 33039532-33039532 on Build GRCh38
rs200258385 Chr.20: 33039288-33039288 on Build GRCh38
rs201153947 Chr.20: 33039460-33039460 on Build GRCh38
rs750803333 Chr.20: 33039649-33039649 on Build GRCh38
rs976159200 Chr.20: 33039710-33039710 on Build GRCh38
rs4911287 Chr.20: 33039485-33039485 on Build GRCh38
rs760956234 Chr.20: 33039520-33039520 on Build GRCh38
rs796669572 Chr.20: 33039604-33039604 on Build GRCh38
rs897859345 Chr.20: 33039602-33039602 on Build GRCh38
rs761895203 Chr.20: 33039450-33039450 on Build GRCh38
rs775834459 Chr.20: 33039429-33039429 on Build GRCh38
rs770854127 Chr.20: 33039293-33039293 on Build GRCh38
rs925032674 Chr.20: 33039648-33039648 on Build GRCh38
rs754718394 Chr.20: 33039384-33039384 on Build GRCh38
rs764033664 Chr.20: 33039548-33039548 on Build GRCh38
rs762158268 Chr.20: 33039362-33039362 on Build GRCh38
rs753717166 Chr.20: 33039477-33039477 on Build GRCh38
rs200171093 Chr.20: 33039330-33039330 on Build GRCh38
rs1034208822 Chr.20: 33039679-33039679 on Build GRCh38
rs867999560 Chr.20: 33039453-33039453 on Build GRCh38
rs767530991 Chr.20: 33039364-33039364 on Build GRCh38
rs933126243 Chr.20: 33039695-33039695 on Build GRCh38

View Full Product Details