Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 42854470-42854969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR4
Entrez Gene ID
10785
Gene Name
WD repeat domain 4
Gene Aliases
TRM82, TRMT82
UniGene
Hs.248815
Gene Chromosome Location
Chr.21: 42843075-42879865 on Build GRCh38
Transcripts
NM_001260474.1 NM_001260475.1 NM_001260476.1 NM_001260477.1 NM_018669.5 NM_033661.4 NR_048535.1 XM_011529433.1 XM_017028262.1 XM_017028263.1 XM_017028264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2819959 c.753C>T p.F251F Chr.21: 42854600-42854600 on Build GRCh38
1714052 c.743_744GG>AT p.R248N Chr.21: 42854609-42854610 on Build GRCh38
2819960 c.736G>A p.A246T Chr.21: 42854617-42854617 on Build GRCh38
6526307 c.734C>T p.A245V Chr.21: 42854619-42854619 on Build GRCh38
5813233 c.761A>T p.Q254L Chr.21: 42854592-42854592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941399780 Chr.21: 42854738-42854738 on Build GRCh38
rs778707578 Chr.21: 42854603-42854603 on Build GRCh38
rs375518933 Chr.21: 42854618-42854618 on Build GRCh38
rs574347863 Chr.21: 42854631-42854631 on Build GRCh38
rs569127512 Chr.21: 42854673-42854673 on Build GRCh38
rs751961120 Chr.21: 42854510-42854510 on Build GRCh38
rs751039981 Chr.21: 42854641-42854641 on Build GRCh38
rs986142254 Chr.21: 42854583-42854583 on Build GRCh38
rs757560527 Chr.21: 42854550-42854550 on Build GRCh38
rs1051048359 Chr.21: 42854782-42854782 on Build GRCh38
rs376553203 Chr.21: 42854656-42854656 on Build GRCh38
rs149569805 Chr.21: 42854640-42854640 on Build GRCh38
rs866434656 Chr.21: 42854619-42854619 on Build GRCh38
rs1045481415 Chr.21: 42854679-42854679 on Build GRCh38
rs761691540 Chr.21: 42854556-42854556 on Build GRCh38
rs986810355 Chr.21: 42854499-42854499 on Build GRCh38
rs367663245 Chr.21: 42854615-42854615 on Build GRCh38
rs772891386 Chr.21: 42854559-42854559 on Build GRCh38
rs372394728 Chr.21: 42854848-42854848 on Build GRCh38
rs955075056 Chr.21: 42854658-42854658 on Build GRCh38
rs147779866 Chr.21: 42854592-42854592 on Build GRCh38
rs566709496 Chr.21: 42854723-42854723 on Build GRCh38
rs956267467 Chr.21: 42854886-42854886 on Build GRCh38
rs751918320 Chr.21: 42854633-42854633 on Build GRCh38
rs754598104 Chr.21: 42854601-42854601 on Build GRCh38
rs1031164971 Chr.21: 42854885-42854885 on Build GRCh38
rs201736090 Chr.21: 42854572-42854572 on Build GRCh38
rs953497327 Chr.21: 42854749-42854749 on Build GRCh38
rs1038621449 Chr.21: 42854746-42854746 on Build GRCh38
rs1008017109 Chr.21: 42854766-42854766 on Build GRCh38
rs62215010 Chr.21: 42854697-42854697 on Build GRCh38
rs891021370 Chr.21: 42854800-42854800 on Build GRCh38
rs2298666 Chr.21: 42854532-42854532 on Build GRCh38
rs1006153868 Chr.21: 42854701-42854701 on Build GRCh38
rs1000580833 Chr.21: 42854884-42854884 on Build GRCh38
rs373669142 Chr.21: 42854730-42854730 on Build GRCh38
rs778512946 Chr.21: 42854649-42854649 on Build GRCh38
rs763182331 Chr.21: 42854629-42854629 on Build GRCh38
rs770778502 Chr.21: 42854671-42854671 on Build GRCh38
rs759606940 Chr.21: 42854620-42854620 on Build GRCh38
rs776652538 Chr.21: 42854566-42854566 on Build GRCh38
rs980114194 Chr.21: 42854920-42854921 on Build GRCh38
rs781033216 Chr.21: 42854708-42854708 on Build GRCh38
rs1032363106 Chr.21: 42854777-42854777 on Build GRCh38
rs747656609 Chr.21: 42854709-42854709 on Build GRCh38
rs758552318 Chr.21: 42854573-42854573 on Build GRCh38
rs768898240 Chr.21: 42854678-42854678 on Build GRCh38
rs1044533826 Chr.21: 42854604-42854604 on Build GRCh38
rs1055465221 Chr.21: 42854684-42854684 on Build GRCh38
rs774199021 Chr.21: 42854553-42854553 on Build GRCh38
rs756697954 Chr.21: 42854646-42854646 on Build GRCh38
rs779659365 Chr.21: 42854544-42854544 on Build GRCh38
rs543989584 Chr.21: 42854591-42854591 on Build GRCh38
rs928403450 Chr.21: 42854680-42854680 on Build GRCh38
rs464854 Chr.21: 42854923-42854923 on Build GRCh38
rs779137633 Chr.21: 42854517-42854517 on Build GRCh38
rs200123512 Chr.21: 42854676-42854676 on Build GRCh38
rs762979207 Chr.21: 42854677-42854677 on Build GRCh38
rs770871520 Chr.21: 42854617-42854617 on Build GRCh38
rs775559748 Chr.21: 42854628-42854628 on Build GRCh38
rs896949262 Chr.21: 42854750-42854750 on Build GRCh38
rs549530535 Chr.21: 42854887-42854887 on Build GRCh38
rs765233947 Chr.21: 42854625-42854625 on Build GRCh38
rs532557737 Chr.21: 42854581-42854581 on Build GRCh38
rs753030086 Chr.21: 42854522-42854522 on Build GRCh38
rs758855785 Chr.21: 42854526-42854526 on Build GRCh38
rs759653974 Chr.21: 42854567-42854567 on Build GRCh38
rs779672432 Chr.21: 42854674-42854674 on Build GRCh38
rs961844200 Chr.21: 42854562-42854562 on Build GRCh38
rs566044049 Chr.21: 42854552-42854552 on Build GRCh38
rs367904468 Chr.21: 42854657-42854657 on Build GRCh38
rs1044399276 Chr.21: 42854719-42854719 on Build GRCh38
rs747635327 Chr.21: 42854651-42854651 on Build GRCh38
rs558641644 Chr.21: 42854784-42854784 on Build GRCh38
rs747772807 Chr.21: 42854609-42854609 on Build GRCh38
rs553853128 Chr.21: 42854551-42854551 on Build GRCh38
rs377106680 Chr.21: 42854549-42854549 on Build GRCh38
rs931987803 Chr.21: 42854491-42854491 on Build GRCh38
rs1029303779 Chr.21: 42854500-42854500 on Build GRCh38
rs757609425 Chr.21: 42854636-42854636 on Build GRCh38
rs763304797 Chr.21: 42854740-42854740 on Build GRCh38
rs763720766 Chr.21: 42854663-42854675 on Build GRCh38
rs925241616 Chr.21: 42854613-42854613 on Build GRCh38
rs199713565 Chr.21: 42854616-42854616 on Build GRCh38
rs988383502 Chr.21: 42854824-42854824 on Build GRCh38
rs746822433 Chr.21: 42854683-42854683 on Build GRCh38
rs1006528091 Chr.21: 42854810-42854810 on Build GRCh38
rs548385277 Chr.21: 42854721-42854721 on Build GRCh38
rs772723860 Chr.21: 42854839-42854839 on Build GRCh38
rs367704779 Chr.21: 42854558-42854558 on Build GRCh38
rs567974305 Chr.21: 42854508-42854508 on Build GRCh38
rs370186752 Chr.21: 42854563-42854563 on Build GRCh38
rs755146161 Chr.21: 42854514-42854514 on Build GRCh38
rs138194752 Chr.21: 42854564-42854564 on Build GRCh38
rs111539710 Chr.21: 42854590-42854590 on Build GRCh38
rs536367076 Chr.21: 42854798-42854799 on Build GRCh38
rs917529039 Chr.21: 42854903-42854903 on Build GRCh38
rs1017867126 Chr.21: 42854823-42854823 on Build GRCh38
rs774940569 Chr.21: 42854599-42854599 on Build GRCh38
rs765431906 Chr.21: 42854569-42854569 on Build GRCh38
rs140790002 Chr.21: 42854576-42854576 on Build GRCh38
rs563834005 Chr.21: 42854715-42854715 on Build GRCh38
rs145747081 Chr.21: 42854577-42854577 on Build GRCh38
rs762681235 Chr.21: 42854635-42854637 on Build GRCh38
rs1107831 Chr.21: 42854735-42854735 on Build GRCh38
rs943567107 Chr.21: 42854541-42854541 on Build GRCh38
rs777100186 Chr.21: 42854539-42854540 on Build GRCh38
rs373989563 Chr.21: 42854540-42854540 on Build GRCh38
rs376829512 Chr.21: 42854531-42854531 on Build GRCh38
rs771503114 Chr.21: 42854868-42854868 on Build GRCh38
rs757740830 Chr.21: 42854585-42854585 on Build GRCh38
rs970911915 Chr.21: 42854898-42854898 on Build GRCh38
rs148700675 Chr.21: 42854910-42854910 on Build GRCh38

View Full Product Details