Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45456012-45456502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL18A1
Entrez Gene ID
80781
Gene Name
collagen type XVIII alpha 1 chain
Gene Aliases
KNO, KNO1, KS
UniGene
Hs.517356
Gene Chromosome Location
Chr.21: 45405137-45513720 on Build GRCh38
Transcripts
NM_030582.3 NM_130444.2 NM_130445.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6632683 c.617C>T p.T206M Chr.21: 45456147-45456147 on Build GRCh38
6333780 c.518G>T p.G173V Chr.21: 45456048-45456048 on Build GRCh38
4501385 c.587C>T p.P196L Chr.21: 45456117-45456117 on Build GRCh38
1592797 c.861C>T p.G287G Chr.21: 45456391-45456391 on Build GRCh38
3551580 c.590C>T p.S197F Chr.21: 45456120-45456120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760993489 Chr.21: 45456478-45456478 on Build GRCh38
rs956658327 Chr.21: 45456243-45456243 on Build GRCh38
rs760202762 Chr.21: 45456141-45456141 on Build GRCh38
rs746650638 Chr.21: 45456267-45456267 on Build GRCh38
rs200284308 Chr.21: 45456045-45456045 on Build GRCh38
rs868032921 Chr.21: 45456230-45456230 on Build GRCh38
rs760683399 Chr.21: 45456088-45456088 on Build GRCh38
rs376644375 Chr.21: 45456308-45456308 on Build GRCh38
rs1005409090 Chr.21: 45456185-45456185 on Build GRCh38
rs772165396 Chr.21: 45456061-45456061 on Build GRCh38
rs748461576 Chr.21: 45456347-45456347 on Build GRCh38
rs771496335 Chr.21: 45456064-45456064 on Build GRCh38
rs770112416 Chr.21: 45456123-45456123 on Build GRCh38
rs988273624 Chr.21: 45456433-45456433 on Build GRCh38
rs185291015 Chr.21: 45456407-45456407 on Build GRCh38
rs1047563425 Chr.21: 45456465-45456465 on Build GRCh38
rs771686771 Chr.21: 45456137-45456137 on Build GRCh38
rs774274082 Chr.21: 45456406-45456406 on Build GRCh38
rs533274673 Chr.21: 45456367-45456367 on Build GRCh38
rs73386835 Chr.21: 45456067-45456067 on Build GRCh38
rs760431847 Chr.21: 45456199-45456199 on Build GRCh38
rs189589098 Chr.21: 45456242-45456242 on Build GRCh38
rs758849391 Chr.21: 45456356-45456356 on Build GRCh38
rs1045315350 Chr.21: 45456344-45456344 on Build GRCh38
rs577697894 Chr.21: 45456277-45456277 on Build GRCh38
rs181274677 Chr.21: 45456263-45456263 on Build GRCh38
rs563547981 Chr.21: 45456395-45456395 on Build GRCh38
rs780365285 Chr.21: 45456262-45456262 on Build GRCh38
rs754114479 Chr.21: 45456426-45456426 on Build GRCh38
rs777802002 Chr.21: 45456167-45456167 on Build GRCh38
rs185215745 Chr.21: 45456196-45456196 on Build GRCh38
rs753055018 Chr.21: 45456063-45456063 on Build GRCh38
rs552922627 Chr.21: 45456276-45456276 on Build GRCh38
rs905567567 Chr.21: 45456238-45456238 on Build GRCh38
rs946947490 Chr.21: 45456340-45456340 on Build GRCh38
rs758609359 Chr.21: 45456111-45456111 on Build GRCh38
rs573085272 Chr.21: 45456303-45456303 on Build GRCh38
rs11702494 Chr.21: 45456392-45456392 on Build GRCh38
rs201419516 Chr.21: 45456130-45456130 on Build GRCh38
rs374441004 Chr.21: 45456298-45456298 on Build GRCh38
rs534367236 Chr.21: 45456269-45456269 on Build GRCh38
rs930039985 Chr.21: 45456286-45456286 on Build GRCh38
rs749957649 Chr.21: 45456080-45456080 on Build GRCh38
rs557847592 Chr.21: 45456368-45456368 on Build GRCh38
rs1042697947 Chr.21: 45456205-45456205 on Build GRCh38
rs745318918 Chr.21: 45456270-45456270 on Build GRCh38
rs963368953 Chr.21: 45456105-45456105 on Build GRCh38
rs373353645 Chr.21: 45456070-45456070 on Build GRCh38
rs751750214 Chr.21: 45456253-45456253 on Build GRCh38
rs905515344 Chr.21: 45456355-45456355 on Build GRCh38
rs549161026 Chr.21: 45456434-45456434 on Build GRCh38
rs767571894 Chr.21: 45456256-45456256 on Build GRCh38
rs142892701 Chr.21: 45456090-45456090 on Build GRCh38
rs930096071 Chr.21: 45456427-45456427 on Build GRCh38
rs770343212 Chr.21: 45456182-45456182 on Build GRCh38
rs62000960 Chr.21: 45456047-45456047 on Build GRCh38
rs199725915 Chr.21: 45456131-45456131 on Build GRCh38
rs764239495 Chr.21: 45456394-45456394 on Build GRCh38
rs770778212 Chr.21: 45456268-45456268 on Build GRCh38
rs780945725 Chr.21: 45456358-45456358 on Build GRCh38
rs761412846 Chr.21: 45456073-45456073 on Build GRCh38
rs773218086 Chr.21: 45456062-45456062 on Build GRCh38
rs746995849 Chr.21: 45456113-45456113 on Build GRCh38
rs372854150 Chr.21: 45456112-45456112 on Build GRCh38
rs959202481 Chr.21: 45456396-45456396 on Build GRCh38
rs931441771 Chr.21: 45456469-45456490 on Build GRCh38
rs537362206 Chr.21: 45456250-45456250 on Build GRCh38
rs746406313 Chr.21: 45456121-45456121 on Build GRCh38
rs752503890 Chr.21: 45456163-45456163 on Build GRCh38
rs772920092 Chr.21: 45456138-45456138 on Build GRCh38
rs1028927736 Chr.21: 45456387-45456387 on Build GRCh38
rs774188278 Chr.21: 45456289-45456289 on Build GRCh38
rs994732533 Chr.21: 45456377-45456377 on Build GRCh38
rs538715485 Chr.21: 45456290-45456290 on Build GRCh38
rs769156194 Chr.21: 45456369-45456369 on Build GRCh38
rs763981744 Chr.21: 45456252-45456252 on Build GRCh38
rs956505177 Chr.21: 45456364-45456364 on Build GRCh38
rs953325335 Chr.21: 45456391-45456391 on Build GRCh38
rs752945899 Chr.21: 45456107-45456107 on Build GRCh38
rs746351498 Chr.21: 45456181-45456181 on Build GRCh38
rs76871966 Chr.21: 45456437-45456437 on Build GRCh38
rs557208339 Chr.21: 45456291-45456291 on Build GRCh38
rs2236452 Chr.21: 45456259-45456259 on Build GRCh38
rs201773944 Chr.21: 45456032-45456032 on Build GRCh38
rs78240331 Chr.21: 45456312-45456312 on Build GRCh38
rs1028978765 Chr.21: 45456281-45456281 on Build GRCh38
rs773381032 Chr.21: 45456424-45456424 on Build GRCh38
rs766006804 Chr.21: 45456148-45456148 on Build GRCh38
rs751830446 Chr.21: 45456168-45456168 on Build GRCh38
rs568717643 Chr.21: 45456057-45456057 on Build GRCh38
rs755673225 Chr.21: 45456148-45456148 on Build GRCh38
rs780511511 Chr.21: 45456360-45456360 on Build GRCh38
rs779419485 Chr.21: 45456313-45456313 on Build GRCh38
rs915378479 Chr.21: 45456452-45456452 on Build GRCh38
rs754264615 Chr.21: 45456307-45456307 on Build GRCh38
rs200930983 Chr.21: 45456224-45456224 on Build GRCh38
rs775116003 Chr.21: 45456383-45456383 on Build GRCh38
rs542947545 Chr.21: 45456299-45456299 on Build GRCh38
rs771013645 Chr.21: 45456114-45456114 on Build GRCh38
rs561076797 Chr.21: 45456302-45456302 on Build GRCh38
rs748336591 Chr.21: 45456048-45456048 on Build GRCh38
rs771874035 Chr.21: 45456418-45456418 on Build GRCh38
rs779372007 Chr.21: 45456266-45456266 on Build GRCh38
rs530848809 Chr.21: 45456091-45456091 on Build GRCh38
rs989023296 Chr.21: 45456251-45456252 on Build GRCh38
rs199717521 Chr.21: 45456118-45456118 on Build GRCh38
rs1057181306 Chr.21: 45456147-45456147 on Build GRCh38
rs756054515 Chr.21: 45456043-45456043 on Build GRCh38
rs757523045 Chr.21: 45456255-45456255 on Build GRCh38
rs369474928 Chr.21: 45456101-45456101 on Build GRCh38
rs780460277 Chr.21: 45456188-45456188 on Build GRCh38
rs763775854 Chr.21: 45456284-45456284 on Build GRCh38
rs770530671 Chr.21: 45456209-45456209 on Build GRCh38
rs756222197 Chr.21: 45456260-45456260 on Build GRCh38
rs781289837 Chr.21: 45456175-45456175 on Build GRCh38
rs2236451 Chr.21: 45456169-45456169 on Build GRCh38
rs765021555 Chr.21: 45456237-45456237 on Build GRCh38
rs533453650 Chr.21: 45456195-45456195 on Build GRCh38
rs368985827 Chr.21: 45456044-45456044 on Build GRCh38
rs767031366 Chr.21: 45456079-45456079 on Build GRCh38
rs559679925 Chr.21: 45456357-45456357 on Build GRCh38
rs899510965 Chr.21: 45456309-45456309 on Build GRCh38
rs547769296 Chr.21: 45456150-45456150 on Build GRCh38
rs758219432 Chr.21: 45456165-45456165 on Build GRCh38
rs754435895 Chr.21: 45456460-45456461 on Build GRCh38
rs759599651 Chr.21: 45456153-45456153 on Build GRCh38
rs765071259 Chr.21: 45456160-45456160 on Build GRCh38
rs762988380 Chr.21: 45456282-45456282 on Build GRCh38
rs148957774 Chr.21: 45456068-45456068 on Build GRCh38
rs749547449 Chr.21: 45456192-45456192 on Build GRCh38
rs776343723 Chr.21: 45456212-45456212 on Build GRCh38

View Full Product Details