Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23825103-23825596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMARCB1
Entrez Gene ID
6598
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1
Gene Aliases
BAF47, CSS3, INI1, MRD15, PPP1R144, RDT, RTPS1, SNF5, SNF5L1, SWNTS1, Sfh1p, Snr1, hSNFS
UniGene
Hs.534350
Gene Chromosome Location
Chr.22: 23786931-23834518 on Build GRCh38
Transcripts
NM_001007468.2 NM_001317946.1 NM_003073.4 XM_011530345.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1009 c.897G>A p.S299S Chr.22: 23825326-23825326 on Build GRCh38
327308 c.891C>A p.N297K Chr.22: 23825293-23825293 on Build GRCh38
3964140 c.855G>C p.E285D Chr.22: 23825284-23825284 on Build GRCh38
4766059 c.906G>A p.G302G Chr.22: 23825335-23825335 on Build GRCh38
402259 c.1005C>A p.Y335* Chr.22: 23825407-23825407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112493794 Chr.22: 23825515-23825515 on Build GRCh38
rs565395305 Chr.22: 23825196-23825196 on Build GRCh38
rs973142334 Chr.22: 23825172-23825172 on Build GRCh38
rs138735048 Chr.22: 23825281-23825281 on Build GRCh38
rs775635513 Chr.22: 23825189-23825189 on Build GRCh38
rs753334172 Chr.22: 23825269-23825269 on Build GRCh38
rs546466028 Chr.22: 23825466-23825466 on Build GRCh38
rs891918145 Chr.22: 23825468-23825468 on Build GRCh38
rs764318057 Chr.22: 23825191-23825191 on Build GRCh38
rs766845537 Chr.22: 23825443-23825443 on Build GRCh38
rs1030998446 Chr.22: 23825221-23825221 on Build GRCh38
rs34624937 Chr.22: 23825558-23825560 on Build GRCh38
rs749348930 Chr.22: 23825363-23825363 on Build GRCh38
rs868003456 Chr.22: 23825516-23825516 on Build GRCh38
rs756643970 Chr.22: 23825212-23825212 on Build GRCh38
rs530036261 Chr.22: 23825462-23825462 on Build GRCh38
rs774688910 Chr.22: 23825479-23825479 on Build GRCh38
rs377260020 Chr.22: 23825179-23825179 on Build GRCh38
rs35838942 Chr.22: 23825163-23825163 on Build GRCh38
rs370768337 Chr.22: 23825318-23825318 on Build GRCh38
rs781521647 Chr.22: 23825149-23825149 on Build GRCh38
rs5844569 Chr.22: 23825470-23825471 on Build GRCh38
rs397897182 Chr.22: 23825473-23825474 on Build GRCh38
rs773617664 Chr.22: 23825424-23825424 on Build GRCh38
rs1053215058 Chr.22: 23825365-23825365 on Build GRCh38
rs760381148 Chr.22: 23825501-23825501 on Build GRCh38
rs765865422 Chr.22: 23825216-23825216 on Build GRCh38
rs561124304 Chr.22: 23825432-23825432 on Build GRCh38
rs35702954 Chr.22: 23825556-23825556 on Build GRCh38
rs587776677 Chr.22: 23825380-23825380 on Build GRCh38
rs985748293 Chr.22: 23825162-23825162 on Build GRCh38
rs753696710 Chr.22: 23825542-23825542 on Build GRCh38
rs769322487 Chr.22: 23825317-23825317 on Build GRCh38
rs963557868 Chr.22: 23825359-23825359 on Build GRCh38
rs768065617 Chr.22: 23825410-23825410 on Build GRCh38
rs761102377 Chr.22: 23825439-23825439 on Build GRCh38
rs4050151 Chr.22: 23825565-23825567 on Build GRCh38
rs573456379 Chr.22: 23825169-23825169 on Build GRCh38
rs150722737 Chr.22: 23825362-23825362 on Build GRCh38
rs200362024 Chr.22: 23825566-23825568 on Build GRCh38
rs767971910 Chr.22: 23825199-23825199 on Build GRCh38
rs919036744 Chr.22: 23825476-23825478 on Build GRCh38
rs777512879 Chr.22: 23825335-23825335 on Build GRCh38
rs769023941 Chr.22: 23825375-23825375 on Build GRCh38
rs374509291 Chr.22: 23825341-23825341 on Build GRCh38
rs185465037 Chr.22: 23825573-23825573 on Build GRCh38
rs766234454 Chr.22: 23825522-23825522 on Build GRCh38
rs772576999 Chr.22: 23825220-23825221 on Build GRCh38
rs754587817 Chr.22: 23825284-23825284 on Build GRCh38
rs774763517 Chr.22: 23825404-23825404 on Build GRCh38
rs1007116669 Chr.22: 23825520-23825520 on Build GRCh38
rs545593020 Chr.22: 23825171-23825171 on Build GRCh38
rs970791302 Chr.22: 23825489-23825489 on Build GRCh38
rs762220738 Chr.22: 23825197-23825197 on Build GRCh38
rs112038099 Chr.22: 23825416-23825416 on Build GRCh38
rs2229354 Chr.22: 23825326-23825326 on Build GRCh38
rs754484256 Chr.22: 23825557-23825557 on Build GRCh38
rs747013879 Chr.22: 23825180-23825181 on Build GRCh38
rs937838534 Chr.22: 23825154-23825154 on Build GRCh38
rs770886017 Chr.22: 23825353-23825353 on Build GRCh38
rs954479313 Chr.22: 23825148-23825148 on Build GRCh38
rs903318541 Chr.22: 23825562-23825562 on Build GRCh38
rs187488637 Chr.22: 23825407-23825407 on Build GRCh38
rs769803365 Chr.22: 23825187-23825187 on Build GRCh38
rs981838241 Chr.22: 23825499-23825499 on Build GRCh38
rs760635703 Chr.22: 23825178-23825178 on Build GRCh38
rs542121952 Chr.22: 23825164-23825164 on Build GRCh38

View Full Product Details