Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38078427-38078917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC16A8
Entrez Gene ID
23539
Gene Name
solute carrier family 16 member 8
Gene Aliases
MCT3, REMP
UniGene
Hs.745256
Gene Chromosome Location
Chr.22: 38078137-38083163 on Build GRCh38
Transcripts
NM_013356.2 XM_017028685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4956948 c.1338C>A p.G446G Chr.22: 38078565-38078565 on Build GRCh38
4801519 c.1403T>G p.V468G Chr.22: 38078500-38078500 on Build GRCh38
1416218 c.1287C>T p.V429V Chr.22: 38078616-38078616 on Build GRCh38
1416217 c.1450G>A p.A484T Chr.22: 38078453-38078453 on Build GRCh38
300162 c.1454G>A p.R485Q Chr.22: 38078449-38078449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569433346 Chr.22: 38078479-38078479 on Build GRCh38
rs971659913 Chr.22: 38078498-38078498 on Build GRCh38
rs201452730 Chr.22: 38078688-38078688 on Build GRCh38
rs747546295 Chr.22: 38078744-38078744 on Build GRCh38
rs561077276 Chr.22: 38078868-38078868 on Build GRCh38
rs867253868 Chr.22: 38078644-38078644 on Build GRCh38
rs201036628 Chr.22: 38078471-38078471 on Build GRCh38
rs1004706407 Chr.22: 38078794-38078794 on Build GRCh38
rs149067515 Chr.22: 38078613-38078613 on Build GRCh38
rs761416594 Chr.22: 38078578-38078578 on Build GRCh38
rs373468406 Chr.22: 38078591-38078591 on Build GRCh38
rs779227503 Chr.22: 38078624-38078624 on Build GRCh38
rs764747371 Chr.22: 38078581-38078581 on Build GRCh38
rs200389946 Chr.22: 38078615-38078615 on Build GRCh38
rs758540386 Chr.22: 38078626-38078626 on Build GRCh38
rs748304965 Chr.22: 38078520-38078520 on Build GRCh38
rs768444330 Chr.22: 38078740-38078742 on Build GRCh38
rs200082757 Chr.22: 38078571-38078571 on Build GRCh38
rs761167954 Chr.22: 38078648-38078648 on Build GRCh38
rs773450414 Chr.22: 38078535-38078535 on Build GRCh38
rs779928832 Chr.22: 38078708-38078709 on Build GRCh38
rs764758514 Chr.22: 38078654-38078654 on Build GRCh38
rs2076371 Chr.22: 38078689-38078689 on Build GRCh38
rs752675265 Chr.22: 38078614-38078614 on Build GRCh38
rs750111343 Chr.22: 38078582-38078582 on Build GRCh38
rs772824497 Chr.22: 38078751-38078751 on Build GRCh38
rs35481940 Chr.22: 38078892-38078892 on Build GRCh38
rs565479077 Chr.22: 38078446-38078446 on Build GRCh38
rs746285383 Chr.22: 38078625-38078625 on Build GRCh38
rs774855917 Chr.22: 38078460-38078460 on Build GRCh38
rs766268798 Chr.22: 38078596-38078596 on Build GRCh38
rs1004764 Chr.22: 38078845-38078845 on Build GRCh38
rs572012866 Chr.22: 38078702-38078702 on Build GRCh38
rs779137755 Chr.22: 38078708-38078708 on Build GRCh38
rs746168595 Chr.22: 38078731-38078731 on Build GRCh38
rs749045610 Chr.22: 38078722-38078724 on Build GRCh38
rs765389705 Chr.22: 38078464-38078465 on Build GRCh38
rs983325337 Chr.22: 38078534-38078534 on Build GRCh38
rs540924144 Chr.22: 38078741-38078741 on Build GRCh38
rs755006883 Chr.22: 38078510-38078510 on Build GRCh38
rs981716487 Chr.22: 38078517-38078518 on Build GRCh38
rs762588282 Chr.22: 38078756-38078756 on Build GRCh38
rs557837757 Chr.22: 38078660-38078660 on Build GRCh38
rs774765843 Chr.22: 38078542-38078542 on Build GRCh38
rs567347112 Chr.22: 38078577-38078577 on Build GRCh38
rs377373294 Chr.22: 38078454-38078454 on Build GRCh38
rs550895893 Chr.22: 38078462-38078462 on Build GRCh38
rs772417968 Chr.22: 38078642-38078642 on Build GRCh38
rs769870593 Chr.22: 38078532-38078532 on Build GRCh38
rs974883817 Chr.22: 38078818-38078821 on Build GRCh38
rs762678368 Chr.22: 38078659-38078659 on Build GRCh38
rs990714160 Chr.22: 38078480-38078480 on Build GRCh38
rs144999316 Chr.22: 38078519-38078519 on Build GRCh38
rs776087361 Chr.22: 38078647-38078647 on Build GRCh38
rs749599605 Chr.22: 38078541-38078541 on Build GRCh38
rs751909314 Chr.22: 38078832-38078832 on Build GRCh38
rs112227645 Chr.22: 38078879-38078879 on Build GRCh38
rs374272776 Chr.22: 38078733-38078733 on Build GRCh38
rs748310060 Chr.22: 38078449-38078449 on Build GRCh38
rs756065931 Chr.22: 38078709-38078711 on Build GRCh38
rs769342488 Chr.22: 38078748-38078748 on Build GRCh38
rs201652278 Chr.22: 38078655-38078655 on Build GRCh38
rs759401754 Chr.22: 38078684-38078684 on Build GRCh38
rs990245913 Chr.22: 38078895-38078895 on Build GRCh38
rs369032590 Chr.22: 38078668-38078668 on Build GRCh38
rs1002887993 Chr.22: 38078673-38078673 on Build GRCh38
rs574936978 Chr.22: 38078811-38078811 on Build GRCh38
rs758288019 Chr.22: 38078494-38078494 on Build GRCh38
rs771201997 Chr.22: 38078619-38078619 on Build GRCh38
rs780080887 Chr.22: 38078499-38078499 on Build GRCh38
rs753019610 Chr.22: 38078621-38078621 on Build GRCh38
rs780475697 Chr.22: 38078785-38078785 on Build GRCh38
rs746945015 Chr.22: 38078508-38078508 on Build GRCh38
rs200126325 Chr.22: 38078450-38078450 on Build GRCh38
rs371223340 Chr.22: 38078682-38078682 on Build GRCh38
rs757440454 Chr.22: 38078706-38078706 on Build GRCh38
rs538880672 Chr.22: 38078790-38078790 on Build GRCh38
rs763509384 Chr.22: 38078466-38078466 on Build GRCh38
rs759948243 Chr.22: 38078555-38078555 on Build GRCh38
rs756076991 Chr.22: 38078700-38078700 on Build GRCh38
rs758199673 Chr.22: 38078593-38078593 on Build GRCh38
rs919511239 Chr.22: 38078743-38078743 on Build GRCh38
rs778563367 Chr.22: 38078745-38078754 on Build GRCh38
rs901874649 Chr.22: 38078632-38078632 on Build GRCh38
rs369688090 Chr.22: 38078458-38078458 on Build GRCh38
rs751613208 Chr.22: 38078706-38078707 on Build GRCh38
rs777819488 Chr.22: 38078701-38078701 on Build GRCh38
rs191863883 Chr.22: 38078808-38078808 on Build GRCh38
rs777866924 Chr.22: 38078631-38078632 on Build GRCh38
rs183953135 Chr.22: 38078816-38078816 on Build GRCh38
rs754845253 Chr.22: 38078611-38078611 on Build GRCh38
rs149941944 Chr.22: 38078483-38078483 on Build GRCh38
rs763344401 Chr.22: 38078455-38078455 on Build GRCh38
rs1049188249 Chr.22: 38078570-38078570 on Build GRCh38
rs751454907 Chr.22: 38078597-38078597 on Build GRCh38
rs756837794 Chr.22: 38078475-38078475 on Build GRCh38
rs1014738006 Chr.22: 38078798-38078798 on Build GRCh38
rs537158986 Chr.22: 38078484-38078484 on Build GRCh38

View Full Product Details