Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47622082-47622595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMARCC1
Entrez Gene ID
6599
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1
Gene Aliases
BAF155, CRACC1, Rsc8, SRG3, SWI3
UniGene
Hs.476179
Gene Chromosome Location
Chr.3: 47585888-47781915 on Build GRCh38
Transcripts
NM_003074.3 XM_011534034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6097888 c.2669G>C p.R890T Chr.3: 47622319-47622319 on Build GRCh38
7152101 c.2665G>C p.E889Q Chr.3: 47622323-47622323 on Build GRCh38
4152464 c.2674A>G p.I892V Chr.3: 47622314-47622314 on Build GRCh38
6784986 c.2735G>A p.R912Q Chr.3: 47622253-47622253 on Build GRCh38
7152100 c.2715G>C p.K905N Chr.3: 47622273-47622273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968932935 Chr.3: 47622449-47622449 on Build GRCh38
rs773745845 Chr.3: 47622136-47622136 on Build GRCh38
rs948182779 Chr.3: 47622470-47622470 on Build GRCh38
rs757303353 Chr.3: 47622197-47622197 on Build GRCh38
rs140035331 Chr.3: 47622267-47622267 on Build GRCh38
rs533879628 Chr.3: 47622540-47622540 on Build GRCh38
rs145327328 Chr.3: 47622448-47622448 on Build GRCh38
rs749259441 Chr.3: 47622163-47622163 on Build GRCh38
rs948346200 Chr.3: 47622114-47622114 on Build GRCh38
rs373975345 Chr.3: 47622348-47622348 on Build GRCh38
rs752386588 Chr.3: 47622273-47622273 on Build GRCh38
rs987138455 Chr.3: 47622469-47622469 on Build GRCh38
rs773306917 Chr.3: 47622389-47622389 on Build GRCh38
rs146336326 Chr.3: 47622332-47622332 on Build GRCh38
rs1004248867 Chr.3: 47622364-47622364 on Build GRCh38
rs978880192 Chr.3: 47622452-47622452 on Build GRCh38
rs370321862 Chr.3: 47622186-47622186 on Build GRCh38
rs968494064 Chr.3: 47622244-47622244 on Build GRCh38
rs939614133 Chr.3: 47622558-47622558 on Build GRCh38
rs570787075 Chr.3: 47622153-47622153 on Build GRCh38
rs960301162 Chr.3: 47622404-47622404 on Build GRCh38
rs924394314 Chr.3: 47622460-47622460 on Build GRCh38
rs754254733 Chr.3: 47622552-47622552 on Build GRCh38
rs981056831 Chr.3: 47622510-47622510 on Build GRCh38
rs911845216 Chr.3: 47622155-47622155 on Build GRCh38
rs778692102 Chr.3: 47622357-47622357 on Build GRCh38
rs771650118 Chr.3: 47622222-47622222 on Build GRCh38
rs757103087 Chr.3: 47622165-47622165 on Build GRCh38
rs768982036 Chr.3: 47622178-47622178 on Build GRCh38
rs763701944 Chr.3: 47622318-47622318 on Build GRCh38
rs745713299 Chr.3: 47622373-47622373 on Build GRCh38
rs551796701 Chr.3: 47622122-47622122 on Build GRCh38
rs758120877 Chr.3: 47622378-47622378 on Build GRCh38
rs1021441668 Chr.3: 47622228-47622228 on Build GRCh38
rs745592166 Chr.3: 47622174-47622174 on Build GRCh38
rs535019779 Chr.3: 47622411-47622411 on Build GRCh38
rs769932085 Chr.3: 47622387-47622387 on Build GRCh38
rs765316683 Chr.3: 47622351-47622352 on Build GRCh38
rs148929306 Chr.3: 47622445-47622445 on Build GRCh38
rs185037010 Chr.3: 47622117-47622117 on Build GRCh38
rs769846386 Chr.3: 47622198-47622198 on Build GRCh38
rs778774219 Chr.3: 47622169-47622169 on Build GRCh38
rs767347785 Chr.3: 47622270-47622270 on Build GRCh38
rs750648415 Chr.3: 47622376-47622377 on Build GRCh38
rs557133065 Chr.3: 47622520-47622520 on Build GRCh38
rs757239864 Chr.3: 47622354-47622354 on Build GRCh38
rs763516708 Chr.3: 47622252-47622252 on Build GRCh38
rs375971966 Chr.3: 47622182-47622182 on Build GRCh38
rs553266135 Chr.3: 47622424-47622424 on Build GRCh38
rs943320270 Chr.3: 47622555-47622555 on Build GRCh38
rs373548235 Chr.3: 47622160-47622160 on Build GRCh38
rs541575568 Chr.3: 47622383-47622383 on Build GRCh38
rs755770734 Chr.3: 47622300-47622300 on Build GRCh38
rs759035316 Chr.3: 47622162-47622162 on Build GRCh38
rs766734543 Chr.3: 47622253-47622253 on Build GRCh38
rs760376844 Chr.3: 47622464-47622464 on Build GRCh38
rs983844767 Chr.3: 47622453-47622453 on Build GRCh38

View Full Product Details