Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56994531-56995070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR2B
Entrez Gene ID
5431
Gene Name
RNA polymerase II subunit B
Gene Aliases
POL2RB, RPB2, hRPB140
UniGene
Hs.602757
Gene Chromosome Location
Chr.4: 56978640-57031162 on Build GRCh38
Transcripts
NM_000938.2 NM_001303268.1 NM_001303269.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5995547 c.397A>G p.I133V Chr.4: 56994687-56994687 on Build GRCh38
6878566 c.541C>T p.P181S Chr.4: 56994831-56994831 on Build GRCh38
1056364 c.421C>G p.Q141E Chr.4: 56994711-56994711 on Build GRCh38
4922221 c.400A>G p.K134E Chr.4: 56994690-56994690 on Build GRCh38
4417857 c.406G>T p.G136C Chr.4: 56994696-56994696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755585636 Chr.4: 56994616-56994616 on Build GRCh38
rs781004445 Chr.4: 56994670-56994670 on Build GRCh38
rs56888342 Chr.4: 56994945-56994946 on Build GRCh38
rs78191447 Chr.4: 56994939-56994939 on Build GRCh38
rs113145499 Chr.4: 56994892-56994892 on Build GRCh38
rs773594158 Chr.4: 56994700-56994700 on Build GRCh38
rs982369812 Chr.4: 56994964-56994964 on Build GRCh38
rs41560115 Chr.4: 56994939-56994946 on Build GRCh38
rs763760157 Chr.4: 56994770-56994770 on Build GRCh38
rs570633791 Chr.4: 56994901-56994901 on Build GRCh38
rs144234799 Chr.4: 56994818-56994818 on Build GRCh38
rs749556740 Chr.4: 56994939-56994939 on Build GRCh38
rs766824339 Chr.4: 56994707-56994707 on Build GRCh38
rs747215044 Chr.4: 56994874-56994874 on Build GRCh38
rs755996731 Chr.4: 56994932-56994939 on Build GRCh38
rs758067808 Chr.4: 56994636-56994636 on Build GRCh38
rs781196327 Chr.4: 56994930-56994937 on Build GRCh38
rs754304123 Chr.4: 56994708-56994708 on Build GRCh38
rs769673916 Chr.4: 56994673-56994673 on Build GRCh38
rs760842686 Chr.4: 56994911-56994915 on Build GRCh38
rs368609522 Chr.4: 56994947-56994947 on Build GRCh38
rs751181447 Chr.4: 56994779-56994779 on Build GRCh38
rs760107067 Chr.4: 56994728-56994728 on Build GRCh38
rs761084252 Chr.4: 56994701-56994701 on Build GRCh38
rs929785221 Chr.4: 56994751-56994751 on Build GRCh38
rs1055621830 Chr.4: 56994575-56994575 on Build GRCh38
rs201474145 Chr.4: 56994838-56994838 on Build GRCh38
rs7659855 Chr.4: 56994880-56994880 on Build GRCh38
rs772078945 Chr.4: 56994870-56994870 on Build GRCh38
rs1031685509 Chr.4: 56994945-56994945 on Build GRCh38
rs145350715 Chr.4: 56994957-56994957 on Build GRCh38
rs572121141 Chr.4: 56995044-56995044 on Build GRCh38
rs1713951 Chr.4: 56994951-56994951 on Build GRCh38
rs1008851563 Chr.4: 56994656-56994656 on Build GRCh38
rs979728652 Chr.4: 56994594-56994594 on Build GRCh38
rs766947033 Chr.4: 56994555-56994555 on Build GRCh38
rs752185601 Chr.4: 56994734-56994734 on Build GRCh38
rs778465434 Chr.4: 56994620-56994620 on Build GRCh38
rs201940173 Chr.4: 56994622-56994622 on Build GRCh38
rs748213016 Chr.4: 56994869-56994869 on Build GRCh38
rs184313866 Chr.4: 56995008-56995008 on Build GRCh38
rs757867254 Chr.4: 56994761-56994761 on Build GRCh38
rs771338153 Chr.4: 56994875-56994875 on Build GRCh38
rs145225188 Chr.4: 56994931-56994932 on Build GRCh38
rs776082383 Chr.4: 56994848-56994849 on Build GRCh38
rs773078271 Chr.4: 56994931-56994938 on Build GRCh38
rs375114461 Chr.4: 56994659-56994659 on Build GRCh38
rs548284937 Chr.4: 56994719-56994719 on Build GRCh38
rs780948955 Chr.4: 56994810-56994810 on Build GRCh38
rs1010383358 Chr.4: 56994940-56994951 on Build GRCh38
rs746756090 Chr.4: 56994657-56994657 on Build GRCh38
rs199707667 Chr.4: 56994947-56994947 on Build GRCh38
rs192612210 Chr.4: 56994978-56994978 on Build GRCh38
rs899217148 Chr.4: 56994593-56994597 on Build GRCh38
rs562550093 Chr.4: 56994692-56994692 on Build GRCh38
rs71889795 Chr.4: 56994931-56994931 on Build GRCh38
rs201408632 Chr.4: 56994929-56994929 on Build GRCh38
rs746229704 Chr.4: 56994623-56994624 on Build GRCh38
rs779866742 Chr.4: 56994867-56994867 on Build GRCh38
rs759826731 Chr.4: 56994884-56994884 on Build GRCh38
rs530202082 Chr.4: 56994946-56994946 on Build GRCh38
rs149588434 Chr.4: 56994773-56994773 on Build GRCh38
rs756952360 Chr.4: 56994801-56994801 on Build GRCh38
rs745781732 Chr.4: 56994671-56994671 on Build GRCh38
rs777297320 Chr.4: 56994642-56994642 on Build GRCh38
rs532686013 Chr.4: 56994567-56994567 on Build GRCh38
rs140288467 Chr.4: 56994696-56994696 on Build GRCh38
rs752207344 Chr.4: 56994627-56994627 on Build GRCh38
rs928881250 Chr.4: 56994997-56994997 on Build GRCh38
rs535480505 Chr.4: 56994990-56994990 on Build GRCh38
rs1022492816 Chr.4: 56995019-56995019 on Build GRCh38
rs776142668 Chr.4: 56994931-56994939 on Build GRCh38
rs773469780 Chr.4: 56994871-56994871 on Build GRCh38
rs926968230 Chr.4: 56994596-56994596 on Build GRCh38
rs751127947 Chr.4: 56994909-56994909 on Build GRCh38
rs774480932 Chr.4: 56994681-56994681 on Build GRCh38
rs78320117 Chr.4: 56994676-56994676 on Build GRCh38
rs765732690 Chr.4: 56994732-56994732 on Build GRCh38
rs938012550 Chr.4: 56994561-56994561 on Build GRCh38
rs763581646 Chr.4: 56994930-56994938 on Build GRCh38
rs201102254 Chr.4: 56994896-56994896 on Build GRCh38
rs756011094 Chr.4: 56994851-56994851 on Build GRCh38
rs569958063 Chr.4: 56994979-56994979 on Build GRCh38
rs772763389 Chr.4: 56994801-56994801 on Build GRCh38
rs569227791 Chr.4: 56994933-56994933 on Build GRCh38

View Full Product Details