Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134559719-134560266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JADE2
Entrez Gene ID
23338
Gene Name
jade family PHD finger 2
Gene Aliases
JADE-2, PHF15
UniGene
Hs.732155
Gene Chromosome Location
Chr.5: 134524154-134583230 on Build GRCh38
Transcripts
NM_001289984.1 NM_001289985.1 NM_001308143.1 NM_015288.5 XM_005271945.1 XM_005271946.1 XM_005271948.4 XM_011543291.2 XM_017009282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3138090 c.354G>A p.P118P Chr.5: 134559872-134559872 on Build GRCh38
5830022 c.449_450insC p.S151fs*4 Chr.5: 134559919-134559920 on Build GRCh38
5625681 c.363C>T p.T121T Chr.5: 134559881-134559881 on Build GRCh38
3138091 c.466G>A p.E156K Chr.5: 134559984-134559984 on Build GRCh38
5867290 c.370C>T p.P124S Chr.5: 134559840-134559840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748474174 Chr.5: 134559863-134559863 on Build GRCh38
rs759573430 Chr.5: 134559790-134559790 on Build GRCh38
rs895936505 Chr.5: 134559770-134559770 on Build GRCh38
rs1033246911 Chr.5: 134559843-134559843 on Build GRCh38
rs114147622 Chr.5: 134559957-134559957 on Build GRCh38
rs780381856 Chr.5: 134560054-134560054 on Build GRCh38
rs1000347068 Chr.5: 134560081-134560081 on Build GRCh38
rs1028913843 Chr.5: 134560035-134560035 on Build GRCh38
rs745910827 Chr.5: 134560061-134560061 on Build GRCh38
rs953419498 Chr.5: 134560048-134560048 on Build GRCh38
rs750057398 Chr.5: 134560002-134560002 on Build GRCh38
rs763856577 Chr.5: 134559973-134559973 on Build GRCh38
rs569107238 Chr.5: 134560069-134560069 on Build GRCh38
rs752390192 Chr.5: 134559765-134559765 on Build GRCh38
rs755616899 Chr.5: 134560006-134560006 on Build GRCh38
rs965224144 Chr.5: 134560075-134560075 on Build GRCh38
rs756720812 Chr.5: 134559831-134559831 on Build GRCh38
rs987509538 Chr.5: 134560053-134560053 on Build GRCh38
rs140822723 Chr.5: 134560025-134560025 on Build GRCh38
rs755902280 Chr.5: 134559776-134559776 on Build GRCh38
rs867252462 Chr.5: 134559980-134559980 on Build GRCh38
rs1033132367 Chr.5: 134560131-134560131 on Build GRCh38
rs926940619 Chr.5: 134560176-134560176 on Build GRCh38
rs987519504 Chr.5: 134559749-134559749 on Build GRCh38
rs777606989 Chr.5: 134559819-134559819 on Build GRCh38
rs761022325 Chr.5: 134559868-134559868 on Build GRCh38
rs778257053 Chr.5: 134559938-134559938 on Build GRCh38
rs368048328 Chr.5: 134559805-134559805 on Build GRCh38
rs762938739 Chr.5: 134559965-134559965 on Build GRCh38
rs755335433 Chr.5: 134560160-134560160 on Build GRCh38
rs751161699 Chr.5: 134559908-134559908 on Build GRCh38
rs759357300 Chr.5: 134559884-134559884 on Build GRCh38
rs747173757 Chr.5: 134559942-134559942 on Build GRCh38
rs199670348 Chr.5: 134559974-134559974 on Build GRCh38
rs931188811 Chr.5: 134560142-134560142 on Build GRCh38
rs200711171 Chr.5: 134559925-134559925 on Build GRCh38
rs762672702 Chr.5: 134559895-134559895 on Build GRCh38
rs35545286 Chr.5: 134559913-134559913 on Build GRCh38
rs780813914 Chr.5: 134559839-134559839 on Build GRCh38
rs777351343 Chr.5: 134559820-134559820 on Build GRCh38
rs777965831 Chr.5: 134560042-134560042 on Build GRCh38
rs767149887 Chr.5: 134560000-134560000 on Build GRCh38
rs115657900 Chr.5: 134559871-134559871 on Build GRCh38
rs528867392 Chr.5: 134560023-134560023 on Build GRCh38
rs764992378 Chr.5: 134559793-134559793 on Build GRCh38
rs965386365 Chr.5: 134560230-134560230 on Build GRCh38
rs775424432 Chr.5: 134559891-134559891 on Build GRCh38
rs748668343 Chr.5: 134559924-134559924 on Build GRCh38
rs747378372 Chr.5: 134560063-134560063 on Build GRCh38
rs1010864342 Chr.5: 134560172-134560172 on Build GRCh38
rs776817859 Chr.5: 134559872-134559872 on Build GRCh38
rs772577491 Chr.5: 134559865-134559865 on Build GRCh38
rs145325630 Chr.5: 134559928-134559928 on Build GRCh38
rs1020623344 Chr.5: 134560192-134560192 on Build GRCh38
rs937023107 Chr.5: 134560226-134560226 on Build GRCh38
rs749737763 Chr.5: 134559842-134559842 on Build GRCh38
rs1037075972 Chr.5: 134559996-134559996 on Build GRCh38
rs568103937 Chr.5: 134559993-134559993 on Build GRCh38
rs765115289 Chr.5: 134559885-134559885 on Build GRCh38
rs200309089 Chr.5: 134559961-134559961 on Build GRCh38
rs779505300 Chr.5: 134559919-134559919 on Build GRCh38
rs776746894 Chr.5: 134559952-134559952 on Build GRCh38
rs370946545 Chr.5: 134559780-134559780 on Build GRCh38
rs771221323 Chr.5: 134559947-134559947 on Build GRCh38
rs763946544 Chr.5: 134559899-134559899 on Build GRCh38
rs751232762 Chr.5: 134559821-134559821 on Build GRCh38
rs749909598 Chr.5: 134559914-134559914 on Build GRCh38
rs753713197 Chr.5: 134559781-134559781 on Build GRCh38
rs759020152 Chr.5: 134560039-134560039 on Build GRCh38
rs758215822 Chr.5: 134559815-134559815 on Build GRCh38
rs371625180 Chr.5: 134559814-134559814 on Build GRCh38
rs769247538 Chr.5: 134559856-134559856 on Build GRCh38
rs749170830 Chr.5: 134559902-134559902 on Build GRCh38
rs139883151 Chr.5: 134559915-134559915 on Build GRCh38
rs796539908 Chr.5: 134559748-134559748 on Build GRCh38
rs775404385 Chr.5: 134559963-134559963 on Build GRCh38

View Full Product Details