Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140694035-140694438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HARS2
Entrez Gene ID
23438
Gene Name
histidyl-tRNA synthetase 2, mitochondrial
Gene Aliases
HARSL, HARSR, HO3, PRLTS2
UniGene
Hs.432560
Gene Chromosome Location
Chr.5: 140691426-140699318 on Build GRCh38
Transcripts
NM_001278731.1 NM_001278732.1 NM_012208.3 XM_011537619.1 XM_011537620.1 XM_017009287.1 XM_017009288.1 XM_017009289.1 XM_017009290.1 XM_017009291.1 XM_017009292.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5804497 c.304-2A>T p.? Chr.5: 140694183-140694183 on Build GRCh38
4609123 c.352C>A p.L118M Chr.5: 140694233-140694233 on Build GRCh38
3381106 c.303+2T>C p.? Chr.5: 140694056-140694056 on Build GRCh38
6346870 c.399+1G>T p.? Chr.5: 140694281-140694281 on Build GRCh38
6283190 c.338G>A p.G113E Chr.5: 140694219-140694219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555954745 Chr.5: 140694405-140694405 on Build GRCh38
rs778535058 Chr.5: 140694237-140694237 on Build GRCh38
rs377730406 Chr.5: 140694100-140694100 on Build GRCh38
rs533472697 Chr.5: 140694266-140694266 on Build GRCh38
rs748677831 Chr.5: 140694140-140694140 on Build GRCh38
rs78426773 Chr.5: 140694088-140694088 on Build GRCh38
rs376860640 Chr.5: 140694223-140694223 on Build GRCh38
rs374827942 Chr.5: 140694328-140694328 on Build GRCh38
rs975734645 Chr.5: 140694307-140694307 on Build GRCh38
rs755600265 Chr.5: 140694105-140694105 on Build GRCh38
rs10515510 Chr.5: 140694160-140694160 on Build GRCh38
rs373905676 Chr.5: 140694189-140694189 on Build GRCh38
rs565008309 Chr.5: 140694330-140694330 on Build GRCh38
rs527673882 Chr.5: 140694351-140694351 on Build GRCh38
rs745560786 Chr.5: 140694156-140694156 on Build GRCh38
rs192714181 Chr.5: 140694399-140694399 on Build GRCh38
rs771782336 Chr.5: 140694242-140694242 on Build GRCh38
rs372656235 Chr.5: 140694293-140694293 on Build GRCh38
rs756692287 Chr.5: 140694230-140694230 on Build GRCh38
rs748722079 Chr.5: 140694228-140694228 on Build GRCh38
rs745511545 Chr.5: 140694240-140694240 on Build GRCh38
rs767966107 Chr.5: 140694301-140694301 on Build GRCh38
rs758738918 Chr.5: 140694219-140694219 on Build GRCh38
rs775326420 Chr.5: 140694161-140694161 on Build GRCh38
rs759280022 Chr.5: 140694066-140694066 on Build GRCh38
rs1001631468 Chr.5: 140694395-140694395 on Build GRCh38
rs755372694 Chr.5: 140694225-140694225 on Build GRCh38
rs759376945 Chr.5: 140694216-140694216 on Build GRCh38
rs151312766 Chr.5: 140694267-140694267 on Build GRCh38
rs763874405 Chr.5: 140694177-140694177 on Build GRCh38
rs140855017 Chr.5: 140694104-140694104 on Build GRCh38
rs941574922 Chr.5: 140694363-140694363 on Build GRCh38
rs746751794 Chr.5: 140694259-140694259 on Build GRCh38
rs751841748 Chr.5: 140694295-140694295 on Build GRCh38
rs752600766 Chr.5: 140694076-140694076 on Build GRCh38
rs760604782 Chr.5: 140694176-140694176 on Build GRCh38
rs935398039 Chr.5: 140694311-140694311 on Build GRCh38
rs369816856 Chr.5: 140694264-140694264 on Build GRCh38
rs922835626 Chr.5: 140694308-140694308 on Build GRCh38
rs983026856 Chr.5: 140694344-140694344 on Build GRCh38
rs6883035 Chr.5: 140694298-140694298 on Build GRCh38
rs750638675 Chr.5: 140694212-140694212 on Build GRCh38
rs373375372 Chr.5: 140694084-140694084 on Build GRCh38
rs757648883 Chr.5: 140694180-140694181 on Build GRCh38
rs368989873 Chr.5: 140694181-140694181 on Build GRCh38
rs753123211 Chr.5: 140694302-140694302 on Build GRCh38
rs775131788 Chr.5: 140694252-140694252 on Build GRCh38
rs766639750 Chr.5: 140694280-140694280 on Build GRCh38
rs781492646 Chr.5: 140694226-140694226 on Build GRCh38
rs768516014 Chr.5: 140694260-140694260 on Build GRCh38
rs773483704 Chr.5: 140694269-140694269 on Build GRCh38
rs773836159 Chr.5: 140694147-140694147 on Build GRCh38
rs758792128 Chr.5: 140694098-140694098 on Build GRCh38
rs764224258 Chr.5: 140694082-140694082 on Build GRCh38
rs145259405 Chr.5: 140694090-140694090 on Build GRCh38
rs767332898 Chr.5: 140694067-140694067 on Build GRCh38
rs145215203 Chr.5: 140694373-140694373 on Build GRCh38
rs916004467 Chr.5: 140694374-140694374 on Build GRCh38
rs777331269 Chr.5: 140694139-140694139 on Build GRCh38
rs763666118 Chr.5: 140694356-140694356 on Build GRCh38
rs778395061 Chr.5: 140694329-140694329 on Build GRCh38
rs143687204 Chr.5: 140694205-140694205 on Build GRCh38
rs768791952 Chr.5: 140694124-140694124 on Build GRCh38
rs760659695 Chr.5: 140694077-140694077 on Build GRCh38
rs184394978 Chr.5: 140694331-140694331 on Build GRCh38
rs187686304 Chr.5: 140694129-140694129 on Build GRCh38
rs774055964 Chr.5: 140694060-140694060 on Build GRCh38
rs1052487955 Chr.5: 140694343-140694343 on Build GRCh38
rs188829067 Chr.5: 140694355-140694355 on Build GRCh38
rs763261995 Chr.5: 140694270-140694270 on Build GRCh38
rs113407877 Chr.5: 140694342-140694342 on Build GRCh38
rs770499453 Chr.5: 140694146-140694146 on Build GRCh38

View Full Product Details